Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 13112823-13113340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPDZ
Entrez Gene ID
8777
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases
HYC2, MUPP1
UniGene
Hs.169378
Gene Chromosome Location
Chr.9: 13105296-13279687 on Build GRCh38
Transcripts
NM_001261406.1 NM_001261407.1 NM_003829.4 XM_005251622.4 XM_005251623.4 XM_006716885.3 XM_006716886.3 XM_006716887.3 XM_006716888.3 XM_006716889.3 XM_006716891.3 XM_017015252.1 XM_017015253.1 XM_017015254.1 XM_017015255.1 XM_017015256.1 XM_017015257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
276105 c.5505C>T p.V1835V Chr.9: 13113023-13113023 on Build GRCh38
1460398 c.5497A>C p.R1833R Chr.9: 13113031-13113031 on Build GRCh38
1105558 c.5506G>A p.E1836K Chr.9: 13113022-13113022 on Build GRCh38
6874324 c.5517+1G>A p.? Chr.9: 13113010-13113010 on Build GRCh38
1460397 c.5494A>C p.R1832R Chr.9: 13113031-13113031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756212652 Chr.9: 13113086-13113086 on Build GRCh38
rs569045295 Chr.9: 13112883-13112883 on Build GRCh38
rs760456120 Chr.9: 13112930-13112930 on Build GRCh38
rs886433908 Chr.9: 13113160-13113163 on Build GRCh38
rs752498486 Chr.9: 13113067-13113067 on Build GRCh38
rs879131328 Chr.9: 13113124-13113124 on Build GRCh38
rs750970272 Chr.9: 13113010-13113010 on Build GRCh38
rs750623791 Chr.9: 13113077-13113077 on Build GRCh38
rs566960858 Chr.9: 13113254-13113254 on Build GRCh38
rs767848483 Chr.9: 13112965-13112965 on Build GRCh38
rs145384556 Chr.9: 13112862-13112862 on Build GRCh38
rs371434505 Chr.9: 13113005-13113005 on Build GRCh38
rs532066583 Chr.9: 13113137-13113137 on Build GRCh38
rs749795326 Chr.9: 13113027-13113027 on Build GRCh38
rs922089567 Chr.9: 13113313-13113313 on Build GRCh38
rs956483620 Chr.9: 13113257-13113257 on Build GRCh38
rs1021985056 Chr.9: 13112938-13112938 on Build GRCh38
rs567988740 Chr.9: 13113186-13113186 on Build GRCh38
rs76297322 Chr.9: 13112871-13112871 on Build GRCh38
rs774867671 Chr.9: 13113041-13113041 on Build GRCh38
rs769199723 Chr.9: 13113028-13113028 on Build GRCh38
rs769289499 Chr.9: 13113092-13113092 on Build GRCh38
rs368364908 Chr.9: 13112975-13112975 on Build GRCh38
rs758152595 Chr.9: 13113070-13113070 on Build GRCh38
rs368576356 Chr.9: 13113000-13113000 on Build GRCh38
rs533719367 Chr.9: 13112870-13112870 on Build GRCh38
rs772005880 Chr.9: 13112912-13112912 on Build GRCh38
rs377025202 Chr.9: 13113002-13113002 on Build GRCh38
rs562388389 Chr.9: 13113150-13113150 on Build GRCh38
rs760192031 Chr.9: 13112998-13112998 on Build GRCh38
rs1056475827 Chr.9: 13113206-13113206 on Build GRCh38
rs895170834 Chr.9: 13113258-13113258 on Build GRCh38
rs753787253 Chr.9: 13113054-13113054 on Build GRCh38
rs910899576 Chr.9: 13113113-13113113 on Build GRCh38
rs369015690 Chr.9: 13113080-13113080 on Build GRCh38
rs547235788 Chr.9: 13113245-13113245 on Build GRCh38
rs779521959 Chr.9: 13113094-13113094 on Build GRCh38
rs930741134 Chr.9: 13112954-13112954 on Build GRCh38
rs1033765429 Chr.9: 13112957-13112957 on Build GRCh38
rs912058577 Chr.9: 13112928-13112928 on Build GRCh38
rs61753786 Chr.9: 13113011-13113011 on Build GRCh38
rs891614943 Chr.9: 13113182-13113182 on Build GRCh38
rs773024102 Chr.9: 13112929-13112929 on Build GRCh38
rs764519893 Chr.9: 13112972-13112972 on Build GRCh38
rs375992264 Chr.9: 13113057-13113057 on Build GRCh38
rs375001551 Chr.9: 13113023-13113023 on Build GRCh38
rs554246821 Chr.9: 13113033-13113033 on Build GRCh38
rs56002100 Chr.9: 13113255-13113255 on Build GRCh38
rs367604771 Chr.9: 13113035-13113035 on Build GRCh38
rs767219040 Chr.9: 13112982-13112982 on Build GRCh38
rs776416799 Chr.9: 13113022-13113022 on Build GRCh38
rs886464891 Chr.9: 13112980-13112980 on Build GRCh38
rs370721108 Chr.9: 13112976-13112976 on Build GRCh38
rs192589235 Chr.9: 13112909-13112909 on Build GRCh38
rs559001342 Chr.9: 13113271-13113271 on Build GRCh38
rs1040832328 Chr.9: 13113175-13113175 on Build GRCh38
rs776013591 Chr.9: 13112963-13112963 on Build GRCh38
rs368217290 Chr.9: 13113135-13113135 on Build GRCh38
rs1004789209 Chr.9: 13112987-13112987 on Build GRCh38
rs1001305639 Chr.9: 13112952-13112952 on Build GRCh38
rs552151635 Chr.9: 13113140-13113140 on Build GRCh38
rs973353635 Chr.9: 13113315-13113315 on Build GRCh38
rs141024279 Chr.9: 13112876-13112876 on Build GRCh38
rs74973826 Chr.9: 13113174-13113175 on Build GRCh38
rs374639569 Chr.9: 13112988-13112988 on Build GRCh38
rs373441968 Chr.9: 13113291-13113291 on Build GRCh38
rs780165815 Chr.9: 13113090-13113090 on Build GRCh38
rs943738633 Chr.9: 13113120-13113120 on Build GRCh38
rs144590212 Chr.9: 13112948-13112948 on Build GRCh38
rs199730853 Chr.9: 13113045-13113045 on Build GRCh38
rs943772904 Chr.9: 13112890-13112890 on Build GRCh38
rs533757186 Chr.9: 13113192-13113192 on Build GRCh38
rs765104820 Chr.9: 13113066-13113066 on Build GRCh38
rs544272098 Chr.9: 13113007-13113007 on Build GRCh38
rs1044904217 Chr.9: 13113246-13113246 on Build GRCh38
rs531195897 Chr.9: 13113169-13113169 on Build GRCh38
rs956396544 Chr.9: 13113043-13113043 on Build GRCh38
rs1019288872 Chr.9: 13113273-13113273 on Build GRCh38
rs560208273 Chr.9: 13113112-13113112 on Build GRCh38
rs967767927 Chr.9: 13113021-13113021 on Build GRCh38
rs763272827 Chr.9: 13112968-13112968 on Build GRCh38
rs548107981 Chr.9: 13113176-13113176 on Build GRCh38
rs749268450 Chr.9: 13113091-13113091 on Build GRCh38
rs201173952 Chr.9: 13113074-13113074 on Build GRCh38
rs760152518 Chr.9: 13113049-13113049 on Build GRCh38
rs765006530 Chr.9: 13113215-13113215 on Build GRCh38
rs199904060 Chr.9: 13113160-13113160 on Build GRCh38
rs765883510 Chr.9: 13113050-13113050 on Build GRCh38
rs1025044647 Chr.9: 13113287-13113287 on Build GRCh38
rs762202328 Chr.9: 13113043-13113043 on Build GRCh38
rs1042970639 Chr.9: 13113210-13113210 on Build GRCh38
rs979064571 Chr.9: 13112880-13112880 on Build GRCh38
rs1034061962 Chr.9: 13113256-13113256 on Build GRCh38

View Full Product Details