Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 174999677-175000171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2

SNP Detail

SNP ID Chromosome Location
rs1035990277 Chr.1: 175000074-175000074 on Build GRCh38
rs1002355854 Chr.1: 174999982-174999982 on Build GRCh38
rs747663581 Chr.1: 175000150-175000150 on Build GRCh38
rs921379735 Chr.1: 175000023-175000023 on Build GRCh38
rs111585397 Chr.1: 175000141-175000141 on Build GRCh38
rs574197445 Chr.1: 175000133-175000133 on Build GRCh38
rs576845016 Chr.1: 174999801-174999801 on Build GRCh38
rs199809264 Chr.1: 175000145-175000145 on Build GRCh38
rs546624993 Chr.1: 174999940-174999940 on Build GRCh38
rs536236832 Chr.1: 174999769-174999769 on Build GRCh38
rs1048784673 Chr.1: 175000085-175000085 on Build GRCh38
rs1046439 Chr.1: 175000135-175000135 on Build GRCh38
rs1017220063 Chr.1: 175000147-175000147 on Build GRCh38
rs571380880 Chr.1: 175000086-175000086 on Build GRCh38
rs943168144 Chr.1: 174999702-174999702 on Build GRCh38
rs944629242 Chr.1: 175000071-175000071 on Build GRCh38
rs915049676 Chr.1: 175000082-175000082 on Build GRCh38
rs996101811 Chr.1: 174999740-174999740 on Build GRCh38
rs752889549 Chr.1: 175000136-175000136 on Build GRCh38
rs569377458 Chr.1: 174999721-174999721 on Build GRCh38
rs988569205 Chr.1: 174999839-174999839 on Build GRCh38
rs994544957 Chr.1: 175000078-175000078 on Build GRCh38
rs201546478 Chr.1: 174999766-174999769 on Build GRCh38
rs925775553 Chr.1: 174999909-174999909 on Build GRCh38
rs998813145 Chr.1: 174999895-174999895 on Build GRCh38
rs931838694 Chr.1: 174999927-174999927 on Build GRCh38
rs184014500 Chr.1: 174999771-174999771 on Build GRCh38
rs962119545 Chr.1: 174999783-174999783 on Build GRCh38
rs771406293 Chr.1: 175000132-175000132 on Build GRCh38
rs924612845 Chr.1: 174999964-174999964 on Build GRCh38
rs528821623 Chr.1: 174999950-174999950 on Build GRCh38
rs1017505418 Chr.1: 175000044-175000044 on Build GRCh38
rs571403717 Chr.1: 174999716-174999739 on Build GRCh38
rs890119532 Chr.1: 174999761-174999761 on Build GRCh38
rs987353185 Chr.1: 174999879-174999879 on Build GRCh38
rs765660881 Chr.1: 174999732-174999732 on Build GRCh38
rs879084400 Chr.1: 174999900-174999900 on Build GRCh38
rs565522582 Chr.1: 174999877-174999877 on Build GRCh38
rs764512900 Chr.1: 174999845-174999845 on Build GRCh38
rs1028247144 Chr.1: 174999802-174999802 on Build GRCh38
rs948978871 Chr.1: 175000109-175000109 on Build GRCh38
rs188762152 Chr.1: 174999807-174999807 on Build GRCh38
rs993662280 Chr.1: 175000001-175000001 on Build GRCh38
rs548602563 Chr.1: 174999965-174999965 on Build GRCh38
rs960909161 Chr.1: 175000115-175000115 on Build GRCh38
rs969520096 Chr.1: 174999967-174999967 on Build GRCh38
rs527888806 Chr.1: 174999931-174999931 on Build GRCh38
rs1042861093 Chr.1: 174999904-174999904 on Build GRCh38
rs960681425 Chr.1: 174999836-174999836 on Build GRCh38
rs966533030 Chr.1: 175000090-175000090 on Build GRCh38
rs971796171 Chr.1: 174999838-174999838 on Build GRCh38
rs968119662 Chr.1: 175000007-175000007 on Build GRCh38
rs756674983 Chr.1: 175000148-175000148 on Build GRCh38
rs1035162570 Chr.1: 175000113-175000113 on Build GRCh38
rs778151297 Chr.1: 175000138-175000138 on Build GRCh38
rs1035216613 Chr.1: 174999987-174999987 on Build GRCh38
rs946801962 Chr.1: 174999863-174999863 on Build GRCh38
rs1013066102 Chr.1: 174999823-174999823 on Build GRCh38
rs994999233 Chr.1: 174999958-174999958 on Build GRCh38
rs533752524 Chr.1: 175000106-175000106 on Build GRCh38
rs755859164 Chr.1: 175000066-175000066 on Build GRCh38
rs554281043 Chr.1: 175000119-175000119 on Build GRCh38
rs945880339 Chr.1: 174999705-174999705 on Build GRCh38
rs985928464 Chr.1: 175000153-175000153 on Build GRCh38
rs376540752 Chr.1: 174999943-174999943 on Build GRCh38
rs763469564 Chr.1: 174999785-174999785 on Build GRCh38
rs551275986 Chr.1: 175000076-175000076 on Build GRCh38
rs71645225 Chr.1: 174999758-174999758 on Build GRCh38
rs904409360 Chr.1: 174999718-174999718 on Build GRCh38
rs988128180 Chr.1: 174999991-174999991 on Build GRCh38
rs146318167 Chr.1: 174999809-174999809 on Build GRCh38
rs1035909458 Chr.1: 174999814-174999814 on Build GRCh38
rs780536365 Chr.1: 175000149-175000149 on Build GRCh38
rs1027444297 Chr.1: 174999960-174999960 on Build GRCh38
rs773613992 Chr.1: 175000102-175000102 on Build GRCh38
rs752330128 Chr.1: 174999843-174999843 on Build GRCh38
rs180782165 Chr.1: 175000146-175000146 on Build GRCh38
rs913898531 Chr.1: 174999992-174999992 on Build GRCh38
rs556625983 Chr.1: 175000139-175000139 on Build GRCh38
rs1050271244 Chr.1: 175000062-175000062 on Build GRCh38
rs1016027683 Chr.1: 174999829-174999829 on Build GRCh38
rs910446888 Chr.1: 174999894-174999894 on Build GRCh38
rs1028390241 Chr.1: 175000144-175000144 on Build GRCh38
rs954345812 Chr.1: 174999866-174999866 on Build GRCh38
rs931787746 Chr.1: 175000029-175000029 on Build GRCh38
rs1043298384 Chr.1: 174999709-174999709 on Build GRCh38
rs967703671 Chr.1: 174999890-174999890 on Build GRCh38
rs377745745 Chr.1: 174999988-174999988 on Build GRCh38
rs1008491070 Chr.1: 175000029-175000030 on Build GRCh38
rs1036452143 Chr.1: 174999944-174999944 on Build GRCh38
rs767510080 Chr.1: 175000131-175000133 on Build GRCh38
rs758482512 Chr.1: 175000137-175000137 on Build GRCh38
rs771514371 Chr.1: 175000152-175000152 on Build GRCh38
rs753544362 Chr.1: 175000132-175000132 on Build GRCh38
rs112052593 Chr.1: 174999949-174999949 on Build GRCh38
rs1013109061 Chr.1: 174999990-174999990 on Build GRCh38
rs928252934 Chr.1: 174999822-174999822 on Build GRCh38
rs115063638 Chr.1: 175000012-175000012 on Build GRCh38
rs781653545 Chr.1: 175000142-175000142 on Build GRCh38
rs904974053 Chr.1: 175000098-175000098 on Build GRCh38
rs938676835 Chr.1: 175000099-175000099 on Build GRCh38

View Full Product Details