Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 88994863-88995378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf47
Entrez Gene ID
286223
Gene Name
chromosome 9 open reading frame 47
Gene Aliases
C9orf108, bA791O21.3
UniGene
Hs.585118
Gene Chromosome Location
Chr.9: 88990863-88996142 on Build GRCh38
Transcripts
NM_001001938.3 NM_001142413.1 NM_005226.3

Gene Symbol
S1PR3
Entrez Gene ID
1903
Gene Name
sphingosine-1-phosphate receptor 3
Gene Aliases
EDG-3, EDG3, LPB3, S1P3
UniGene
Hs.585118
Gene Chromosome Location
Chr.9: 88991409-89005155 on Build GRCh38
Transcripts
NM_001001938.3 NM_001142413.1 NM_005226.3

SNP Detail

SNP ID Chromosome Location
rs1027010353 Chr.9: 88995118-88995118 on Build GRCh38
rs574606999 Chr.9: 88995012-88995012 on Build GRCh38
rs1054079509 Chr.9: 88995324-88995324 on Build GRCh38
rs561075711 Chr.9: 88995344-88995344 on Build GRCh38
rs559000323 Chr.9: 88994947-88994947 on Build GRCh38
rs980607549 Chr.9: 88994982-88994982 on Build GRCh38
rs148717819 Chr.9: 88995177-88995177 on Build GRCh38
rs34199294 Chr.9: 88994925-88994926 on Build GRCh38
rs752758325 Chr.9: 88995209-88995209 on Build GRCh38
rs923346987 Chr.9: 88995193-88995193 on Build GRCh38
rs974825556 Chr.9: 88995187-88995187 on Build GRCh38
rs901684924 Chr.9: 88994944-88994944 on Build GRCh38
rs547365171 Chr.9: 88995293-88995293 on Build GRCh38
rs1049470282 Chr.9: 88995080-88995080 on Build GRCh38
rs377209772 Chr.9: 88994924-88994924 on Build GRCh38
rs528291833 Chr.9: 88995262-88995262 on Build GRCh38
rs374722323 Chr.9: 88995100-88995100 on Build GRCh38
rs1042291684 Chr.9: 88995042-88995042 on Build GRCh38
rs973007640 Chr.9: 88994917-88994917 on Build GRCh38
rs185785834 Chr.9: 88995043-88995043 on Build GRCh38
rs374145344 Chr.9: 88995123-88995123 on Build GRCh38
rs936341350 Chr.9: 88995348-88995348 on Build GRCh38
rs750936255 Chr.9: 88994991-88994991 on Build GRCh38
rs554613478 Chr.9: 88994971-88994971 on Build GRCh38
rs933100507 Chr.9: 88994945-88994945 on Build GRCh38
rs1034444419 Chr.9: 88995240-88995240 on Build GRCh38
rs971578467 Chr.9: 88995046-88995046 on Build GRCh38
rs561733442 Chr.9: 88995273-88995273 on Build GRCh38
rs1041236418 Chr.9: 88995130-88995130 on Build GRCh38
rs917768772 Chr.9: 88995272-88995273 on Build GRCh38
rs1032026071 Chr.9: 88995005-88995005 on Build GRCh38
rs141825917 Chr.9: 88995289-88995289 on Build GRCh38
rs182430974 Chr.9: 88995034-88995034 on Build GRCh38
rs142250356 Chr.9: 88995195-88995195 on Build GRCh38
rs535270023 Chr.9: 88994948-88994948 on Build GRCh38
rs755199190 Chr.9: 88995088-88995088 on Build GRCh38
rs1039109409 Chr.9: 88994942-88994942 on Build GRCh38
rs937831403 Chr.9: 88995011-88995011 on Build GRCh38
rs985421979 Chr.9: 88995127-88995127 on Build GRCh38
rs978430709 Chr.9: 88995281-88995281 on Build GRCh38
rs1027209870 Chr.9: 88995193-88995193 on Build GRCh38
rs542464907 Chr.9: 88995264-88995264 on Build GRCh38
rs952731227 Chr.9: 88994938-88994938 on Build GRCh38
rs964672899 Chr.9: 88995183-88995183 on Build GRCh38
rs961965387 Chr.9: 88994890-88994890 on Build GRCh38
rs1042205810 Chr.9: 88995261-88995261 on Build GRCh38
rs1027423738 Chr.9: 88994927-88994927 on Build GRCh38
rs917750346 Chr.9: 88995029-88995029 on Build GRCh38
rs767111699 Chr.9: 88995202-88995202 on Build GRCh38
rs370740644 Chr.9: 88994904-88994904 on Build GRCh38
rs897303761 Chr.9: 88995154-88995154 on Build GRCh38
rs12336695 Chr.9: 88995208-88995208 on Build GRCh38

View Full Product Details