Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97460356-97460852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD7
Entrez Gene ID
23424
Gene Name
tudor domain containing 7
Gene Aliases
CATC4, PCTAIRE2BP, TRAP
UniGene
Hs.193842
Gene Chromosome Location
Chr.9: 97412020-97496125 on Build GRCh38
Transcripts
NM_001302884.1 NM_014290.2 XM_006717026.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1464308 c.1374G>T p.M458I Chr.9: 97460696-97460696 on Build GRCh38
4774711 c.1049T>G p.V350G Chr.9: 97460371-97460371 on Build GRCh38
6113979 c.1264G>T p.G422C Chr.9: 97460586-97460586 on Build GRCh38
4464603 c.1341C>T p.T447T Chr.9: 97460663-97460663 on Build GRCh38
2793026 c.1136A>C p.K379T Chr.9: 97460458-97460458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760073934 Chr.9: 97460703-97460703 on Build GRCh38
rs913246549 Chr.9: 97460550-97460550 on Build GRCh38
rs747766997 Chr.9: 97460543-97460543 on Build GRCh38
rs767958212 Chr.9: 97460705-97460705 on Build GRCh38
rs145545604 Chr.9: 97460557-97460557 on Build GRCh38
rs376371748 Chr.9: 97460621-97460621 on Build GRCh38
rs530610768 Chr.9: 97460535-97460535 on Build GRCh38
rs778768321 Chr.9: 97460782-97460782 on Build GRCh38
rs779276379 Chr.9: 97460386-97460386 on Build GRCh38
rs771147038 Chr.9: 97460449-97460449 on Build GRCh38
rs186876252 Chr.9: 97460627-97460627 on Build GRCh38
rs887898280 Chr.9: 97460511-97460511 on Build GRCh38
rs752270932 Chr.9: 97460710-97460710 on Build GRCh38
rs146047308 Chr.9: 97460675-97460675 on Build GRCh38
rs772576151 Chr.9: 97460399-97460399 on Build GRCh38
rs17852595 Chr.9: 97460689-97460689 on Build GRCh38
rs372005786 Chr.9: 97460379-97460379 on Build GRCh38
rs762022926 Chr.9: 97460579-97460579 on Build GRCh38
rs772101100 Chr.9: 97460480-97460480 on Build GRCh38
rs749145383 Chr.9: 97460554-97460554 on Build GRCh38
rs762726176 Chr.9: 97460712-97460724 on Build GRCh38
rs780574930 Chr.9: 97460526-97460526 on Build GRCh38
rs755645925 Chr.9: 97460753-97460753 on Build GRCh38
rs374773107 Chr.9: 97460408-97460408 on Build GRCh38
rs768158623 Chr.9: 97460407-97460407 on Build GRCh38
rs755443044 Chr.9: 97460616-97460616 on Build GRCh38
rs1043175351 Chr.9: 97460378-97460378 on Build GRCh38
rs367568709 Chr.9: 97460435-97460435 on Build GRCh38
rs772012019 Chr.9: 97460812-97460812 on Build GRCh38
rs552015878 Chr.9: 97460555-97460555 on Build GRCh38
rs778589649 Chr.9: 97460647-97460647 on Build GRCh38
rs746318646 Chr.9: 97460388-97460388 on Build GRCh38
rs143511562 Chr.9: 97460663-97460663 on Build GRCh38
rs747498675 Chr.9: 97460532-97460532 on Build GRCh38
rs775411557 Chr.9: 97460467-97460467 on Build GRCh38
rs150323833 Chr.9: 97460604-97460604 on Build GRCh38
rs758720057 Chr.9: 97460587-97460587 on Build GRCh38
rs758877384 Chr.9: 97460513-97460513 on Build GRCh38
rs113226995 Chr.9: 97460778-97460778 on Build GRCh38
rs751933537 Chr.9: 97460702-97460702 on Build GRCh38
rs369891285 Chr.9: 97460551-97460551 on Build GRCh38
rs779876593 Chr.9: 97460481-97460481 on Build GRCh38
rs765628635 Chr.9: 97460451-97460451 on Build GRCh38
rs763509024 Chr.9: 97460728-97460731 on Build GRCh38
rs752046246 Chr.9: 97460605-97460605 on Build GRCh38
rs985346487 Chr.9: 97460490-97460490 on Build GRCh38
rs745710762 Chr.9: 97460788-97460788 on Build GRCh38
rs752225106 Chr.9: 97460737-97460737 on Build GRCh38
rs765522687 Chr.9: 97460499-97460499 on Build GRCh38
rs199686400 Chr.9: 97460577-97460577 on Build GRCh38
rs780748474 Chr.9: 97460617-97460617 on Build GRCh38
rs960352420 Chr.9: 97460402-97460402 on Build GRCh38
rs753486062 Chr.9: 97460781-97460781 on Build GRCh38
rs372232770 Chr.9: 97460497-97460497 on Build GRCh38
rs943991492 Chr.9: 97460530-97460530 on Build GRCh38
rs780579998 Chr.9: 97460538-97460538 on Build GRCh38
rs763172476 Chr.9: 97460676-97460676 on Build GRCh38
rs961564135 Chr.9: 97460607-97460607 on Build GRCh38
rs760849386 Chr.9: 97460471-97460471 on Build GRCh38
rs961088321 Chr.9: 97460424-97460424 on Build GRCh38
rs766673237 Chr.9: 97460598-97460598 on Build GRCh38
rs979573593 Chr.9: 97460828-97460828 on Build GRCh38
rs764179156 Chr.9: 97460571-97460571 on Build GRCh38
rs777608846 Chr.9: 97460630-97460630 on Build GRCh38
rs746575726 Chr.9: 97460403-97460403 on Build GRCh38
rs776583414 Chr.9: 97460574-97460574 on Build GRCh38
rs754045537 Chr.9: 97460487-97460487 on Build GRCh38
rs1026781684 Chr.9: 97460473-97460473 on Build GRCh38
rs965642123 Chr.9: 97460803-97460803 on Build GRCh38
rs769637406 Chr.9: 97460544-97460544 on Build GRCh38
rs768091013 Chr.9: 97460804-97460804 on Build GRCh38
rs757605890 Chr.9: 97460381-97460381 on Build GRCh38
rs183136059 Chr.9: 97460564-97460564 on Build GRCh38
rs770802468 Chr.9: 97460643-97460643 on Build GRCh38
rs912468692 Chr.9: 97460523-97460523 on Build GRCh38
rs566332281 Chr.9: 97460586-97460586 on Build GRCh38
rs373534828 Chr.9: 97460658-97460658 on Build GRCh38
rs773639105 Chr.9: 97460400-97460400 on Build GRCh38
rs1001754965 Chr.9: 97460383-97460383 on Build GRCh38
rs754458500 Chr.9: 97460534-97460534 on Build GRCh38
rs747777898 Chr.9: 97460427-97460427 on Build GRCh38
rs748981064 Chr.9: 97460640-97460640 on Build GRCh38
rs761917089 Chr.9: 97460672-97460672 on Build GRCh38
rs137960660 Chr.9: 97460659-97460659 on Build GRCh38
rs773127507 Chr.9: 97460547-97460547 on Build GRCh38
rs112811088 Chr.9: 97460583-97460583 on Build GRCh38
rs990524723 Chr.9: 97460710-97460710 on Build GRCh38
rs377406077 Chr.9: 97460688-97460688 on Build GRCh38
rs138152263 Chr.9: 97460815-97460815 on Build GRCh38

View Full Product Details