Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101540558-101541060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF20
Entrez Gene ID
56254
Gene Name
ring finger protein 20
Gene Aliases
BRE1, BRE1A, hBRE1
UniGene
Hs.729085
Gene Chromosome Location
Chr.9: 101533849-101563344 on Build GRCh38
Transcripts
NM_019592.6 XM_011518862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979726 c.425G>A p.R142H Chr.9: 101540617-101540617 on Build GRCh38
4909458 c.563T>C p.I188T Chr.9: 101540910-101540910 on Build GRCh38
1103365 c.436_437insGA p.G149fs*42 Chr.9: 101540628-101540629 on Build GRCh38
3952133 c.463G>A p.A155T Chr.9: 101540810-101540810 on Build GRCh38
3952132 c.382G>A p.V128I Chr.9: 101540574-101540574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043844970 Chr.9: 101540582-101540582 on Build GRCh38
rs1055728697 Chr.9: 101540715-101540715 on Build GRCh38
rs370469701 Chr.9: 101540919-101540919 on Build GRCh38
rs768212285 Chr.9: 101540616-101540616 on Build GRCh38
rs759536223 Chr.9: 101540657-101540657 on Build GRCh38
rs750591860 Chr.9: 101540671-101540671 on Build GRCh38
rs780693484 Chr.9: 101540628-101540628 on Build GRCh38
rs777140211 Chr.9: 101540921-101540921 on Build GRCh38
rs748659375 Chr.9: 101540776-101540777 on Build GRCh38
rs745505185 Chr.9: 101541016-101541016 on Build GRCh38
rs146790041 Chr.9: 101540892-101540892 on Build GRCh38
rs1016680910 Chr.9: 101540732-101540732 on Build GRCh38
rs747590185 Chr.9: 101540635-101540635 on Build GRCh38
rs145275546 Chr.9: 101540957-101540957 on Build GRCh38
rs377247805 Chr.9: 101540891-101540891 on Build GRCh38
rs748876328 Chr.9: 101540767-101540767 on Build GRCh38
rs774289110 Chr.9: 101540787-101540787 on Build GRCh38
rs769468033 Chr.9: 101540636-101540636 on Build GRCh38
rs745701386 Chr.9: 101540944-101540944 on Build GRCh38
rs778672639 Chr.9: 101540925-101540925 on Build GRCh38
rs139560201 Chr.9: 101540895-101540895 on Build GRCh38
rs771973871 Chr.9: 101540955-101540955 on Build GRCh38
rs748762561 Chr.9: 101540922-101540922 on Build GRCh38
rs759342177 Chr.9: 101540810-101540810 on Build GRCh38
rs551561548 Chr.9: 101540678-101540678 on Build GRCh38
rs753306889 Chr.9: 101541002-101541002 on Build GRCh38
rs780279559 Chr.9: 101540673-101540673 on Build GRCh38
rs576092601 Chr.9: 101540999-101540999 on Build GRCh38
rs373452971 Chr.9: 101540660-101540660 on Build GRCh38
rs200797084 Chr.9: 101540967-101540967 on Build GRCh38
rs901859035 Chr.9: 101540988-101540988 on Build GRCh38
rs374568065 Chr.9: 101540985-101540985 on Build GRCh38
rs762036789 Chr.9: 101540875-101540875 on Build GRCh38
rs779556391 Chr.9: 101540759-101540760 on Build GRCh38
rs769903491 Chr.9: 101540598-101540598 on Build GRCh38
rs560105998 Chr.9: 101540958-101540958 on Build GRCh38
rs766540553 Chr.9: 101540979-101540979 on Build GRCh38
rs373105826 Chr.9: 101540845-101540845 on Build GRCh38
rs756764813 Chr.9: 101541004-101541004 on Build GRCh38
rs1021031299 Chr.9: 101540749-101540767 on Build GRCh38
rs753179127 Chr.9: 101540899-101540899 on Build GRCh38
rs534210601 Chr.9: 101540722-101540722 on Build GRCh38
rs773488421 Chr.9: 101540782-101540782 on Build GRCh38
rs753267177 Chr.9: 101540996-101540996 on Build GRCh38
rs769154201 Chr.9: 101540755-101540755 on Build GRCh38
rs554493254 Chr.9: 101540852-101540852 on Build GRCh38
rs763041367 Chr.9: 101540968-101540968 on Build GRCh38
rs758143655 Chr.9: 101540641-101540641 on Build GRCh38
rs7037267 Chr.9: 101540676-101540676 on Build GRCh38
rs116409222 Chr.9: 101541018-101541018 on Build GRCh38
rs992437192 Chr.9: 101540803-101540803 on Build GRCh38
rs376380494 Chr.9: 101540672-101540672 on Build GRCh38
rs200632178 Chr.9: 101540991-101540991 on Build GRCh38
rs747687487 Chr.9: 101540681-101540681 on Build GRCh38
rs772459590 Chr.9: 101540781-101540782 on Build GRCh38
rs542335592 Chr.9: 101540719-101540719 on Build GRCh38
rs192822144 Chr.9: 101540648-101540648 on Build GRCh38
rs960628674 Chr.9: 101540785-101540785 on Build GRCh38
rs777528551 Chr.9: 101540758-101540758 on Build GRCh38
rs61739680 Chr.9: 101540923-101540923 on Build GRCh38
rs756580481 Chr.9: 101540997-101540997 on Build GRCh38
rs772693155 Chr.9: 101540637-101540637 on Build GRCh38
rs753997642 Chr.9: 101540667-101540667 on Build GRCh38
rs766565572 Chr.9: 101540888-101540888 on Build GRCh38
rs962429847 Chr.9: 101540747-101540747 on Build GRCh38
rs761932485 Chr.9: 101540670-101540670 on Build GRCh38
rs916833534 Chr.9: 101540718-101540718 on Build GRCh38
rs763091222 Chr.9: 101540887-101540887 on Build GRCh38
rs567126384 Chr.9: 101540882-101540882 on Build GRCh38
rs759501625 Chr.9: 101540930-101540930 on Build GRCh38
rs778760631 Chr.9: 101541005-101541005 on Build GRCh38
rs199855488 Chr.9: 101540794-101540794 on Build GRCh38

View Full Product Details