Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105364470-105364975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A1
Entrez Gene ID
23446
Gene Name
solute carrier family 44 member 1
Gene Aliases
CD92, CDW92, CHTL1, CTL1
UniGene
Hs.573495
Gene Chromosome Location
Chr.9: 105244608-105438504 on Build GRCh38
Transcripts
NM_001286730.1 NM_080546.4 XM_005251855.4 XM_006717027.3 XM_006717028.3 XM_006717029.2 XM_017014559.1 XM_017014560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669233 c.1117G>T p.V373L Chr.9: 105364584-105364584 on Build GRCh38
4593767 c.1139C>T p.P380L Chr.9: 105364606-105364606 on Build GRCh38
1459134 c.1139C>A p.P380H Chr.9: 105364606-105364606 on Build GRCh38
3395483 c.1114T>G p.F372V Chr.9: 105364581-105364581 on Build GRCh38
3902749 c.1104T>C p.N368N Chr.9: 105364571-105364571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775034722 Chr.9: 105364543-105364543 on Build GRCh38
rs769514964 Chr.9: 105364711-105364711 on Build GRCh38
rs370728577 Chr.9: 105364531-105364531 on Build GRCh38
rs764333862 Chr.9: 105364764-105364764 on Build GRCh38
rs754326586 Chr.9: 105364768-105364768 on Build GRCh38
rs776591327 Chr.9: 105364517-105364517 on Build GRCh38
rs532425035 Chr.9: 105364895-105364895 on Build GRCh38
rs983590752 Chr.9: 105364680-105364680 on Build GRCh38
rs931333155 Chr.9: 105364718-105364718 on Build GRCh38
rs147396113 Chr.9: 105364665-105364665 on Build GRCh38
rs950914576 Chr.9: 105364640-105364640 on Build GRCh38
rs561216950 Chr.9: 105364749-105364749 on Build GRCh38
rs553788572 Chr.9: 105364930-105364930 on Build GRCh38
rs900507926 Chr.9: 105364940-105364940 on Build GRCh38
rs564943012 Chr.9: 105364862-105364862 on Build GRCh38
rs1020689050 Chr.9: 105364524-105364526 on Build GRCh38
rs1000902417 Chr.9: 105364916-105364916 on Build GRCh38
rs540389491 Chr.9: 105364791-105364791 on Build GRCh38
rs745465281 Chr.9: 105364617-105364617 on Build GRCh38
rs964990196 Chr.9: 105364556-105364556 on Build GRCh38
rs138719984 Chr.9: 105364772-105364772 on Build GRCh38
rs538745406 Chr.9: 105364494-105364494 on Build GRCh38
rs1055353256 Chr.9: 105364903-105364903 on Build GRCh38
rs768507199 Chr.9: 105364508-105364508 on Build GRCh38
rs762645487 Chr.9: 105364829-105364829 on Build GRCh38
rs767667699 Chr.9: 105364744-105364744 on Build GRCh38
rs761573641 Chr.9: 105364526-105364526 on Build GRCh38
rs776538234 Chr.9: 105364675-105364675 on Build GRCh38
rs748152049 Chr.9: 105364694-105364694 on Build GRCh38
rs997886082 Chr.9: 105364946-105364946 on Build GRCh38
rs757688984 Chr.9: 105364594-105364594 on Build GRCh38
rs542681852 Chr.9: 105364670-105364670 on Build GRCh38
rs957676947 Chr.9: 105364498-105364498 on Build GRCh38
rs763190255 Chr.9: 105364727-105364728 on Build GRCh38
rs908990838 Chr.9: 105364690-105364690 on Build GRCh38
rs771294682 Chr.9: 105364496-105364496 on Build GRCh38
rs753610123 Chr.9: 105364678-105364682 on Build GRCh38
rs377413388 Chr.9: 105364881-105364881 on Build GRCh38
rs759759019 Chr.9: 105364742-105364742 on Build GRCh38
rs1018055909 Chr.9: 105364939-105364939 on Build GRCh38
rs764672978 Chr.9: 105364589-105364589 on Build GRCh38
rs903969253 Chr.9: 105364915-105364915 on Build GRCh38
rs373011030 Chr.9: 105364840-105364840 on Build GRCh38
rs1007619453 Chr.9: 105364866-105364866 on Build GRCh38
rs754449376 Chr.9: 105364590-105364590 on Build GRCh38
rs1030964746 Chr.9: 105364600-105364600 on Build GRCh38
rs756608403 Chr.9: 105364576-105364576 on Build GRCh38
rs765076434 Chr.9: 105364528-105364528 on Build GRCh38
rs773927594 Chr.9: 105364904-105364904 on Build GRCh38
rs1042855602 Chr.9: 105364911-105364911 on Build GRCh38
rs974664793 Chr.9: 105364557-105364557 on Build GRCh38
rs1033765759 Chr.9: 105364497-105364497 on Build GRCh38
rs894048544 Chr.9: 105364947-105364947 on Build GRCh38
rs758006393 Chr.9: 105364620-105364620 on Build GRCh38
rs575304981 Chr.9: 105364569-105364569 on Build GRCh38
rs779421658 Chr.9: 105364610-105364610 on Build GRCh38
rs763709911 Chr.9: 105364863-105364863 on Build GRCh38
rs779575730 Chr.9: 105364624-105364624 on Build GRCh38
rs773006685 Chr.9: 105364726-105364726 on Build GRCh38
rs921279110 Chr.9: 105364560-105364560 on Build GRCh38
rs979805794 Chr.9: 105364763-105364763 on Build GRCh38

View Full Product Details