Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108879952-108880446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
UniGene
Hs.494738
Gene Chromosome Location
Chr.9: 108867520-108934332 on Build GRCh38
Transcripts
NM_001318360.1 NM_003640.4 XM_011519136.1 XM_011519137.1 XM_017015238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6684735 c.3363G>A p.M1121I Chr.9: 108880149-108880149 on Build GRCh38
3902862 c.3348C>T p.A1116A Chr.9: 108880164-108880164 on Build GRCh38
6476998 c.3368T>G p.F1123C Chr.9: 108880144-108880144 on Build GRCh38
1103678 c.3397C>T p.R1133C Chr.9: 108880115-108880115 on Build GRCh38
5808114 c.3442G>A p.A1148T Chr.9: 108880070-108880070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576315331 Chr.9: 108880040-108880040 on Build GRCh38
rs914289496 Chr.9: 108880326-108880326 on Build GRCh38
rs750980956 Chr.9: 108880183-108880183 on Build GRCh38
rs139054753 Chr.9: 108879976-108879976 on Build GRCh38
rs1002307742 Chr.9: 108880294-108880294 on Build GRCh38
rs764894465 Chr.9: 108880017-108880017 on Build GRCh38
rs202080366 Chr.9: 108880095-108880095 on Build GRCh38
rs572321768 Chr.9: 108880270-108880270 on Build GRCh38
rs767780798 Chr.9: 108880021-108880021 on Build GRCh38
rs1035572109 Chr.9: 108880187-108880187 on Build GRCh38
rs779392021 Chr.9: 108880041-108880041 on Build GRCh38
rs779796058 Chr.9: 108880071-108880071 on Build GRCh38
rs747077309 Chr.9: 108880407-108880407 on Build GRCh38
rs551705504 Chr.9: 108880416-108880416 on Build GRCh38
rs12342757 Chr.9: 108880422-108880422 on Build GRCh38
rs768015419 Chr.9: 108880088-108880088 on Build GRCh38
rs746597014 Chr.9: 108880213-108880213 on Build GRCh38
rs752925874 Chr.9: 108880024-108880024 on Build GRCh38
rs769833448 Chr.9: 108880109-108880109 on Build GRCh38
rs1012394977 Chr.9: 108880195-108880195 on Build GRCh38
rs562128767 Chr.9: 108880359-108880359 on Build GRCh38
rs952030874 Chr.9: 108879989-108879989 on Build GRCh38
rs77154805 Chr.9: 108880218-108880218 on Build GRCh38
rs931836170 Chr.9: 108880074-108880074 on Build GRCh38
rs7874280 Chr.9: 108880255-108880255 on Build GRCh38
rs1040572432 Chr.9: 108880031-108880031 on Build GRCh38
rs761843947 Chr.9: 108880014-108880014 on Build GRCh38
rs565686965 Chr.9: 108880377-108880377 on Build GRCh38
rs148372329 Chr.9: 108880408-108880408 on Build GRCh38
rs1056702193 Chr.9: 108880385-108880385 on Build GRCh38
rs555598184 Chr.9: 108880193-108880193 on Build GRCh38
rs41278353 Chr.9: 108880291-108880291 on Build GRCh38
rs776369407 Chr.9: 108880004-108880004 on Build GRCh38
rs560506429 Chr.9: 108880069-108880069 on Build GRCh38
rs779927751 Chr.9: 108880215-108880216 on Build GRCh38
rs753992200 Chr.9: 108880137-108880137 on Build GRCh38
rs547151441 Chr.9: 108880375-108880375 on Build GRCh38
rs201862301 Chr.9: 108880020-108880020 on Build GRCh38
rs771838754 Chr.9: 108880058-108880058 on Build GRCh38
rs769748960 Chr.9: 108880127-108880130 on Build GRCh38
rs746303238 Chr.9: 108880056-108880056 on Build GRCh38
rs780600094 Chr.9: 108880198-108880198 on Build GRCh38
rs377679012 Chr.9: 108880103-108880103 on Build GRCh38
rs373328501 Chr.9: 108880001-108880001 on Build GRCh38
rs945779566 Chr.9: 108880331-108880331 on Build GRCh38
rs779223676 Chr.9: 108880258-108880258 on Build GRCh38
rs143580972 Chr.9: 108880114-108880114 on Build GRCh38
rs895428158 Chr.9: 108880398-108880398 on Build GRCh38
rs766875245 Chr.9: 108880152-108880152 on Build GRCh38
rs983457144 Chr.9: 108879998-108879998 on Build GRCh38
rs746668947 Chr.9: 108880087-108880087 on Build GRCh38
rs146862578 Chr.9: 108880295-108880295 on Build GRCh38
rs56255674 Chr.9: 108880346-108880346 on Build GRCh38
rs907873245 Chr.9: 108880231-108880231 on Build GRCh38
rs756080414 Chr.9: 108880032-108880032 on Build GRCh38
rs541942558 Chr.9: 108880229-108880229 on Build GRCh38
rs910335280 Chr.9: 108880149-108880149 on Build GRCh38
rs907776120 Chr.9: 108879999-108879999 on Build GRCh38
rs372499865 Chr.9: 108880115-108880115 on Build GRCh38
rs143838889 Chr.9: 108880309-108880309 on Build GRCh38
rs764056417 Chr.9: 108880134-108880134 on Build GRCh38
rs1027694965 Chr.9: 108880202-108880202 on Build GRCh38
rs200154542 Chr.9: 108880180-108880180 on Build GRCh38
rs149897637 Chr.9: 108880039-108880039 on Build GRCh38
rs180876404 Chr.9: 108880360-108880360 on Build GRCh38
rs140712847 Chr.9: 108880304-108880304 on Build GRCh38
rs921868292 Chr.9: 108880292-108880292 on Build GRCh38
rs760972192 Chr.9: 108880125-108880125 on Build GRCh38

View Full Product Details