Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 42186916-42186409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31A6
Entrez Gene ID
389730
Gene Name
SPATA31 subfamily A member 6
Gene Aliases
FAM75A6
UniGene
Hs.455299
Gene Chromosome Location
Chr.9: 42183659-42189887 on Build GRCh38
Transcripts
NM_001145196.1 XM_011517871.2

View Full Product Details