Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 70140031-70140544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMDC2
Entrez Gene ID
256691
Gene Name
MAM domain containing 2
Gene Aliases
-
UniGene
Hs.547172
Gene Chromosome Location
Chr.9: 70043519-70226972 on Build GRCh38
Transcripts
NM_153267.4 NR_125850.1 XM_011518471.1 XM_011518475.2 XM_017014587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6706430 c.995-1G>T p.? Chr.9: 70140144-70140144 on Build GRCh38
5806576 c.1069G>T p.G357C Chr.9: 70140219-70140219 on Build GRCh38
4393557 c.1098A>T p.K366N Chr.9: 70140248-70140248 on Build GRCh38
6476629 c.1062T>C p.D354D Chr.9: 70140212-70140212 on Build GRCh38
6346085 c.1014G>T p.V338V Chr.9: 70140164-70140164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201926844 Chr.9: 70140322-70140322 on Build GRCh38
rs527377179 Chr.9: 70140464-70140464 on Build GRCh38
rs976064540 Chr.9: 70140375-70140375 on Build GRCh38
rs763822507 Chr.9: 70140263-70140263 on Build GRCh38
rs765858332 Chr.9: 70140161-70140161 on Build GRCh38
rs1018496287 Chr.9: 70140330-70140330 on Build GRCh38
rs114288682 Chr.9: 70140391-70140391 on Build GRCh38
rs761964221 Chr.9: 70140245-70140245 on Build GRCh38
rs753411697 Chr.9: 70140198-70140198 on Build GRCh38
rs555534315 Chr.9: 70140349-70140349 on Build GRCh38
rs79430954 Chr.9: 70140213-70140213 on Build GRCh38
rs917734319 Chr.9: 70140475-70140475 on Build GRCh38
rs569254371 Chr.9: 70140063-70140063 on Build GRCh38
rs61609258 Chr.9: 70140213-70140215 on Build GRCh38
rs746839312 Chr.9: 70140335-70140335 on Build GRCh38
rs1027279828 Chr.9: 70140390-70140390 on Build GRCh38
rs955381738 Chr.9: 70140304-70140304 on Build GRCh38
rs1008282106 Chr.9: 70140351-70140351 on Build GRCh38
rs986956757 Chr.9: 70140092-70140092 on Build GRCh38
rs892519855 Chr.9: 70140345-70140345 on Build GRCh38
rs777250680 Chr.9: 70140183-70140183 on Build GRCh38
rs763412261 Chr.9: 70140163-70140163 on Build GRCh38
rs148444015 Chr.9: 70140261-70140261 on Build GRCh38
rs771986746 Chr.9: 70140233-70140233 on Build GRCh38
rs750086220 Chr.9: 70140117-70140117 on Build GRCh38
rs776665442 Chr.9: 70140239-70140239 on Build GRCh38
rs773420406 Chr.9: 70140255-70140255 on Build GRCh38
rs768743168 Chr.9: 70140122-70140122 on Build GRCh38
rs757982814 Chr.9: 70140254-70140254 on Build GRCh38
rs759771697 Chr.9: 70140262-70140262 on Build GRCh38
rs980428904 Chr.9: 70140378-70140379 on Build GRCh38
rs752971650 Chr.9: 70140136-70140136 on Build GRCh38
rs200527108 Chr.9: 70140162-70140162 on Build GRCh38
rs756665675 Chr.9: 70140205-70140205 on Build GRCh38
rs761756187 Chr.9: 70140160-70140160 on Build GRCh38
rs755839072 Chr.9: 70140180-70140180 on Build GRCh38
rs759982191 Chr.9: 70140212-70140213 on Build GRCh38
rs907551983 Chr.9: 70140433-70140433 on Build GRCh38
rs778775307 Chr.9: 70140102-70140102 on Build GRCh38
rs144097810 Chr.9: 70140264-70140264 on Build GRCh38
rs544084018 Chr.9: 70140370-70140370 on Build GRCh38
rs766971462 Chr.9: 70140257-70140257 on Build GRCh38
rs1031030199 Chr.9: 70140342-70140342 on Build GRCh38
rs936602798 Chr.9: 70140493-70140493 on Build GRCh38
rs751270472 Chr.9: 70140307-70140308 on Build GRCh38
rs947950872 Chr.9: 70140476-70140476 on Build GRCh38
rs559996712 Chr.9: 70140448-70140448 on Build GRCh38
rs776807614 Chr.9: 70140130-70140130 on Build GRCh38
rs940383329 Chr.9: 70140463-70140463 on Build GRCh38
rs956850094 Chr.9: 70140054-70140054 on Build GRCh38
rs539816273 Chr.9: 70140097-70140097 on Build GRCh38
rs968217823 Chr.9: 70140082-70140082 on Build GRCh38
rs947930396 Chr.9: 70140313-70140313 on Build GRCh38
rs770932881 Chr.9: 70140099-70140099 on Build GRCh38
rs763789719 Chr.9: 70140301-70140304 on Build GRCh38
rs751855886 Chr.9: 70140175-70140175 on Build GRCh38
rs546918674 Chr.9: 70140489-70140489 on Build GRCh38
rs778456323 Chr.9: 70140285-70140285 on Build GRCh38
rs557984489 Chr.9: 70140256-70140256 on Build GRCh38
rs771790477 Chr.9: 70140116-70140116 on Build GRCh38
rs766916360 Chr.9: 70140169-70140169 on Build GRCh38
rs368297806 Chr.9: 70140235-70140235 on Build GRCh38
rs573636222 Chr.9: 70140359-70140359 on Build GRCh38
rs921562982 Chr.9: 70140191-70140191 on Build GRCh38
rs896322402 Chr.9: 70140300-70140300 on Build GRCh38
rs144542381 Chr.9: 70140238-70140238 on Build GRCh38
rs779922459 Chr.9: 70140332-70140332 on Build GRCh38
rs1045023109 Chr.9: 70140316-70140316 on Build GRCh38
rs779877522 Chr.9: 70140234-70140234 on Build GRCh38
rs1045366396 Chr.9: 70140482-70140482 on Build GRCh38
rs183002053 Chr.9: 70140430-70140430 on Build GRCh38
rs745875443 Chr.9: 70140228-70140228 on Build GRCh38
rs749916218 Chr.9: 70140310-70140310 on Build GRCh38
rs149704650 Chr.9: 70140225-70140225 on Build GRCh38
rs562170531 Chr.9: 70140374-70140374 on Build GRCh38
rs754669150 Chr.9: 70140340-70140340 on Build GRCh38
rs745679124 Chr.9: 70140110-70140110 on Build GRCh38
rs1019704390 Chr.9: 70140354-70140354 on Build GRCh38
rs761019857 Chr.9: 70140120-70140120 on Build GRCh38
rs988496529 Chr.9: 70140353-70140353 on Build GRCh38

View Full Product Details