Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76612781-76613277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
UniGene
Hs.262857
Gene Chromosome Location
Chr.9: 76611376-76906220 on Build GRCh38
Transcripts
NM_001308047.1 NM_001308048.1 NM_001308049.1 NM_001308050.1 NM_001308051.1 NM_015225.2 NR_131751.1 XM_005251745.1 XM_005251746.1 XM_005251748.1 XM_005251750.1 XM_005251751.1 XM_005251754.1 XM_006716982.1 XM_006716983.1 XM_006716985.1 XM_006716986.1 XM_011518323.1 XM_011518327.1 XM_017014345.1 XM_017014346.1 XM_017014347.1 XM_017014348.1 XM_017014349.1 XM_017014350.1 XM_017014351.1 XM_017014352.1 XM_017014354.1 XM_017014355.1 XM_017014356.1 XM_017014357.1 XM_017014358.1 XM_017014359.1

SNP Detail

SNP ID Chromosome Location
rs140693448 Chr.9: 76612948-76612948 on Build GRCh38
rs952685331 Chr.9: 76613012-76613012 on Build GRCh38
rs892837347 Chr.9: 76613197-76613197 on Build GRCh38
rs541812927 Chr.9: 76612926-76612926 on Build GRCh38
rs887585032 Chr.9: 76612972-76612972 on Build GRCh38
rs576087622 Chr.9: 76613024-76613024 on Build GRCh38
rs901599197 Chr.9: 76612810-76612810 on Build GRCh38
rs561602118 Chr.9: 76613012-76613012 on Build GRCh38
rs1006036594 Chr.9: 76613009-76613009 on Build GRCh38
rs7028529 Chr.9: 76613126-76613126 on Build GRCh38
rs1052787806 Chr.9: 76613161-76613161 on Build GRCh38
rs1033502885 Chr.9: 76613030-76613030 on Build GRCh38
rs1044026886 Chr.9: 76613223-76613223 on Build GRCh38
rs114094277 Chr.9: 76613209-76613209 on Build GRCh38
rs539271219 Chr.9: 76612806-76612806 on Build GRCh38
rs575312713 Chr.9: 76613135-76613135 on Build GRCh38
rs934432268 Chr.9: 76613157-76613157 on Build GRCh38
rs890021359 Chr.9: 76612913-76612913 on Build GRCh38
rs1040052220 Chr.9: 76612927-76612927 on Build GRCh38
rs144654713 Chr.9: 76612983-76612983 on Build GRCh38
rs780448397 Chr.9: 76613025-76613025 on Build GRCh38
rs998585307 Chr.9: 76612844-76612844 on Build GRCh38
rs959275647 Chr.9: 76613114-76613114 on Build GRCh38
rs1003121886 Chr.9: 76612936-76612936 on Build GRCh38
rs945450636 Chr.9: 76612815-76612815 on Build GRCh38
rs142874325 Chr.9: 76612874-76612874 on Build GRCh38
rs578218996 Chr.9: 76612922-76612922 on Build GRCh38
rs112066929 Chr.9: 76612933-76612933 on Build GRCh38
rs985751611 Chr.9: 76613026-76613026 on Build GRCh38
rs966743438 Chr.9: 76613208-76613208 on Build GRCh38
rs564735281 Chr.9: 76613205-76613205 on Build GRCh38
rs911467945 Chr.9: 76613051-76613051 on Build GRCh38
rs944271269 Chr.9: 76613079-76613079 on Build GRCh38
rs879058856 Chr.9: 76613125-76613133 on Build GRCh38
rs777099370 Chr.9: 76612894-76612894 on Build GRCh38
rs749004157 Chr.9: 76612911-76612911 on Build GRCh38
rs993621479 Chr.9: 76612942-76612942 on Build GRCh38
rs763122431 Chr.9: 76612893-76612893 on Build GRCh38

View Full Product Details