Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133684970-133685493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARDH
Entrez Gene ID
1757
Gene Name
sarcosine dehydrogenase
Gene Aliases
BPR-2, DMGDHL1, SAR, SARD, SDH
UniGene
Hs.198003
Gene Chromosome Location
Chr.9: 133663074-133739958 on Build GRCh38
Transcripts
NM_001134707.1 NM_007101.3 XM_017014367.1 XM_017014368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3952457 c.373G>T p.V125L Chr.9: 133685267-133685267 on Build GRCh38
6771883 c.435C>T p.A145A Chr.9: 133685205-133685205 on Build GRCh38
3952456 c.2089G>T p.V697L Chr.9: 133685267-133685267 on Build GRCh38
4815200 c.447G>A p.L149L Chr.9: 133685193-133685193 on Build GRCh38
455555 c.2163G>A p.L721L Chr.9: 133685193-133685193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980036362 Chr.9: 133685171-133685171 on Build GRCh38
rs774983027 Chr.9: 133685328-133685328 on Build GRCh38
rs201684078 Chr.9: 133685204-133685204 on Build GRCh38
rs202085298 Chr.9: 133685155-133685155 on Build GRCh38
rs774582897 Chr.9: 133685254-133685254 on Build GRCh38
rs545421698 Chr.9: 133685305-133685305 on Build GRCh38
rs371152061 Chr.9: 133685216-133685216 on Build GRCh38
rs143735079 Chr.9: 133685133-133685133 on Build GRCh38
rs762307263 Chr.9: 133685258-133685258 on Build GRCh38
rs748136846 Chr.9: 133685207-133685207 on Build GRCh38
rs531925998 Chr.9: 133685333-133685333 on Build GRCh38
rs148100733 Chr.9: 133685306-133685306 on Build GRCh38
rs896534279 Chr.9: 133685111-133685111 on Build GRCh38
rs751296591 Chr.9: 133685298-133685298 on Build GRCh38
rs759149932 Chr.9: 133685160-133685160 on Build GRCh38
rs776020646 Chr.9: 133685157-133685157 on Build GRCh38
rs999969007 Chr.9: 133685229-133685229 on Build GRCh38
rs370488983 Chr.9: 133685467-133685467 on Build GRCh38
rs758427155 Chr.9: 133685285-133685285 on Build GRCh38
rs775931677 Chr.9: 133685243-133685243 on Build GRCh38
rs7859013 Chr.9: 133685130-133685130 on Build GRCh38
rs770969732 Chr.9: 133685310-133685316 on Build GRCh38
rs904123645 Chr.9: 133685056-133685056 on Build GRCh38
rs547821837 Chr.9: 133685076-133685076 on Build GRCh38
rs973952740 Chr.9: 133685008-133685008 on Build GRCh38
rs769037548 Chr.9: 133685327-133685327 on Build GRCh38
rs758232417 Chr.9: 133685213-133685213 on Build GRCh38
rs1043978430 Chr.9: 133685041-133685041 on Build GRCh38
rs9409845 Chr.9: 133685354-133685354 on Build GRCh38
rs374259091 Chr.9: 133685284-133685284 on Build GRCh38
rs922111256 Chr.9: 133684997-133684997 on Build GRCh38
rs916748281 Chr.9: 133685456-133685456 on Build GRCh38
rs746490253 Chr.9: 133685279-133685287 on Build GRCh38
rs535586479 Chr.9: 133685039-133685039 on Build GRCh38
rs777559327 Chr.9: 133685291-133685291 on Build GRCh38
rs776023318 Chr.9: 133685168-133685168 on Build GRCh38
rs1023761475 Chr.9: 133685125-133685125 on Build GRCh38
rs376087668 Chr.9: 133685149-133685149 on Build GRCh38
rs144844844 Chr.9: 133685244-133685244 on Build GRCh38
rs770497060 Chr.9: 133685232-133685232 on Build GRCh38
rs757433211 Chr.9: 133685189-133685189 on Build GRCh38
rs558168499 Chr.9: 133685098-133685098 on Build GRCh38
rs544052060 Chr.9: 133685352-133685352 on Build GRCh38
rs749726623 Chr.9: 133685312-133685312 on Build GRCh38
rs764474282 Chr.9: 133685182-133685182 on Build GRCh38
rs780952882 Chr.9: 133685233-133685233 on Build GRCh38
rs769330871 Chr.9: 133685242-133685242 on Build GRCh38
rs1028014363 Chr.9: 133685444-133685444 on Build GRCh38
rs184547854 Chr.9: 133685174-133685174 on Build GRCh38
rs946923773 Chr.9: 133685470-133685470 on Build GRCh38
rs982444621 Chr.9: 133685139-133685139 on Build GRCh38
rs139740832 Chr.9: 133685259-133685259 on Build GRCh38
rs368425650 Chr.9: 133685310-133685310 on Build GRCh38
rs975069646 Chr.9: 133685375-133685375 on Build GRCh38
rs760806131 Chr.9: 133685265-133685265 on Build GRCh38
rs905306905 Chr.9: 133685300-133685300 on Build GRCh38
rs188167418 Chr.9: 133685446-133685446 on Build GRCh38
rs111780848 Chr.9: 133685445-133685445 on Build GRCh38
rs750467189 Chr.9: 133685200-133685200 on Build GRCh38
rs539883039 Chr.9: 133685086-133685086 on Build GRCh38
rs762063137 Chr.9: 133685331-133685331 on Build GRCh38
rs995770621 Chr.9: 133685131-133685131 on Build GRCh38
rs937106249 Chr.9: 133685381-133685381 on Build GRCh38
rs932174253 Chr.9: 133685038-133685038 on Build GRCh38
rs746618437 Chr.9: 133685219-133685219 on Build GRCh38
rs45469492 Chr.9: 133685138-133685138 on Build GRCh38
rs529538329 Chr.9: 133685355-133685355 on Build GRCh38
rs757121220 Chr.9: 133685299-133685299 on Build GRCh38
rs921049381 Chr.9: 133685161-133685161 on Build GRCh38
rs778972959 Chr.9: 133685205-133685205 on Build GRCh38
rs571790478 Chr.9: 133685185-133685185 on Build GRCh38
rs779552074 Chr.9: 133685311-133685311 on Build GRCh38
rs768950787 Chr.9: 133685248-133685248 on Build GRCh38
rs750707201 Chr.9: 133685264-133685264 on Build GRCh38
rs201589789 Chr.9: 133685162-133685162 on Build GRCh38
rs928299169 Chr.9: 133685360-133685360 on Build GRCh38
rs772972407 Chr.9: 133685148-133685148 on Build GRCh38
rs1013686668 Chr.9: 133685117-133685117 on Build GRCh38
rs149347502 Chr.9: 133685078-133685078 on Build GRCh38
rs376438251 Chr.9: 133685196-133685196 on Build GRCh38
rs761897698 Chr.9: 133685134-133685134 on Build GRCh38
rs769244988 Chr.9: 133685307-133685307 on Build GRCh38

View Full Product Details