Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133771870-133772367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV2
Entrez Gene ID
7410
Gene Name
vav guanine nucleotide exchange factor 2
Gene Aliases
VAV-2
UniGene
Hs.369921
Gene Chromosome Location
Chr.9: 133761894-133992374 on Build GRCh38
Transcripts
NM_001134398.1 NM_003371.3 XM_005272213.1 XM_017015108.1 XM_017015109.1 XM_017015110.1 XM_017015111.1 XM_017015112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5432639 c.2155C>T p.H719Y Chr.9: 133772027-133772027 on Build GRCh38
5346118 c.2132A>T p.K711M Chr.9: 133772020-133772020 on Build GRCh38
7409092 c.2135+3_2137del2888 p.? Chr.9: 133772045-133774932 on Build GRCh38
7093444 c.2181C>A p.F727L Chr.9: 133771971-133771971 on Build GRCh38
7298657 c.2164A>G p.T722A Chr.9: 133771988-133771988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869167831 Chr.9: 133772101-133772102 on Build GRCh38
rs760375847 Chr.9: 133771990-133771990 on Build GRCh38
rs757488344 Chr.9: 133772060-133772061 on Build GRCh38
rs1012487581 Chr.9: 133772024-133772024 on Build GRCh38
rs945730995 Chr.9: 133772107-133772107 on Build GRCh38
rs749234864 Chr.9: 133771944-133771944 on Build GRCh38
rs950747594 Chr.9: 133772277-133772277 on Build GRCh38
rs747942896 Chr.9: 133771999-133772000 on Build GRCh38
rs653967 Chr.9: 133772076-133772076 on Build GRCh38
rs367642598 Chr.9: 133772075-133772076 on Build GRCh38
rs954890569 Chr.9: 133771926-133771926 on Build GRCh38
rs948291085 Chr.9: 133772201-133772201 on Build GRCh38
rs902218411 Chr.9: 133772126-133772126 on Build GRCh38
rs535219394 Chr.9: 133772163-133772163 on Build GRCh38
rs765599332 Chr.9: 133772089-133772089 on Build GRCh38
rs374536021 Chr.9: 133772048-133772048 on Build GRCh38
rs142210678 Chr.9: 133772291-133772291 on Build GRCh38
rs771885409 Chr.9: 133772077-133772077 on Build GRCh38
rs964872438 Chr.9: 133772221-133772221 on Build GRCh38
rs753384021 Chr.9: 133772013-133772013 on Build GRCh38
rs56171261 Chr.9: 133772055-133772056 on Build GRCh38
rs370767093 Chr.9: 133771950-133771950 on Build GRCh38
rs898516474 Chr.9: 133772051-133772051 on Build GRCh38
rs144160596 Chr.9: 133772037-133772037 on Build GRCh38
rs551049227 Chr.9: 133772102-133772102 on Build GRCh38
rs774621815 Chr.9: 133772091-133772096 on Build GRCh38
rs1006618030 Chr.9: 133772170-133772170 on Build GRCh38
rs547132645 Chr.9: 133772069-133772069 on Build GRCh38
rs1018115223 Chr.9: 133772318-133772318 on Build GRCh38
rs758505295 Chr.9: 133771936-133771936 on Build GRCh38
rs1008347798 Chr.9: 133772303-133772303 on Build GRCh38
rs766811886 Chr.9: 133771924-133771924 on Build GRCh38
rs780571195 Chr.9: 133772076-133772077 on Build GRCh38
rs377063152 Chr.9: 133771919-133771919 on Build GRCh38
rs745949559 Chr.9: 133772071-133772072 on Build GRCh38
rs911117323 Chr.9: 133771915-133771915 on Build GRCh38
rs749748745 Chr.9: 133772077-133772078 on Build GRCh38
rs56259882 Chr.9: 133772202-133772202 on Build GRCh38
rs1056550380 Chr.9: 133772148-133772148 on Build GRCh38
rs991148385 Chr.9: 133772117-133772117 on Build GRCh38
rs773055597 Chr.9: 133772078-133772078 on Build GRCh38
rs143593335 Chr.9: 133772016-133772016 on Build GRCh38
rs927947144 Chr.9: 133772304-133772304 on Build GRCh38
rs902666283 Chr.9: 133771970-133771970 on Build GRCh38
rs747020960 Chr.9: 133771938-133771938 on Build GRCh38
rs751820840 Chr.9: 133772042-133772042 on Build GRCh38
rs372643451 Chr.9: 133772091-133772091 on Build GRCh38
rs1026122205 Chr.9: 133772065-133772065 on Build GRCh38
rs567892114 Chr.9: 133772138-133772138 on Build GRCh38
rs977126154 Chr.9: 133772108-133772108 on Build GRCh38
rs772851620 Chr.9: 133772094-133772104 on Build GRCh38
rs373500982 Chr.9: 133771935-133771935 on Build GRCh38
rs990887483 Chr.9: 133772313-133772313 on Build GRCh38
rs757621399 Chr.9: 133772047-133772047 on Build GRCh38
rs1046506225 Chr.9: 133772269-133772269 on Build GRCh38
rs773995312 Chr.9: 133771959-133771959 on Build GRCh38
rs1050857683 Chr.9: 133772157-133772157 on Build GRCh38
rs778899508 Chr.9: 133771929-133771930 on Build GRCh38
rs995537215 Chr.9: 133772241-133772241 on Build GRCh38
rs902626136 Chr.9: 133772286-133772286 on Build GRCh38
rs146097221 Chr.9: 133771962-133771962 on Build GRCh38
rs112734792 Chr.9: 133771904-133771904 on Build GRCh38
rs532946154 Chr.9: 133772083-133772083 on Build GRCh38
rs758798561 Chr.9: 133772041-133772041 on Build GRCh38
rs1025645154 Chr.9: 133772285-133772285 on Build GRCh38
rs199810832 Chr.9: 133772067-133772067 on Build GRCh38
rs761341497 Chr.9: 133771913-133771913 on Build GRCh38
rs749895383 Chr.9: 133771914-133771914 on Build GRCh38
rs962820666 Chr.9: 133771895-133771895 on Build GRCh38
rs867032627 Chr.9: 133771964-133771964 on Build GRCh38
rs764550755 Chr.9: 133772098-133772098 on Build GRCh38
rs369497454 Chr.9: 133772164-133772164 on Build GRCh38
rs200281508 Chr.9: 133771940-133771940 on Build GRCh38
rs999182445 Chr.9: 133772149-133772149 on Build GRCh38
rs370959712 Chr.9: 133771934-133771934 on Build GRCh38
rs762237887 Chr.9: 133772090-133772091 on Build GRCh38
rs769132741 Chr.9: 133772087-133772090 on Build GRCh38
rs879638550 Chr.9: 133772055-133772056 on Build GRCh38
rs565635701 Chr.9: 133772070-133772070 on Build GRCh38
rs755573609 Chr.9: 133771932-133771932 on Build GRCh38
rs781463003 Chr.9: 133772069-133772070 on Build GRCh38
rs367787989 Chr.9: 133772086-133772086 on Build GRCh38
rs772862738 Chr.9: 133771980-133771980 on Build GRCh38
rs771684457 Chr.9: 133772092-133772093 on Build GRCh38
rs543238251 Chr.9: 133771971-133771971 on Build GRCh38
rs553174775 Chr.9: 133771901-133771901 on Build GRCh38
rs149500041 Chr.9: 133771908-133771908 on Build GRCh38
rs200612399 Chr.9: 133771928-133771928 on Build GRCh38
rs898762143 Chr.9: 133772247-133772247 on Build GRCh38
rs541565274 Chr.9: 133771896-133771896 on Build GRCh38
rs754327178 Chr.9: 133771930-133771930 on Build GRCh38
rs771150259 Chr.9: 133771939-133771939 on Build GRCh38
rs961090432 Chr.9: 133772307-133772307 on Build GRCh38
rs2510244 Chr.9: 133772120-133772120 on Build GRCh38
rs938443276 Chr.9: 133772142-133772142 on Build GRCh38
rs914224910 Chr.9: 133772097-133772097 on Build GRCh38
rs949400494 Chr.9: 133772230-133772230 on Build GRCh38
rs765866512 Chr.9: 133772008-133772008 on Build GRCh38
rs537711503 Chr.9: 133772116-133772116 on Build GRCh38
rs759935502 Chr.9: 133772014-133772014 on Build GRCh38
rs140004145 Chr.9: 133771967-133771967 on Build GRCh38
rs201571757 Chr.9: 133772316-133772317 on Build GRCh38
rs373654300 Chr.9: 133772079-133772079 on Build GRCh38
rs570343548 Chr.9: 133772111-133772111 on Build GRCh38
rs994553876 Chr.9: 133772266-133772266 on Build GRCh38
rs994252596 Chr.9: 133772055-133772055 on Build GRCh38
rs368060780 Chr.9: 133772060-133772060 on Build GRCh38

View Full Product Details