Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127710501-127711007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP157
Entrez Gene ID
286207
Gene Name
cilia and flagella associated protein 157
Gene Aliases
C9orf117
UniGene
Hs.652519
Gene Chromosome Location
Chr.9: 127706261-127716002 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014629.1 XM_017014630.1 XM_017014277.1

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
UniGene
Hs.653265
Gene Chromosome Location
Chr.9: 127695024-127716714 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014629.1 XM_017014630.1 XM_017014277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4640890 c.460T>G p.F154V Chr.9: 127710627-127710627 on Build GRCh38
4553157 c.582G>A p.K194K Chr.9: 127710749-127710749 on Build GRCh38
1105371 c.486G>A p.T162T Chr.9: 127710653-127710653 on Build GRCh38
4450069 c.434-2A>C p.? Chr.9: 127710599-127710599 on Build GRCh38
1460105 c.469C>T p.Q157* Chr.9: 127710636-127710636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975481871 Chr.9: 127710867-127710867 on Build GRCh38
rs779859762 Chr.9: 127710738-127710738 on Build GRCh38
rs750995835 Chr.9: 127710587-127710587 on Build GRCh38
rs532840158 Chr.9: 127710713-127710713 on Build GRCh38
rs754330945 Chr.9: 127710577-127710577 on Build GRCh38
rs1038282524 Chr.9: 127710960-127710960 on Build GRCh38
rs894848826 Chr.9: 127710562-127710562 on Build GRCh38
rs1046053615 Chr.9: 127710843-127710843 on Build GRCh38
rs773156489 Chr.9: 127710594-127710594 on Build GRCh38
rs542140134 Chr.9: 127710578-127710578 on Build GRCh38
rs905350154 Chr.9: 127710767-127710767 on Build GRCh38
rs912623993 Chr.9: 127710727-127710729 on Build GRCh38
rs531810174 Chr.9: 127710969-127710969 on Build GRCh38
rs375545874 Chr.9: 127710586-127710586 on Build GRCh38
rs760642552 Chr.9: 127710655-127710655 on Build GRCh38
rs180978665 Chr.9: 127710672-127710672 on Build GRCh38
rs986767347 Chr.9: 127710907-127710907 on Build GRCh38
rs1012610063 Chr.9: 127710762-127710762 on Build GRCh38
rs549250735 Chr.9: 127710910-127710910 on Build GRCh38
rs76294682 Chr.9: 127710765-127710765 on Build GRCh38
rs966782691 Chr.9: 127710920-127710920 on Build GRCh38
rs201004785 Chr.9: 127710928-127710928 on Build GRCh38
rs538016055 Chr.9: 127710825-127710825 on Build GRCh38
rs946746259 Chr.9: 127710647-127710647 on Build GRCh38
rs933870019 Chr.9: 127710688-127710688 on Build GRCh38
rs956945542 Chr.9: 127710792-127710792 on Build GRCh38
rs1012621613 Chr.9: 127710543-127710543 on Build GRCh38
rs915256378 Chr.9: 127710646-127710646 on Build GRCh38
rs996141005 Chr.9: 127710868-127710868 on Build GRCh38
rs1055196822 Chr.9: 127710522-127710522 on Build GRCh38
rs374960552 Chr.9: 127710716-127710716 on Build GRCh38
rs77175267 Chr.9: 127710708-127710708 on Build GRCh38
rs948411463 Chr.9: 127710979-127710979 on Build GRCh38
rs921373238 Chr.9: 127710870-127710870 on Build GRCh38
rs745560232 Chr.9: 127710595-127710595 on Build GRCh38
rs968074204 Chr.9: 127710850-127710850 on Build GRCh38
rs754825954 Chr.9: 127710764-127710764 on Build GRCh38
rs190081607 Chr.9: 127710652-127710652 on Build GRCh38
rs761005152 Chr.9: 127710571-127710571 on Build GRCh38
rs954292706 Chr.9: 127710871-127710871 on Build GRCh38
rs755420258 Chr.9: 127710582-127710582 on Build GRCh38
rs779226277 Chr.9: 127710875-127710875 on Build GRCh38
rs370956723 Chr.9: 127710574-127710574 on Build GRCh38
rs769703138 Chr.9: 127710596-127710596 on Build GRCh38
rs202023603 Chr.9: 127710601-127710601 on Build GRCh38
rs765610028 Chr.9: 127710685-127710685 on Build GRCh38
rs754134625 Chr.9: 127710653-127710653 on Build GRCh38
rs760945846 Chr.9: 127710650-127710650 on Build GRCh38
rs1016167511 Chr.9: 127710848-127710848 on Build GRCh38
rs945454167 Chr.9: 127710736-127710736 on Build GRCh38
rs919578686 Chr.9: 127710967-127710967 on Build GRCh38
rs908373986 Chr.9: 127710919-127710919 on Build GRCh38
rs367946787 Chr.9: 127710737-127710737 on Build GRCh38
rs934088675 Chr.9: 127710873-127710873 on Build GRCh38
rs773275895 Chr.9: 127710631-127710631 on Build GRCh38
rs185882520 Chr.9: 127710714-127710714 on Build GRCh38
rs1037961580 Chr.9: 127710750-127710750 on Build GRCh38
rs200486914 Chr.9: 127710607-127710607 on Build GRCh38
rs529571369 Chr.9: 127710876-127710876 on Build GRCh38
rs866067906 Chr.9: 127710630-127710630 on Build GRCh38
rs988803081 Chr.9: 127710812-127710812 on Build GRCh38
rs986615800 Chr.9: 127710878-127710878 on Build GRCh38
rs368488988 Chr.9: 127710769-127710769 on Build GRCh38
rs749054423 Chr.9: 127710757-127710757 on Build GRCh38
rs1035612650 Chr.9: 127710779-127710779 on Build GRCh38
rs772040393 Chr.9: 127710786-127710786 on Build GRCh38
rs79991050 Chr.9: 127710606-127710606 on Build GRCh38
rs1013277414 Chr.9: 127710569-127710569 on Build GRCh38
rs756816294 Chr.9: 127710592-127710592 on Build GRCh38
rs780049540 Chr.9: 127710600-127710600 on Build GRCh38
rs761752193 Chr.9: 127710604-127710604 on Build GRCh38

View Full Product Details