Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127712621-127713129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP157
Entrez Gene ID
286207
Gene Name
cilia and flagella associated protein 157
Gene Aliases
C9orf117
UniGene
Hs.652519
Gene Chromosome Location
Chr.9: 127706261-127716002 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 XM_017014277.1

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
UniGene
Hs.653265
Gene Chromosome Location
Chr.9: 127695024-127716714 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 XM_017014277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616783 c.112G>A p.V38I Chr.9: 127712736-127712736 on Build GRCh38
3654454 c.1195G>A p.E399K Chr.9: 127712766-127712766 on Build GRCh38
6226728 c.1379G>T p.G460V Chr.9: 127713094-127713094 on Build GRCh38
6616784 c.55G>T p.G19C Chr.9: 127712679-127712679 on Build GRCh38
3685501 c.1161T>G p.S387R Chr.9: 127712732-127712732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78968934 Chr.9: 127713092-127713092 on Build GRCh38
rs780458865 Chr.9: 127713108-127713108 on Build GRCh38
rs972437141 Chr.9: 127713013-127713013 on Build GRCh38
rs753629552 Chr.9: 127712826-127712826 on Build GRCh38
rs972848416 Chr.9: 127712736-127712736 on Build GRCh38
rs575929587 Chr.9: 127712906-127712906 on Build GRCh38
rs375679788 Chr.9: 127712681-127712681 on Build GRCh38
rs374413353 Chr.9: 127712811-127712811 on Build GRCh38
rs771071414 Chr.9: 127712797-127712797 on Build GRCh38
rs781021008 Chr.9: 127712656-127712656 on Build GRCh38
rs748181351 Chr.9: 127712700-127712700 on Build GRCh38
rs543912514 Chr.9: 127713051-127713051 on Build GRCh38
rs775800130 Chr.9: 127713017-127713017 on Build GRCh38
rs767253698 Chr.9: 127712742-127712742 on Build GRCh38
rs768608600 Chr.9: 127712692-127712692 on Build GRCh38
rs777719317 Chr.9: 127712986-127712986 on Build GRCh38
rs772785644 Chr.9: 127712919-127712919 on Build GRCh38
rs935896423 Chr.9: 127712997-127712997 on Build GRCh38
rs374561061 Chr.9: 127713106-127713106 on Build GRCh38
rs375153055 Chr.9: 127712985-127712985 on Build GRCh38
rs541652643 Chr.9: 127712972-127712972 on Build GRCh38
rs960997152 Chr.9: 127713006-127713006 on Build GRCh38
rs1000115192 Chr.9: 127712714-127712714 on Build GRCh38
rs746139886 Chr.9: 127713022-127713022 on Build GRCh38
rs370271059 Chr.9: 127712717-127712717 on Build GRCh38
rs750531847 Chr.9: 127713103-127713103 on Build GRCh38
rs763991791 Chr.9: 127713091-127713091 on Build GRCh38
rs781442675 Chr.9: 127713021-127713021 on Build GRCh38
rs776332587 Chr.9: 127713075-127713075 on Build GRCh38
rs767206683 Chr.9: 127712813-127712813 on Build GRCh38
rs544952716 Chr.9: 127712910-127712910 on Build GRCh38
rs746718039 Chr.9: 127712835-127712835 on Build GRCh38
rs781288795 Chr.9: 127712896-127712896 on Build GRCh38
rs773446696 Chr.9: 127712715-127712715 on Build GRCh38
rs746744439 Chr.9: 127712854-127712855 on Build GRCh38
rs751586501 Chr.9: 127713003-127713003 on Build GRCh38
rs377334421 Chr.9: 127712819-127712819 on Build GRCh38
rs572650181 Chr.9: 127712966-127712966 on Build GRCh38
rs79537388 Chr.9: 127713033-127713033 on Build GRCh38
rs759346417 Chr.9: 127712975-127712975 on Build GRCh38
rs374070528 Chr.9: 127713062-127713062 on Build GRCh38
rs61738788 Chr.9: 127712718-127712718 on Build GRCh38
rs917308947 Chr.9: 127712930-127712930 on Build GRCh38
rs770167060 Chr.9: 127712984-127712984 on Build GRCh38
rs776139567 Chr.9: 127712913-127712913 on Build GRCh38
rs370617973 Chr.9: 127712983-127712983 on Build GRCh38
rs199630039 Chr.9: 127713004-127713004 on Build GRCh38
rs776985946 Chr.9: 127712800-127712800 on Build GRCh38
rs764999071 Chr.9: 127712977-127712977 on Build GRCh38
rs777965475 Chr.9: 127712777-127712777 on Build GRCh38
rs769303957 Chr.9: 127712918-127712918 on Build GRCh38
rs377150093 Chr.9: 127712840-127712840 on Build GRCh38
rs1031808249 Chr.9: 127712722-127712722 on Build GRCh38
rs370613192 Chr.9: 127712812-127712812 on Build GRCh38
rs774434087 Chr.9: 127712698-127712698 on Build GRCh38
rs551402620 Chr.9: 127712716-127712716 on Build GRCh38
rs553511710 Chr.9: 127712741-127712741 on Build GRCh38
rs775021171 Chr.9: 127713053-127713053 on Build GRCh38
rs772244241 Chr.9: 127712706-127712706 on Build GRCh38
rs748178735 Chr.9: 127712757-127712757 on Build GRCh38
rs187320210 Chr.9: 127712831-127712831 on Build GRCh38
rs756490318 Chr.9: 127713023-127713023 on Build GRCh38
rs759050313 Chr.9: 127713082-127713082 on Build GRCh38
rs522328 Chr.9: 127712732-127712732 on Build GRCh38
rs760135818 Chr.9: 127712920-127712920 on Build GRCh38
rs1007513979 Chr.9: 127712675-127712675 on Build GRCh38
rs747287030 Chr.9: 127712778-127712778 on Build GRCh38
rs930782828 Chr.9: 127712876-127712876 on Build GRCh38
rs771886876 Chr.9: 127712690-127712690 on Build GRCh38
rs372095224 Chr.9: 127712817-127712817 on Build GRCh38
rs779962115 Chr.9: 127712747-127712747 on Build GRCh38
rs747094996 Chr.9: 127712873-127712873 on Build GRCh38
rs1025767068 Chr.9: 127713040-127713040 on Build GRCh38
rs746219770 Chr.9: 127712907-127712907 on Build GRCh38
rs745605379 Chr.9: 127712657-127712657 on Build GRCh38
rs749054064 Chr.9: 127712749-127712749 on Build GRCh38
rs752500714 Chr.9: 127712982-127712982 on Build GRCh38
rs780510350 Chr.9: 127713029-127713029 on Build GRCh38
rs763615745 Chr.9: 127712915-127712915 on Build GRCh38
rs757350739 Chr.9: 127713096-127713096 on Build GRCh38
rs573445451 Chr.9: 127712765-127712765 on Build GRCh38
rs750143880 Chr.9: 127712744-127712744 on Build GRCh38
rs34950677 Chr.9: 127713010-127713011 on Build GRCh38
rs756018572 Chr.9: 127712659-127712659 on Build GRCh38
rs567391916 Chr.9: 127712845-127712845 on Build GRCh38
rs538167990 Chr.9: 127712680-127712680 on Build GRCh38
rs746506232 Chr.9: 127713061-127713061 on Build GRCh38
rs756900403 Chr.9: 127712654-127712654 on Build GRCh38
rs778747226 Chr.9: 127712750-127712750 on Build GRCh38
rs777669320 Chr.9: 127712840-127712840 on Build GRCh38
rs766008254 Chr.9: 127712969-127712969 on Build GRCh38
rs370352312 Chr.9: 127712828-127712828 on Build GRCh38
rs762756206 Chr.9: 127713085-127713085 on Build GRCh38
rs984895158 Chr.9: 127713072-127713072 on Build GRCh38
rs371140658 Chr.9: 127712776-127712776 on Build GRCh38
rs771141348 Chr.9: 127712877-127712877 on Build GRCh38
rs765955028 Chr.9: 127712825-127712825 on Build GRCh38
rs544030250 Chr.9: 127712802-127712802 on Build GRCh38
rs73600061 Chr.9: 127712937-127712937 on Build GRCh38
rs751036602 Chr.9: 127712735-127712735 on Build GRCh38
rs1031337636 Chr.9: 127712955-127712955 on Build GRCh38
rs73600054 Chr.9: 127712653-127712653 on Build GRCh38
rs770282905 Chr.9: 127712804-127712804 on Build GRCh38
rs755971447 Chr.9: 127712746-127712746 on Build GRCh38
rs775224034 Chr.9: 127713083-127713083 on Build GRCh38
rs767555039 Chr.9: 127713101-127713101 on Build GRCh38

View Full Product Details