Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127927129-127927619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP5KL1
Entrez Gene ID
138429
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Gene Aliases
PIPKH, bA203J24.5
UniGene
Hs.734928
Gene Chromosome Location
Chr.9: 127920886-127931958 on Build GRCh38
Transcripts
NM_001135219.1 NM_173492.1 XM_017014279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6182901 c.587G>T p.G196V Chr.9: 127927304-127927304 on Build GRCh38
6746761 c.560-10C>A p.? Chr.9: 127927341-127927341 on Build GRCh38
3847715 c.29G>A p.R10H Chr.9: 127927165-127927165 on Build GRCh38
6746764 c.631G>A p.A211T Chr.9: 127927172-127927172 on Build GRCh38
1151221 c.604A>G p.I202V Chr.9: 127927199-127927199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746840083 Chr.9: 127927256-127927256 on Build GRCh38
rs756618850 Chr.9: 127927226-127927226 on Build GRCh38
rs570227999 Chr.9: 127927436-127927436 on Build GRCh38
rs925781754 Chr.9: 127927177-127927177 on Build GRCh38
rs941943937 Chr.9: 127927447-127927447 on Build GRCh38
rs770666251 Chr.9: 127927258-127927258 on Build GRCh38
rs1023958471 Chr.9: 127927555-127927555 on Build GRCh38
rs929144548 Chr.9: 127927417-127927417 on Build GRCh38
rs900165808 Chr.9: 127927510-127927510 on Build GRCh38
rs994786672 Chr.9: 127927474-127927474 on Build GRCh38
rs13296170 Chr.9: 127927229-127927229 on Build GRCh38
rs376599654 Chr.9: 127927240-127927240 on Build GRCh38
rs536354970 Chr.9: 127927270-127927270 on Build GRCh38
rs935853175 Chr.9: 127927511-127927511 on Build GRCh38
rs377277858 Chr.9: 127927344-127927344 on Build GRCh38
rs572663772 Chr.9: 127927599-127927599 on Build GRCh38
rs781034682 Chr.9: 127927359-127927359 on Build GRCh38
rs762288628 Chr.9: 127927296-127927296 on Build GRCh38
rs933090684 Chr.9: 127927512-127927512 on Build GRCh38
rs538885465 Chr.9: 127927490-127927490 on Build GRCh38
rs748990664 Chr.9: 127927375-127927375 on Build GRCh38
rs369220019 Chr.9: 127927205-127927205 on Build GRCh38
rs764364952 Chr.9: 127927198-127927198 on Build GRCh38
rs1029875487 Chr.9: 127927336-127927336 on Build GRCh38
rs981760203 Chr.9: 127927587-127927587 on Build GRCh38
rs754403835 Chr.9: 127927157-127927157 on Build GRCh38
rs778323835 Chr.9: 127927233-127927233 on Build GRCh38
rs1009700916 Chr.9: 127927348-127927348 on Build GRCh38
rs757890069 Chr.9: 127927341-127927341 on Build GRCh38
rs900527447 Chr.9: 127927365-127927365 on Build GRCh38
rs761149136 Chr.9: 127927308-127927308 on Build GRCh38
rs62585276 Chr.9: 127927514-127927514 on Build GRCh38
rs752242725 Chr.9: 127927236-127927236 on Build GRCh38
rs774673425 Chr.9: 127927286-127927286 on Build GRCh38
rs751265060 Chr.9: 127927351-127927351 on Build GRCh38
rs768167876 Chr.9: 127927217-127927217 on Build GRCh38
rs779933555 Chr.9: 127927213-127927223 on Build GRCh38
rs763031252 Chr.9: 127927285-127927285 on Build GRCh38
rs931957962 Chr.9: 127927368-127927368 on Build GRCh38
rs1004933781 Chr.9: 127927405-127927405 on Build GRCh38
rs750908459 Chr.9: 127927218-127927218 on Build GRCh38
rs969006108 Chr.9: 127927214-127927214 on Build GRCh38
rs745662155 Chr.9: 127927369-127927369 on Build GRCh38
rs778647599 Chr.9: 127927158-127927158 on Build GRCh38
rs983282877 Chr.9: 127927420-127927420 on Build GRCh38
rs769766043 Chr.9: 127927267-127927267 on Build GRCh38
rs372864040 Chr.9: 127927266-127927266 on Build GRCh38
rs771721727 Chr.9: 127927163-127927163 on Build GRCh38
rs374852003 Chr.9: 127927170-127927170 on Build GRCh38
rs959488815 Chr.9: 127927392-127927392 on Build GRCh38
rs768040970 Chr.9: 127927300-127927300 on Build GRCh38
rs752156374 Chr.9: 127927316-127927316 on Build GRCh38
rs562902378 Chr.9: 127927173-127927173 on Build GRCh38
rs547403774 Chr.9: 127927197-127927197 on Build GRCh38
rs756872946 Chr.9: 127927352-127927352 on Build GRCh38
rs968303443 Chr.9: 127927353-127927353 on Build GRCh38
rs890564466 Chr.9: 127927398-127927398 on Build GRCh38
rs887662300 Chr.9: 127927220-127927220 on Build GRCh38
rs765285168 Chr.9: 127927175-127927175 on Build GRCh38
rs748811078 Chr.9: 127927152-127927152 on Build GRCh38
rs770862697 Chr.9: 127927169-127927169 on Build GRCh38
rs973696274 Chr.9: 127927203-127927203 on Build GRCh38
rs762200437 Chr.9: 127927216-127927216 on Build GRCh38
rs747800765 Chr.9: 127927161-127927161 on Build GRCh38
rs992197309 Chr.9: 127927403-127927403 on Build GRCh38
rs1051738570 Chr.9: 127927386-127927386 on Build GRCh38
rs746753736 Chr.9: 127927166-127927166 on Build GRCh38
rs900280406 Chr.9: 127927319-127927319 on Build GRCh38
rs1014769714 Chr.9: 127927415-127927415 on Build GRCh38
rs913355676 Chr.9: 127927324-127927324 on Build GRCh38
rs775610935 Chr.9: 127927181-127927181 on Build GRCh38
rs13295285 Chr.9: 127927424-127927424 on Build GRCh38
rs750783687 Chr.9: 127927303-127927303 on Build GRCh38
rs969245990 Chr.9: 127927580-127927580 on Build GRCh38
rs200658103 Chr.9: 127927252-127927252 on Build GRCh38
rs766892484 Chr.9: 127927311-127927311 on Build GRCh38

View Full Product Details