Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128255981-128256477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
UniGene
Hs.155827
Gene Chromosome Location
Chr.9: 128255829-128276007 on Build GRCh38
Transcripts
NM_004486.4 XM_005251930.1 XM_005251931.1 XM_005251932.1

SNP Detail

SNP ID Chromosome Location
rs930077502 Chr.9: 128256178-128256178 on Build GRCh38
rs546870312 Chr.9: 128256426-128256426 on Build GRCh38
rs559818868 Chr.9: 128256197-128256197 on Build GRCh38
rs2006280 Chr.9: 128256404-128256404 on Build GRCh38
rs888438919 Chr.9: 128256004-128256004 on Build GRCh38
rs1030762180 Chr.9: 128256045-128256045 on Build GRCh38
rs989226953 Chr.9: 128256101-128256101 on Build GRCh38
rs562171959 Chr.9: 128256273-128256273 on Build GRCh38
rs1039602909 Chr.9: 128256203-128256203 on Build GRCh38
rs888482880 Chr.9: 128256198-128256198 on Build GRCh38
rs771768653 Chr.9: 128256372-128256372 on Build GRCh38
rs77168744 Chr.9: 128256263-128256263 on Build GRCh38
rs778456886 Chr.9: 128256310-128256310 on Build GRCh38
rs4734 Chr.9: 128256360-128256360 on Build GRCh38
rs1018143042 Chr.9: 128256016-128256016 on Build GRCh38
rs1050147081 Chr.9: 128256399-128256399 on Build GRCh38
rs185651594 Chr.9: 128256289-128256289 on Build GRCh38
rs918699601 Chr.9: 128256177-128256178 on Build GRCh38
rs879326177 Chr.9: 128256018-128256018 on Build GRCh38
rs372946084 Chr.9: 128256241-128256241 on Build GRCh38
rs956635815 Chr.9: 128256053-128256053 on Build GRCh38
rs531016383 Chr.9: 128256340-128256340 on Build GRCh38
rs1006792985 Chr.9: 128256015-128256015 on Build GRCh38
rs369732072 Chr.9: 128256115-128256115 on Build GRCh38
rs1040050797 Chr.9: 128256418-128256418 on Build GRCh38
rs570949933 Chr.9: 128256380-128256380 on Build GRCh38
rs901067822 Chr.9: 128256218-128256218 on Build GRCh38
rs997939696 Chr.9: 128256031-128256031 on Build GRCh38
rs1048463822 Chr.9: 128256193-128256193 on Build GRCh38
rs944379145 Chr.9: 128256417-128256417 on Build GRCh38
rs960257276 Chr.9: 128256394-128256394 on Build GRCh38
rs1031274865 Chr.9: 128256249-128256249 on Build GRCh38
rs36105229 Chr.9: 128256192-128256193 on Build GRCh38
rs780713566 Chr.9: 128256265-128256265 on Build GRCh38
rs568839585 Chr.9: 128256262-128256262 on Build GRCh38
rs938840447 Chr.9: 128256175-128256175 on Build GRCh38
rs577915688 Chr.9: 128256066-128256066 on Build GRCh38
rs540523350 Chr.9: 128256117-128256117 on Build GRCh38
rs969034938 Chr.9: 128256130-128256130 on Build GRCh38
rs901461055 Chr.9: 128256429-128256429 on Build GRCh38
rs878887008 Chr.9: 128256176-128256177 on Build GRCh38
rs1011106877 Chr.9: 128256267-128256267 on Build GRCh38
rs1006805697 Chr.9: 128256199-128256199 on Build GRCh38
rs557768709 Chr.9: 128256022-128256022 on Build GRCh38
rs1022043171 Chr.9: 128256352-128256352 on Build GRCh38
rs748850269 Chr.9: 128256176-128256176 on Build GRCh38
rs980390343 Chr.9: 128256143-128256143 on Build GRCh38
rs927492017 Chr.9: 128256171-128256172 on Build GRCh38

View Full Product Details