Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128259914-128260420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
UniGene
Hs.155827
Gene Chromosome Location
Chr.9: 128255829-128276007 on Build GRCh38
Transcripts
NM_004486.4 XM_005251930.1 XM_005251931.1 XM_005251932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672502 c.1692G>T p.M564I Chr.9: 128260175-128260175 on Build GRCh38
7311347 c.1723C>T p.H575Y Chr.9: 128260144-128260144 on Build GRCh38
6672501 c.1727T>G p.V576G Chr.9: 128260140-128260140 on Build GRCh38
3904389 c.1771C>T p.L591L Chr.9: 128260096-128260096 on Build GRCh38
381123 c.1755C>G p.G585G Chr.9: 128260112-128260112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376863046 Chr.9: 128260041-128260041 on Build GRCh38
rs199538572 Chr.9: 128260111-128260111 on Build GRCh38
rs369587258 Chr.9: 128260042-128260042 on Build GRCh38
rs200653956 Chr.9: 128260068-128260068 on Build GRCh38
rs368103650 Chr.9: 128260231-128260231 on Build GRCh38
rs535370019 Chr.9: 128260308-128260308 on Build GRCh38
rs767951870 Chr.9: 128260091-128260091 on Build GRCh38
rs142411612 Chr.9: 128260162-128260162 on Build GRCh38
rs780988029 Chr.9: 128260136-128260136 on Build GRCh38
rs769566882 Chr.9: 128260227-128260227 on Build GRCh38
rs201243209 Chr.9: 128260108-128260108 on Build GRCh38
rs905266713 Chr.9: 128260327-128260327 on Build GRCh38
rs567980136 Chr.9: 128260316-128260316 on Build GRCh38
rs1034965776 Chr.9: 128260076-128260076 on Build GRCh38
rs748473256 Chr.9: 128260039-128260039 on Build GRCh38
rs148778498 Chr.9: 128260120-128260120 on Build GRCh38
rs145670991 Chr.9: 128260023-128260023 on Build GRCh38
rs377705334 Chr.9: 128260297-128260297 on Build GRCh38
rs906909079 Chr.9: 128260275-128260275 on Build GRCh38
rs1027792237 Chr.9: 128260317-128260317 on Build GRCh38
rs1003804193 Chr.9: 128259959-128259959 on Build GRCh38
rs550013838 Chr.9: 128260163-128260163 on Build GRCh38
rs1015590799 Chr.9: 128260008-128260008 on Build GRCh38
rs556783958 Chr.9: 128260351-128260351 on Build GRCh38
rs566264413 Chr.9: 128260306-128260306 on Build GRCh38
rs747836734 Chr.9: 128260183-128260183 on Build GRCh38
rs956818908 Chr.9: 128260078-128260078 on Build GRCh38
rs373870855 Chr.9: 128260151-128260151 on Build GRCh38
rs765799898 Chr.9: 128260085-128260105 on Build GRCh38
rs775563471 Chr.9: 128260070-128260070 on Build GRCh38
rs778584663 Chr.9: 128260040-128260040 on Build GRCh38
rs770736942 Chr.9: 128260044-128260044 on Build GRCh38
rs975124711 Chr.9: 128260122-128260122 on Build GRCh38
rs1003526431 Chr.9: 128260386-128260386 on Build GRCh38
rs750765144 Chr.9: 128260100-128260100 on Build GRCh38
rs149575536 Chr.9: 128260293-128260293 on Build GRCh38
rs768763105 Chr.9: 128260152-128260152 on Build GRCh38
rs1036537621 Chr.9: 128260254-128260254 on Build GRCh38
rs541697888 Chr.9: 128260037-128260037 on Build GRCh38
rs767573741 Chr.9: 128260164-128260164 on Build GRCh38
rs989962292 Chr.9: 128260205-128260205 on Build GRCh38
rs776765819 Chr.9: 128260084-128260085 on Build GRCh38
rs940898192 Chr.9: 128259958-128259958 on Build GRCh38
rs770450224 Chr.9: 128260133-128260133 on Build GRCh38
rs759522002 Chr.9: 128260100-128260102 on Build GRCh38
rs746641261 Chr.9: 128260131-128260131 on Build GRCh38
rs750985465 Chr.9: 128260218-128260218 on Build GRCh38
rs757615879 Chr.9: 128260090-128260090 on Build GRCh38
rs775404277 Chr.9: 128260143-128260143 on Build GRCh38
rs759502737 Chr.9: 128260067-128260067 on Build GRCh38
rs761114875 Chr.9: 128260177-128260177 on Build GRCh38
rs777373664 Chr.9: 128260213-128260213 on Build GRCh38
rs751797429 Chr.9: 128260077-128260077 on Build GRCh38
rs752499337 Chr.9: 128260229-128260230 on Build GRCh38
rs745732253 Chr.9: 128260221-128260221 on Build GRCh38
rs754373393 Chr.9: 128260211-128260211 on Build GRCh38
rs112280200 Chr.9: 128259935-128259935 on Build GRCh38
rs537010813 Chr.9: 128260318-128260318 on Build GRCh38
rs182615718 Chr.9: 128260033-128260033 on Build GRCh38
rs940939323 Chr.9: 128260250-128260250 on Build GRCh38
rs764293854 Chr.9: 128260072-128260072 on Build GRCh38
rs776471946 Chr.9: 128260045-128260045 on Build GRCh38
rs751878112 Chr.9: 128260032-128260032 on Build GRCh38
rs756948826 Chr.9: 128260219-128260219 on Build GRCh38
rs374197968 Chr.9: 128260139-128260139 on Build GRCh38
rs757885655 Chr.9: 128260212-128260212 on Build GRCh38
rs765163529 Chr.9: 128260204-128260213 on Build GRCh38
rs555798317 Chr.9: 128260276-128260276 on Build GRCh38
rs757910820 Chr.9: 128260112-128260112 on Build GRCh38
rs555217790 Chr.9: 128260314-128260314 on Build GRCh38
rs367676933 Chr.9: 128260195-128260195 on Build GRCh38
rs199553242 Chr.9: 128260169-128260169 on Build GRCh38
rs899306565 Chr.9: 128260276-128260276 on Build GRCh38
rs745605906 Chr.9: 128260137-128260137 on Build GRCh38
rs756699959 Chr.9: 128260104-128260104 on Build GRCh38

View Full Product Details