Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152991146-152991688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNMA5
Entrez Gene ID
114824
Gene Name
paraneoplastic Ma antigen family member 5
Gene Aliases
-
UniGene
Hs.573567
Gene Chromosome Location
Chr.X: 152988824-152994458 on Build GRCh38
Transcripts
NM_001103150.1 NM_001103151.1 NM_001184924.1 NM_052926.2 XM_017029252.1 XM_017029253.1

SNP Detail

SNP ID Chromosome Location
rs781996754 Chr.X: 152991470-152991470 on Build GRCh38
rs142107812 Chr.X: 152991171-152991171 on Build GRCh38
rs782746336 Chr.X: 152991514-152991514 on Build GRCh38
rs782579115 Chr.X: 152991581-152991581 on Build GRCh38
rs145394099 Chr.X: 152991313-152991313 on Build GRCh38
rs782541189 Chr.X: 152991426-152991426 on Build GRCh38
rs782232196 Chr.X: 152991308-152991308 on Build GRCh38
rs782122744 Chr.X: 152991316-152991316 on Build GRCh38
rs186142178 Chr.X: 152991252-152991252 on Build GRCh38
rs368167670 Chr.X: 152991650-152991650 on Build GRCh38
rs782012013 Chr.X: 152991206-152991206 on Build GRCh38
rs377260067 Chr.X: 152991367-152991367 on Build GRCh38
rs377604389 Chr.X: 152991253-152991253 on Build GRCh38
rs782351130 Chr.X: 152991312-152991312 on Build GRCh38
rs373163204 Chr.X: 152991274-152991274 on Build GRCh38
rs200533836 Chr.X: 152991399-152991399 on Build GRCh38
rs565458820 Chr.X: 152991406-152991406 on Build GRCh38
rs782698342 Chr.X: 152991170-152991170 on Build GRCh38
rs200173729 Chr.X: 152991457-152991457 on Build GRCh38
rs782353282 Chr.X: 152991635-152991635 on Build GRCh38
rs140534157 Chr.X: 152991478-152991478 on Build GRCh38
rs782265083 Chr.X: 152991448-152991448 on Build GRCh38
rs781906071 Chr.X: 152991547-152991547 on Build GRCh38
rs781838282 Chr.X: 152991374-152991374 on Build GRCh38
rs935733166 Chr.X: 152991611-152991611 on Build GRCh38
rs375427411 Chr.X: 152991458-152991458 on Build GRCh38
rs201729365 Chr.X: 152991466-152991466 on Build GRCh38
rs782182073 Chr.X: 152991234-152991234 on Build GRCh38
rs782686495 Chr.X: 152991268-152991268 on Build GRCh38
rs782586977 Chr.X: 152991429-152991429 on Build GRCh38
rs370616990 Chr.X: 152991271-152991271 on Build GRCh38
rs782539708 Chr.X: 152991194-152991194 on Build GRCh38
rs150452691 Chr.X: 152991490-152991490 on Build GRCh38
rs3810655 Chr.X: 152991280-152991280 on Build GRCh38
rs781997942 Chr.X: 152991563-152991563 on Build GRCh38
rs373123354 Chr.X: 152991184-152991184 on Build GRCh38
rs781813438 Chr.X: 152991510-152991510 on Build GRCh38
rs782600363 Chr.X: 152991446-152991446 on Build GRCh38
rs201596972 Chr.X: 152991384-152991384 on Build GRCh38
rs370233185 Chr.X: 152991537-152991537 on Build GRCh38
rs782379919 Chr.X: 152991454-152991454 on Build GRCh38
rs782085755 Chr.X: 152991180-152991180 on Build GRCh38
rs782775107 Chr.X: 152991386-152991386 on Build GRCh38
rs781864018 Chr.X: 152991186-152991186 on Build GRCh38
rs782649229 Chr.X: 152991546-152991546 on Build GRCh38
rs782530783 Chr.X: 152991565-152991565 on Build GRCh38
rs782518669 Chr.X: 152991255-152991255 on Build GRCh38
rs782176702 Chr.X: 152991439-152991439 on Build GRCh38
rs782053915 Chr.X: 152991513-152991513 on Build GRCh38
rs782072627 Chr.X: 152991168-152991168 on Build GRCh38
rs781830252 Chr.X: 152991536-152991536 on Build GRCh38
rs782616725 Chr.X: 152991246-152991246 on Build GRCh38
rs782803270 Chr.X: 152991203-152991203 on Build GRCh38
rs148357143 Chr.X: 152991464-152991464 on Build GRCh38
rs782175884 Chr.X: 152991630-152991630 on Build GRCh38
rs371164310 Chr.X: 152991440-152991440 on Build GRCh38
rs373727785 Chr.X: 152991603-152991603 on Build GRCh38
rs147665095 Chr.X: 152991326-152991326 on Build GRCh38

View Full Product Details