Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 109871688-109872129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
UniGene
Hs.390623
Gene Chromosome Location
Chr.10: 109864766-109923553 on Build GRCh38
Transcripts
NM_001167604.1 NM_001324128.1 NM_001324131.1 NM_001324132.1 NM_001324133.1 NM_001324134.1 NM_001324135.1 NM_001324136.1 NM_020383.3 XM_011540134.1 XM_017016613.1 XM_017016614.1 XM_017016615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940094 c.1378C>T p.P460S Chr.10: 109871807-109871807 on Build GRCh38
7150350 c.1473C>T p.L491L Chr.10: 109871841-109871841 on Build GRCh38
6571211 c.1506C>A p.F502L Chr.10: 109871808-109871808 on Build GRCh38
7150351 c.1344C>T p.L448L Chr.10: 109871841-109871841 on Build GRCh38
5940093 c.1507C>T p.P503S Chr.10: 109871807-109871807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926486329 Chr.10: 109871954-109871954 on Build GRCh38
rs750611175 Chr.10: 109871779-109871779 on Build GRCh38
rs768673256 Chr.10: 109871870-109871870 on Build GRCh38
rs932749008 Chr.10: 109872022-109872022 on Build GRCh38
rs991895698 Chr.10: 109871990-109871990 on Build GRCh38
rs763868159 Chr.10: 109871743-109871743 on Build GRCh38
rs528677335 Chr.10: 109871896-109871896 on Build GRCh38
rs1029749389 Chr.10: 109871813-109871813 on Build GRCh38
rs778391724 Chr.10: 109872083-109872083 on Build GRCh38
rs571658071 Chr.10: 109871979-109871979 on Build GRCh38
rs775066798 Chr.10: 109871864-109871864 on Build GRCh38
rs1023127989 Chr.10: 109871847-109871847 on Build GRCh38
rs887327277 Chr.10: 109871712-109871712 on Build GRCh38
rs1020889742 Chr.10: 109871885-109871885 on Build GRCh38
rs544662225 Chr.10: 109871720-109871721 on Build GRCh38
rs56930482 Chr.10: 109871743-109871744 on Build GRCh38
rs748547667 Chr.10: 109871814-109871814 on Build GRCh38
rs899922607 Chr.10: 109871762-109871762 on Build GRCh38
rs1038609348 Chr.10: 109872014-109872014 on Build GRCh38
rs192070882 Chr.10: 109872077-109872077 on Build GRCh38
rs2274978 Chr.10: 109871907-109871907 on Build GRCh38
rs908793711 Chr.10: 109872004-109872004 on Build GRCh38
rs753190190 Chr.10: 109871966-109871966 on Build GRCh38
rs756995286 Chr.10: 109871763-109871763 on Build GRCh38
rs944498327 Chr.10: 109872053-109872053 on Build GRCh38
rs540737990 Chr.10: 109871991-109871991 on Build GRCh38
rs979299356 Chr.10: 109871948-109871948 on Build GRCh38
rs1005735524 Chr.10: 109871724-109871727 on Build GRCh38
rs772544859 Chr.10: 109871819-109871819 on Build GRCh38
rs971940567 Chr.10: 109871874-109871874 on Build GRCh38
rs774243244 Chr.10: 109871873-109871873 on Build GRCh38
rs7088138 Chr.10: 109871753-109871753 on Build GRCh38
rs1048691036 Chr.10: 109872099-109872099 on Build GRCh38
rs146223771 Chr.10: 109871805-109871805 on Build GRCh38
rs529639517 Chr.10: 109871728-109871728 on Build GRCh38
rs537502602 Chr.10: 109871996-109871996 on Build GRCh38
rs756214586 Chr.10: 109871791-109871791 on Build GRCh38
rs549666377 Chr.10: 109871734-109871734 on Build GRCh38
rs186055369 Chr.10: 109872006-109872006 on Build GRCh38
rs755514571 Chr.10: 109871806-109871806 on Build GRCh38
rs753973262 Chr.10: 109871911-109871911 on Build GRCh38
rs1013543647 Chr.10: 109871827-109871827 on Build GRCh38
rs2274979 Chr.10: 109872023-109872023 on Build GRCh38
rs993150488 Chr.10: 109872094-109872094 on Build GRCh38
rs941575115 Chr.10: 109872008-109872008 on Build GRCh38
rs182030667 Chr.10: 109871770-109871770 on Build GRCh38
rs745515206 Chr.10: 109871824-109871824 on Build GRCh38
rs761647075 Chr.10: 109871876-109871876 on Build GRCh38
rs74154904 Chr.10: 109871744-109871744 on Build GRCh38
rs769250222 Chr.10: 109871850-109871850 on Build GRCh38
rs536174855 Chr.10: 109872044-109872044 on Build GRCh38
rs756943212 Chr.10: 109872071-109872071 on Build GRCh38
rs762324633 Chr.10: 109871869-109871869 on Build GRCh38
rs139214873 Chr.10: 109871823-109871823 on Build GRCh38
rs552824515 Chr.10: 109872058-109872058 on Build GRCh38
rs760955853 Chr.10: 109871906-109871906 on Build GRCh38
rs763148448 Chr.10: 109871811-109871811 on Build GRCh38
rs914683738 Chr.10: 109871997-109871997 on Build GRCh38

View Full Product Details