Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114122690-114123216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

SNP Detail

SNP ID Chromosome Location
rs925177109 Chr.10: 114122767-114122767 on Build GRCh38
rs935229592 Chr.10: 114122952-114122952 on Build GRCh38
rs900007330 Chr.10: 114122908-114122908 on Build GRCh38
rs995943094 Chr.10: 114122995-114122995 on Build GRCh38
rs781775726 Chr.10: 114123098-114123098 on Build GRCh38
rs530549492 Chr.10: 114123068-114123068 on Build GRCh38
rs961960185 Chr.10: 114123080-114123080 on Build GRCh38
rs916295730 Chr.10: 114122745-114122745 on Build GRCh38
rs1028828332 Chr.10: 114123007-114123007 on Build GRCh38
rs1014760198 Chr.10: 114123045-114123045 on Build GRCh38
rs563422760 Chr.10: 114122954-114122954 on Build GRCh38
rs996980015 Chr.10: 114122927-114122927 on Build GRCh38
rs992923632 Chr.10: 114122714-114122714 on Build GRCh38
rs565787888 Chr.10: 114123054-114123054 on Build GRCh38
rs552137468 Chr.10: 114123070-114123070 on Build GRCh38
rs1046059882 Chr.10: 114122806-114122806 on Build GRCh38
rs532092041 Chr.10: 114123061-114123062 on Build GRCh38
rs1004448566 Chr.10: 114122951-114122951 on Build GRCh38
rs913594353 Chr.10: 114123182-114123186 on Build GRCh38
rs893653542 Chr.10: 114122979-114122979 on Build GRCh38
rs931441002 Chr.10: 114122738-114122738 on Build GRCh38
rs936355733 Chr.10: 114122772-114122772 on Build GRCh38
rs913552321 Chr.10: 114122819-114122819 on Build GRCh38
rs957973568 Chr.10: 114123097-114123097 on Build GRCh38
rs980542653 Chr.10: 114123074-114123074 on Build GRCh38
rs1040792371 Chr.10: 114122845-114122845 on Build GRCh38
rs1052080276 Chr.10: 114122793-114122793 on Build GRCh38
rs969210396 Chr.10: 114123071-114123071 on Build GRCh38
rs1032124253 Chr.10: 114123087-114123087 on Build GRCh38
rs1048696524 Chr.10: 114122930-114122930 on Build GRCh38
rs755577361 Chr.10: 114123086-114123086 on Build GRCh38
rs1053576819 Chr.10: 114122955-114122955 on Build GRCh38
rs575096538 Chr.10: 114122830-114122830 on Build GRCh38
rs541794757 Chr.10: 114122713-114122713 on Build GRCh38
rs990588759 Chr.10: 114122740-114122740 on Build GRCh38
rs1008685878 Chr.10: 114123056-114123056 on Build GRCh38
rs564530689 Chr.10: 114123028-114123028 on Build GRCh38
rs943827114 Chr.10: 114122832-114122832 on Build GRCh38
rs563970701 Chr.10: 114123119-114123119 on Build GRCh38
rs1016049804 Chr.10: 114122953-114122953 on Build GRCh38
rs745337494 Chr.10: 114123012-114123012 on Build GRCh38
rs371149522 Chr.10: 114123134-114123134 on Build GRCh38
rs140513403 Chr.10: 114122798-114122798 on Build GRCh38
rs74581721 Chr.10: 114122942-114122942 on Build GRCh38
rs980657282 Chr.10: 114122764-114122764 on Build GRCh38
rs1027218983 Chr.10: 114123124-114123124 on Build GRCh38
rs746010642 Chr.10: 114122968-114122968 on Build GRCh38
rs952862791 Chr.10: 114123160-114123160 on Build GRCh38

View Full Product Details