Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7510834-7511374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFML1
Entrez Gene ID
283298
Gene Name
olfactomedin like 1
Gene Aliases
UNQ564
UniGene
Hs.503500
Gene Chromosome Location
Chr.11: 7485354-7511897 on Build GRCh38
Transcripts
NM_198474.3 XM_005252878.4

SNP Detail

SNP ID Chromosome Location
rs892102470 Chr.11: 7511334-7511334 on Build GRCh38
rs201289454 Chr.11: 7511293-7511293 on Build GRCh38
rs927400772 Chr.11: 7511072-7511072 on Build GRCh38
rs150808367 Chr.11: 7511143-7511143 on Build GRCh38
rs1050549586 Chr.11: 7511242-7511242 on Build GRCh38
rs891681905 Chr.11: 7510896-7510896 on Build GRCh38
rs940908206 Chr.11: 7510869-7510869 on Build GRCh38
rs1025334940 Chr.11: 7511338-7511338 on Build GRCh38
rs988099550 Chr.11: 7511099-7511099 on Build GRCh38
rs902078157 Chr.11: 7511205-7511205 on Build GRCh38
rs1031959904 Chr.11: 7511021-7511021 on Build GRCh38
rs138369701 Chr.11: 7511290-7511293 on Build GRCh38
rs201122757 Chr.11: 7511283-7511284 on Build GRCh38
rs997286718 Chr.11: 7510920-7510920 on Build GRCh38
rs937402665 Chr.11: 7511096-7511096 on Build GRCh38
rs1000535892 Chr.11: 7511257-7511257 on Build GRCh38
rs371800561 Chr.11: 7510987-7510993 on Build GRCh38
rs1024822807 Chr.11: 7511034-7511034 on Build GRCh38
rs534787916 Chr.11: 7511188-7511188 on Build GRCh38
rs75709911 Chr.11: 7511161-7511161 on Build GRCh38
rs1036536245 Chr.11: 7510902-7510902 on Build GRCh38
rs947258771 Chr.11: 7511129-7511129 on Build GRCh38
rs1000526057 Chr.11: 7511015-7511015 on Build GRCh38
rs548914782 Chr.11: 7511119-7511119 on Build GRCh38
rs956656141 Chr.11: 7511023-7511023 on Build GRCh38
rs1009222451 Chr.11: 7511337-7511337 on Build GRCh38
rs1025233435 Chr.11: 7511327-7511327 on Build GRCh38
rs889641573 Chr.11: 7510951-7510951 on Build GRCh38
rs1032009603 Chr.11: 7511289-7511289 on Build GRCh38
rs141330850 Chr.11: 7511067-7511067 on Build GRCh38
rs571335043 Chr.11: 7511251-7511251 on Build GRCh38
rs909167512 Chr.11: 7511116-7511116 on Build GRCh38
rs888844873 Chr.11: 7511001-7511001 on Build GRCh38
rs970668877 Chr.11: 7511344-7511344 on Build GRCh38
rs933513723 Chr.11: 7511233-7511233 on Build GRCh38
rs1011500911 Chr.11: 7510928-7510928 on Build GRCh38
rs533251351 Chr.11: 7510929-7510929 on Build GRCh38
rs531827908 Chr.11: 7510911-7510911 on Build GRCh38
rs970997557 Chr.11: 7511047-7511047 on Build GRCh38
rs1036991246 Chr.11: 7511179-7511179 on Build GRCh38
rs1041186534 Chr.11: 7511202-7511202 on Build GRCh38
rs984400561 Chr.11: 7511039-7511039 on Build GRCh38
rs993882688 Chr.11: 7511316-7511316 on Build GRCh38
rs984800730 Chr.11: 7511097-7511097 on Build GRCh38
rs993830400 Chr.11: 7511004-7511004 on Build GRCh38
rs965014544 Chr.11: 7510999-7510999 on Build GRCh38
rs3884596 Chr.11: 7510944-7510944 on Build GRCh38
rs372517494 Chr.11: 7510960-7510960 on Build GRCh38
rs901986962 Chr.11: 7510917-7510917 on Build GRCh38
rs1019383832 Chr.11: 7510952-7510952 on Build GRCh38
rs148305607 Chr.11: 7511025-7511025 on Build GRCh38
rs915263047 Chr.11: 7511114-7511114 on Build GRCh38
rs551344532 Chr.11: 7511191-7511191 on Build GRCh38

View Full Product Details