Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18701485-18701991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM86A
Entrez Gene ID
144110
Gene Name
transmembrane protein 86A
Gene Aliases
-
UniGene
Hs.502100
Gene Chromosome Location
Chr.11: 18698735-18704785 on Build GRCh38
Transcripts
NM_153347.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838291 c.491G>A p.G164E Chr.11: 18701777-18701777 on Build GRCh38
1353069 c.408C>T p.Y136Y Chr.11: 18701694-18701694 on Build GRCh38
1229751 c.439G>A p.V147M Chr.11: 18701725-18701725 on Build GRCh38
1288514 c.521G>T p.W174L Chr.11: 18701807-18701807 on Build GRCh38
925697 c.675C>T p.V225V Chr.11: 18701961-18701961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552495816 Chr.11: 18701815-18701815 on Build GRCh38
rs367898132 Chr.11: 18701545-18701545 on Build GRCh38
rs762477511 Chr.11: 18701739-18701739 on Build GRCh38
rs772505526 Chr.11: 18701863-18701863 on Build GRCh38
rs755747323 Chr.11: 18701805-18701805 on Build GRCh38
rs143821414 Chr.11: 18701629-18701629 on Build GRCh38
rs770738074 Chr.11: 18701524-18701524 on Build GRCh38
rs148997299 Chr.11: 18701607-18701607 on Build GRCh38
rs368896190 Chr.11: 18701641-18701641 on Build GRCh38
rs375506271 Chr.11: 18701637-18701637 on Build GRCh38
rs763499201 Chr.11: 18701961-18701961 on Build GRCh38
rs752236177 Chr.11: 18701604-18701604 on Build GRCh38
rs781427046 Chr.11: 18701608-18701608 on Build GRCh38
rs766713755 Chr.11: 18701596-18701596 on Build GRCh38
rs150240202 Chr.11: 18701946-18701946 on Build GRCh38
rs960781612 Chr.11: 18701533-18701533 on Build GRCh38
rs372889645 Chr.11: 18701782-18701782 on Build GRCh38
rs150360512 Chr.11: 18701927-18701927 on Build GRCh38
rs200662128 Chr.11: 18701626-18701626 on Build GRCh38
rs772743402 Chr.11: 18701737-18701737 on Build GRCh38
rs778774495 Chr.11: 18701565-18701566 on Build GRCh38
rs897400761 Chr.11: 18701651-18701651 on Build GRCh38
rs934618920 Chr.11: 18701513-18701513 on Build GRCh38
rs777421140 Chr.11: 18701969-18701969 on Build GRCh38
rs768574130 Chr.11: 18701721-18701721 on Build GRCh38
rs765392913 Chr.11: 18701899-18701899 on Build GRCh38
rs776358550 Chr.11: 18701648-18701648 on Build GRCh38
rs760391160 Chr.11: 18701539-18701539 on Build GRCh38
rs372971219 Chr.11: 18701667-18701667 on Build GRCh38
rs267602813 Chr.11: 18701643-18701643 on Build GRCh38
rs758380144 Chr.11: 18701697-18701697 on Build GRCh38
rs758892093 Chr.11: 18701590-18701590 on Build GRCh38
rs752972183 Chr.11: 18701691-18701691 on Build GRCh38
rs768898557 Chr.11: 18701968-18701968 on Build GRCh38
rs777897440 Chr.11: 18701694-18701694 on Build GRCh38
rs768981881 Chr.11: 18701777-18701777 on Build GRCh38
rs914599958 Chr.11: 18701544-18701544 on Build GRCh38
rs370577203 Chr.11: 18701764-18701764 on Build GRCh38
rs1007258438 Chr.11: 18701510-18701510 on Build GRCh38
rs746361715 Chr.11: 18701859-18701859 on Build GRCh38
rs767579895 Chr.11: 18701688-18701688 on Build GRCh38
rs773920751 Chr.11: 18701857-18701857 on Build GRCh38
rs755020443 Chr.11: 18701799-18701799 on Build GRCh38
rs761159778 Chr.11: 18701658-18701658 on Build GRCh38
rs545745130 Chr.11: 18701557-18701557 on Build GRCh38
rs753914264 Chr.11: 18701696-18701696 on Build GRCh38
rs759920120 Chr.11: 18701683-18701683 on Build GRCh38
rs758131502 Chr.11: 18701883-18701885 on Build GRCh38
rs371309512 Chr.11: 18701969-18701969 on Build GRCh38
rs770435089 Chr.11: 18701848-18701848 on Build GRCh38
rs772592122 Chr.11: 18701948-18701948 on Build GRCh38
rs144559255 Chr.11: 18701663-18701663 on Build GRCh38
rs746777053 Chr.11: 18701748-18701748 on Build GRCh38
rs747332216 Chr.11: 18701962-18701962 on Build GRCh38
rs371769220 Chr.11: 18701562-18701562 on Build GRCh38
rs776997469 Chr.11: 18701873-18701873 on Build GRCh38
rs980122968 Chr.11: 18701612-18701612 on Build GRCh38
rs962564654 Chr.11: 18701512-18701512 on Build GRCh38
rs1016833185 Chr.11: 18701511-18701511 on Build GRCh38
rs534042578 Chr.11: 18701783-18701783 on Build GRCh38
rs1050863904 Chr.11: 18701729-18701729 on Build GRCh38
rs766350384 Chr.11: 18701793-18701793 on Build GRCh38
rs146522347 Chr.11: 18701841-18701841 on Build GRCh38
rs747819934 Chr.11: 18701735-18701735 on Build GRCh38
rs753930789 Chr.11: 18701790-18701790 on Build GRCh38
rs745594079 Chr.11: 18701530-18701530 on Build GRCh38
rs201546830 Chr.11: 18701804-18701804 on Build GRCh38
rs376236030 Chr.11: 18701670-18701670 on Build GRCh38
rs748937191 Chr.11: 18701941-18701941 on Build GRCh38
rs376326393 Chr.11: 18701871-18701871 on Build GRCh38
rs781016058 Chr.11: 18701803-18701803 on Build GRCh38
rs199688904 Chr.11: 18701572-18701572 on Build GRCh38
rs142350678 Chr.11: 18701791-18701791 on Build GRCh38
rs141369005 Chr.11: 18701907-18701907 on Build GRCh38
rs779973876 Chr.11: 18701638-18701638 on Build GRCh38
rs778374576 Chr.11: 18701944-18701944 on Build GRCh38
rs376043899 Chr.11: 18701919-18701919 on Build GRCh38
rs763888387 Chr.11: 18701540-18701540 on Build GRCh38
rs199661354 Chr.11: 18701582-18701582 on Build GRCh38
rs117231481 Chr.11: 18701902-18701902 on Build GRCh38
rs775893136 Chr.11: 18701866-18701866 on Build GRCh38
rs976335687 Chr.11: 18701731-18701731 on Build GRCh38
rs189439292 Chr.11: 18701943-18701943 on Build GRCh38
rs774370640 Chr.11: 18701525-18701525 on Build GRCh38
rs766907992 Chr.11: 18701669-18701669 on Build GRCh38
rs751536581 Chr.11: 18701794-18701794 on Build GRCh38
rs527414310 Chr.11: 18701521-18701521 on Build GRCh38
rs755878488 Chr.11: 18701920-18701920 on Build GRCh38
rs7945285 Chr.11: 18701930-18701930 on Build GRCh38
rs377641163 Chr.11: 18701865-18701865 on Build GRCh38
rs751964903 Chr.11: 18701606-18701606 on Build GRCh38
rs756359755 Chr.11: 18701693-18701693 on Build GRCh38
rs760978178 Chr.11: 18701868-18701868 on Build GRCh38
rs780915125 Chr.11: 18701727-18701727 on Build GRCh38
rs771998603 Chr.11: 18701531-18701531 on Build GRCh38
rs775493778 Chr.11: 18701532-18701532 on Build GRCh38
rs756234407 Chr.11: 18701623-18701623 on Build GRCh38
rs746742687 Chr.11: 18701713-18701713 on Build GRCh38
rs373427296 Chr.11: 18701613-18701613 on Build GRCh38
rs748742737 Chr.11: 18701826-18701826 on Build GRCh38
rs368629045 Chr.11: 18701955-18701955 on Build GRCh38
rs139760572 Chr.11: 18701507-18701507 on Build GRCh38
rs760931137 Chr.11: 18701787-18701787 on Build GRCh38
rs960518981 Chr.11: 18701917-18701917 on Build GRCh38
rs146633157 Chr.11: 18701664-18701664 on Build GRCh38
rs948736248 Chr.11: 18701795-18701795 on Build GRCh38
rs762190507 Chr.11: 18701874-18701874 on Build GRCh38
rs780162534 Chr.11: 18701698-18701698 on Build GRCh38
rs759830586 Chr.11: 18701778-18701778 on Build GRCh38

View Full Product Details