Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65852513-65853003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX32
Entrez Gene ID
254122
Gene Name
sorting nexin 32
Gene Aliases
SNX6B
UniGene
Hs.591950
Gene Chromosome Location
Chr.11: 65833636-65854687 on Build GRCh38
Transcripts
NM_152760.2 XM_006718488.3 XM_017017470.1 XM_017017471.1 XM_017017472.1 XM_017017473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6786605 c.1093C>T p.R365C Chr.11: 65852893-65852893 on Build GRCh38
3810089 c.1044C>T p.F348F Chr.11: 65852761-65852761 on Build GRCh38
5051037 c.1056C>A p.S352S Chr.11: 65852773-65852773 on Build GRCh38
3782666 c.925C>T p.R309W Chr.11: 65852642-65852642 on Build GRCh38
930518 c.1112G>A p.R371Q Chr.11: 65852912-65852912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761868971 Chr.11: 65852778-65852780 on Build GRCh38
rs758073277 Chr.11: 65852690-65852690 on Build GRCh38
rs930355996 Chr.11: 65852826-65852826 on Build GRCh38
rs684372 Chr.11: 65852842-65852842 on Build GRCh38
rs929837431 Chr.11: 65852726-65852727 on Build GRCh38
rs988706616 Chr.11: 65852661-65852661 on Build GRCh38
rs772569166 Chr.11: 65852652-65852652 on Build GRCh38
rs776955754 Chr.11: 65852938-65852938 on Build GRCh38
rs138424932 Chr.11: 65852897-65852897 on Build GRCh38
rs144634991 Chr.11: 65852773-65852773 on Build GRCh38
rs765415612 Chr.11: 65852619-65852619 on Build GRCh38
rs763330648 Chr.11: 65852664-65852666 on Build GRCh38
rs773122832 Chr.11: 65852717-65852717 on Build GRCh38
rs764205010 Chr.11: 65852953-65852953 on Build GRCh38
rs902069100 Chr.11: 65852718-65852718 on Build GRCh38
rs767191029 Chr.11: 65852868-65852868 on Build GRCh38
rs17857243 Chr.11: 65852778-65852778 on Build GRCh38
rs971430249 Chr.11: 65852696-65852697 on Build GRCh38
rs192657950 Chr.11: 65852891-65852891 on Build GRCh38
rs746391168 Chr.11: 65852570-65852571 on Build GRCh38
rs746332200 Chr.11: 65852700-65852700 on Build GRCh38
rs756948683 Chr.11: 65852803-65852803 on Build GRCh38
rs751422727 Chr.11: 65852624-65852624 on Build GRCh38
rs562705743 Chr.11: 65852843-65852843 on Build GRCh38
rs779009523 Chr.11: 65852841-65852841 on Build GRCh38
rs556990632 Chr.11: 65852795-65852795 on Build GRCh38
rs771071939 Chr.11: 65852611-65852611 on Build GRCh38
rs376440890 Chr.11: 65852889-65852889 on Build GRCh38
rs771322730 Chr.11: 65852664-65852664 on Build GRCh38
rs770063297 Chr.11: 65852670-65852670 on Build GRCh38
rs566572862 Chr.11: 65852826-65852827 on Build GRCh38
rs768086999 Chr.11: 65852855-65852855 on Build GRCh38
rs768812681 Chr.11: 65852679-65852690 on Build GRCh38
rs758040391 Chr.11: 65852918-65852921 on Build GRCh38
rs757226342 Chr.11: 65852575-65852575 on Build GRCh38
rs201321656 Chr.11: 65852844-65852844 on Build GRCh38
rs1034000291 Chr.11: 65852882-65852882 on Build GRCh38
rs757579112 Chr.11: 65852761-65852761 on Build GRCh38
rs752427548 Chr.11: 65852689-65852689 on Build GRCh38
rs368132118 Chr.11: 65852651-65852651 on Build GRCh38
rs773868232 Chr.11: 65852604-65852604 on Build GRCh38
rs776436853 Chr.11: 65852728-65852728 on Build GRCh38
rs762539744 Chr.11: 65852802-65852802 on Build GRCh38
rs201795634 Chr.11: 65852542-65852542 on Build GRCh38
rs567216254 Chr.11: 65852677-65852677 on Build GRCh38
rs760606808 Chr.11: 65852825-65852826 on Build GRCh38
rs759505703 Chr.11: 65852729-65852729 on Build GRCh38
rs372344292 Chr.11: 65852698-65852698 on Build GRCh38
rs926923353 Chr.11: 65852817-65852817 on Build GRCh38
rs752019876 Chr.11: 65852757-65852757 on Build GRCh38
rs374052223 Chr.11: 65852980-65852980 on Build GRCh38
rs371535733 Chr.11: 65852616-65852616 on Build GRCh38
rs750841685 Chr.11: 65852874-65852874 on Build GRCh38
rs371148171 Chr.11: 65852912-65852912 on Build GRCh38
rs578235979 Chr.11: 65852780-65852780 on Build GRCh38
rs773865054 Chr.11: 65852856-65852856 on Build GRCh38
rs778469177 Chr.11: 65852907-65852908 on Build GRCh38
rs763096986 Chr.11: 65852674-65852674 on Build GRCh38
rs776813322 Chr.11: 65852668-65852668 on Build GRCh38
rs754721665 Chr.11: 65852650-65852650 on Build GRCh38
rs996239519 Chr.11: 65852806-65852806 on Build GRCh38
rs377266615 Chr.11: 65852798-65852798 on Build GRCh38
rs752790193 Chr.11: 65852893-65852893 on Build GRCh38
rs761490442 Chr.11: 65852820-65852820 on Build GRCh38
rs772763399 Chr.11: 65852599-65852599 on Build GRCh38
rs943962510 Chr.11: 65852923-65852924 on Build GRCh38
rs758167032 Chr.11: 65852756-65852756 on Build GRCh38
rs765768834 Chr.11: 65852940-65852940 on Build GRCh38
rs201463949 Chr.11: 65852737-65852737 on Build GRCh38
rs966013845 Chr.11: 65852598-65852598 on Build GRCh38
rs750440058 Chr.11: 65852802-65852803 on Build GRCh38
rs980033938 Chr.11: 65852811-65852811 on Build GRCh38
rs778241458 Chr.11: 65852655-65852655 on Build GRCh38
rs768168830 Chr.11: 65852594-65852594 on Build GRCh38
rs754281950 Chr.11: 65852832-65852832 on Build GRCh38
rs754778652 Chr.11: 65852901-65852903 on Build GRCh38
rs763973559 Chr.11: 65852571-65852571 on Build GRCh38
rs775510483 Chr.11: 65852707-65852707 on Build GRCh38
rs377041103 Chr.11: 65852643-65852643 on Build GRCh38
rs200391009 Chr.11: 65852574-65852574 on Build GRCh38
rs201325063 Chr.11: 65852569-65852569 on Build GRCh38
rs778130693 Chr.11: 65852896-65852896 on Build GRCh38
rs184757711 Chr.11: 65852941-65852941 on Build GRCh38
rs755464290 Chr.11: 65852888-65852888 on Build GRCh38
rs749538202 Chr.11: 65852711-65852711 on Build GRCh38
rs767576060 Chr.11: 65852798-65852799 on Build GRCh38
rs761585911 Chr.11: 65852626-65852626 on Build GRCh38
rs768920081 Chr.11: 65852779-65852779 on Build GRCh38
rs780523734 Chr.11: 65852831-65852831 on Build GRCh38
rs142204103 Chr.11: 65852952-65852952 on Build GRCh38
rs1010812146 Chr.11: 65852981-65852981 on Build GRCh38
rs892518893 Chr.11: 65852908-65852908 on Build GRCh38
rs749207469 Chr.11: 65852910-65852910 on Build GRCh38
rs775706839 Chr.11: 65852671-65852671 on Build GRCh38
rs751180194 Chr.11: 65852819-65852819 on Build GRCh38
rs774604877 Chr.11: 65852748-65852756 on Build GRCh38
rs756566957 Chr.11: 65852766-65852766 on Build GRCh38
rs199522276 Chr.11: 65852669-65852669 on Build GRCh38
rs757508435 Chr.11: 65852962-65852962 on Build GRCh38
rs376064835 Chr.11: 65852584-65852584 on Build GRCh38
rs754549143 Chr.11: 65852687-65852687 on Build GRCh38
rs766660309 Chr.11: 65852881-65852881 on Build GRCh38
rs752390045 Chr.11: 65852912-65852913 on Build GRCh38
rs754373777 Chr.11: 65852885-65852885 on Build GRCh38
rs760369292 Chr.11: 65852725-65852725 on Build GRCh38
rs760277569 Chr.11: 65852927-65852927 on Build GRCh38
rs760796071 Chr.11: 65852541-65852541 on Build GRCh38
rs931977907 Chr.11: 65852942-65852942 on Build GRCh38
rs758615571 Chr.11: 65852894-65852894 on Build GRCh38
rs766218846 Chr.11: 65852685-65852685 on Build GRCh38
rs140929894 Chr.11: 65852642-65852642 on Build GRCh38
rs779957839 Chr.11: 65852907-65852907 on Build GRCh38
rs770359143 Chr.11: 65852580-65852580 on Build GRCh38
rs569746823 Chr.11: 65852726-65852726 on Build GRCh38
rs746408377 Chr.11: 65852765-65852765 on Build GRCh38
rs200733417 Chr.11: 65852663-65852663 on Build GRCh38
rs370489273 Chr.11: 65852699-65852699 on Build GRCh38
rs377126402 Chr.11: 65852911-65852911 on Build GRCh38
rs148372152 Chr.11: 65852959-65852959 on Build GRCh38
rs771421056 Chr.11: 65852925-65852925 on Build GRCh38
rs753595679 Chr.11: 65852862-65852866 on Build GRCh38
rs371760257 Chr.11: 65852570-65852570 on Build GRCh38
rs767246942 Chr.11: 65852641-65852641 on Build GRCh38
rs747575172 Chr.11: 65852657-65852657 on Build GRCh38
rs554513968 Chr.11: 65852797-65852797 on Build GRCh38
rs752233619 Chr.11: 65852739-65852739 on Build GRCh38
rs748514872 Chr.11: 65852595-65852595 on Build GRCh38
rs199720353 Chr.11: 65852585-65852585 on Build GRCh38
rs201282220 Chr.11: 65852645-65852645 on Build GRCh38
rs755979415 Chr.11: 65852904-65852904 on Build GRCh38
rs367671050 Chr.11: 65852741-65852741 on Build GRCh38
rs757215607 Chr.11: 65852646-65852646 on Build GRCh38
rs372404613 Chr.11: 65852878-65852878 on Build GRCh38
rs755013911 Chr.11: 65852834-65852834 on Build GRCh38
rs926951393 Chr.11: 65852678-65852678 on Build GRCh38
rs187612313 Chr.11: 65852864-65852864 on Build GRCh38
rs1050664488 Chr.11: 65852886-65852886 on Build GRCh38
rs769871121 Chr.11: 65852705-65852705 on Build GRCh38
rs764368258 Chr.11: 65852593-65852593 on Build GRCh38
rs766750558 Chr.11: 65852934-65852934 on Build GRCh38
rs759852970 Chr.11: 65852617-65852617 on Build GRCh38
rs764773365 Chr.11: 65852688-65852688 on Build GRCh38
rs760703986 Chr.11: 65852684-65852684 on Build GRCh38
rs375447568 Chr.11: 65852712-65852712 on Build GRCh38
rs777425477 Chr.11: 65852965-65852992 on Build GRCh38
rs749362753 Chr.11: 65852774-65852774 on Build GRCh38
rs776029399 Chr.11: 65852648-65852648 on Build GRCh38
rs375482473 Chr.11: 65852923-65852923 on Build GRCh38
rs761018745 Chr.11: 65852608-65852608 on Build GRCh38
rs750767835 Chr.11: 65852821-65852821 on Build GRCh38
rs762920128 Chr.11: 65852854-65852854 on Build GRCh38
rs368459965 Chr.11: 65852926-65852926 on Build GRCh38
rs763602707 Chr.11: 65852810-65852810 on Build GRCh38
rs1019185846 Chr.11: 65852596-65852596 on Build GRCh38
rs775583362 Chr.11: 65852851-65852851 on Build GRCh38
rs781557193 Chr.11: 65852762-65852762 on Build GRCh38
rs767334940 Chr.11: 65852675-65852675 on Build GRCh38
rs747306445 Chr.11: 65852697-65852697 on Build GRCh38
rs200713547 Chr.11: 65852829-65852829 on Build GRCh38
rs117804536 Chr.11: 65852785-65852785 on Build GRCh38
rs778333031 Chr.11: 65852693-65852693 on Build GRCh38
rs917211310 Chr.11: 65852662-65852662 on Build GRCh38
rs142949971 Chr.11: 65852924-65852924 on Build GRCh38
rs377655599 Chr.11: 65852686-65852686 on Build GRCh38
rs773488083 Chr.11: 65852539-65852539 on Build GRCh38

View Full Product Details