Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8221595-8222155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM90A1
Entrez Gene ID
55138
Gene Name
family with sequence similarity 90 member A1
Gene Aliases
-
UniGene
Hs.196086
Gene Chromosome Location
Chr.12: 8221260-8227618 on Build GRCh38
Transcripts
NM_001319982.1 NM_018088.3 XM_006719097.3 XM_011520719.1 XM_017019547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466044 c.1364C>T p.S455F Chr.12: 8221853-8221853 on Build GRCh38
3872619 c.1190G>A p.R397Q Chr.12: 8222027-8222027 on Build GRCh38
6660322 c.1091C>T p.P364L Chr.12: 8222126-8222126 on Build GRCh38
3872618 c.1272C>T p.L424L Chr.12: 8221945-8221945 on Build GRCh38
943979 c.1203C>T p.N401N Chr.12: 8222014-8222014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146601021 Chr.12: 8222126-8222126 on Build GRCh38
rs781303049 Chr.12: 8221711-8221711 on Build GRCh38
rs757665819 Chr.12: 8221857-8221857 on Build GRCh38
rs753445954 Chr.12: 8221771-8221771 on Build GRCh38
rs759052041 Chr.12: 8222028-8222028 on Build GRCh38
rs770552492 Chr.12: 8222025-8222025 on Build GRCh38
rs755118266 Chr.12: 8222060-8222060 on Build GRCh38
rs138279289 Chr.12: 8221871-8221871 on Build GRCh38
rs759195005 Chr.12: 8221823-8221823 on Build GRCh38
rs757101298 Chr.12: 8222135-8222135 on Build GRCh38
rs768258971 Chr.12: 8221819-8221819 on Build GRCh38
rs779190607 Chr.12: 8221939-8221939 on Build GRCh38
rs778308345 Chr.12: 8221930-8221930 on Build GRCh38
rs775392172 Chr.12: 8221839-8221839 on Build GRCh38
rs762713055 Chr.12: 8221957-8221957 on Build GRCh38
rs749639617 Chr.12: 8221885-8221885 on Build GRCh38
rs370006970 Chr.12: 8221988-8221988 on Build GRCh38
rs756600448 Chr.12: 8221870-8221870 on Build GRCh38
rs779657328 Chr.12: 8221864-8221864 on Build GRCh38
rs757912518 Chr.12: 8221751-8221751 on Build GRCh38
rs9669398 Chr.12: 8222125-8222125 on Build GRCh38
rs748678882 Chr.12: 8221981-8221981 on Build GRCh38
rs776504177 Chr.12: 8222058-8222058 on Build GRCh38
rs770771398 Chr.12: 8222077-8222077 on Build GRCh38
rs781617670 Chr.12: 8221877-8221878 on Build GRCh38
rs758750761 Chr.12: 8222053-8222053 on Build GRCh38
rs902964093 Chr.12: 8221759-8221759 on Build GRCh38
rs781358874 Chr.12: 8222064-8222064 on Build GRCh38
rs748607191 Chr.12: 8221795-8221795 on Build GRCh38
rs749398650 Chr.12: 8221965-8221965 on Build GRCh38
rs773186775 Chr.12: 8222020-8222020 on Build GRCh38
rs753621078 Chr.12: 8221854-8221854 on Build GRCh38
rs761665959 Chr.12: 8221914-8221914 on Build GRCh38
rs942710275 Chr.12: 8221974-8221974 on Build GRCh38
rs755305051 Chr.12: 8221850-8221852 on Build GRCh38
rs200871644 Chr.12: 8221934-8221934 on Build GRCh38
rs552829155 Chr.12: 8221952-8221952 on Build GRCh38
rs373760588 Chr.12: 8222027-8222027 on Build GRCh38
rs975363447 Chr.12: 8221979-8221979 on Build GRCh38
rs199565639 Chr.12: 8221904-8221904 on Build GRCh38
rs779373331 Chr.12: 8221741-8221741 on Build GRCh38
rs759080179 Chr.12: 8221978-8221978 on Build GRCh38
rs3825299 Chr.12: 8221642-8221642 on Build GRCh38
rs754090539 Chr.12: 8221674-8221674 on Build GRCh38
rs77978823 Chr.12: 8222067-8222067 on Build GRCh38
rs770072005 Chr.12: 8221797-8221797 on Build GRCh38
rs778588868 Chr.12: 8221773-8221773 on Build GRCh38
rs746349496 Chr.12: 8221620-8221620 on Build GRCh38
rs148315042 Chr.12: 8221906-8221906 on Build GRCh38
rs893352464 Chr.12: 8221748-8221748 on Build GRCh38
rs747118222 Chr.12: 8221994-8221994 on Build GRCh38
rs773950236 Chr.12: 8222084-8222084 on Build GRCh38
rs751824794 Chr.12: 8222115-8222115 on Build GRCh38
rs759580401 Chr.12: 8221821-8221821 on Build GRCh38
rs750117192 Chr.12: 8221916-8221916 on Build GRCh38
rs909080771 Chr.12: 8221637-8221637 on Build GRCh38
rs779102150 Chr.12: 8221889-8221889 on Build GRCh38
rs143290511 Chr.12: 8221898-8221898 on Build GRCh38
rs147617845 Chr.12: 8221736-8221736 on Build GRCh38
rs778450828 Chr.12: 8222128-8222128 on Build GRCh38
rs11044098 Chr.12: 8221989-8221989 on Build GRCh38
rs773724484 Chr.12: 8222110-8222110 on Build GRCh38
rs749056652 Chr.12: 8222022-8222022 on Build GRCh38
rs773562652 Chr.12: 8221801-8221801 on Build GRCh38
rs750401253 Chr.12: 8222059-8222059 on Build GRCh38
rs755100794 Chr.12: 8222007-8222007 on Build GRCh38
rs746003433 Chr.12: 8221897-8221897 on Build GRCh38
rs376033532 Chr.12: 8221944-8221944 on Build GRCh38
rs766029368 Chr.12: 8221958-8221958 on Build GRCh38
rs781068871 Chr.12: 8222013-8222013 on Build GRCh38
rs757692705 Chr.12: 8221987-8221987 on Build GRCh38
rs771089165 Chr.12: 8221816-8221816 on Build GRCh38
rs756895633 Chr.12: 8221772-8221772 on Build GRCh38
rs368611708 Chr.12: 8222099-8222099 on Build GRCh38
rs756156518 Chr.12: 8222068-8222068 on Build GRCh38
rs774941532 Chr.12: 8222033-8222033 on Build GRCh38
rs145267317 Chr.12: 8221714-8221714 on Build GRCh38
rs760601310 Chr.12: 8221840-8221840 on Build GRCh38
rs745565777 Chr.12: 8222085-8222085 on Build GRCh38
rs1041842400 Chr.12: 8221756-8221756 on Build GRCh38
rs762356892 Chr.12: 8222040-8222040 on Build GRCh38
rs763304740 Chr.12: 8222112-8222112 on Build GRCh38
rs772370997 Chr.12: 8221888-8221888 on Build GRCh38
rs200086837 Chr.12: 8221927-8221927 on Build GRCh38
rs773105433 Chr.12: 8221955-8221955 on Build GRCh38
rs202187715 Chr.12: 8221698-8221698 on Build GRCh38
rs772700171 Chr.12: 8221749-8221749 on Build GRCh38
rs766256508 Chr.12: 8221710-8221710 on Build GRCh38
rs147470293 Chr.12: 8221933-8221933 on Build GRCh38
rs745352083 Chr.12: 8221784-8221784 on Build GRCh38
rs138275793 Chr.12: 8222050-8222050 on Build GRCh38
rs755583607 Chr.12: 8221786-8221786 on Build GRCh38
rs370693616 Chr.12: 8221855-8221855 on Build GRCh38
rs781764226 Chr.12: 8221790-8221790 on Build GRCh38
rs769810148 Chr.12: 8222014-8222014 on Build GRCh38
rs757971003 Chr.12: 8221860-8221860 on Build GRCh38
rs201043502 Chr.12: 8222029-8222029 on Build GRCh38
rs143102996 Chr.12: 8221921-8221921 on Build GRCh38
rs754380818 Chr.12: 8221982-8221982 on Build GRCh38
rs761787607 Chr.12: 8221953-8221953 on Build GRCh38
rs746640938 Chr.12: 8221853-8221853 on Build GRCh38
rs139145613 Chr.12: 8221623-8221623 on Build GRCh38
rs749028733 Chr.12: 8221685-8221685 on Build GRCh38
rs751354017 Chr.12: 8221709-8221709 on Build GRCh38
rs775021555 Chr.12: 8222097-8222097 on Build GRCh38
rs372449661 Chr.12: 8222104-8222104 on Build GRCh38
rs764285653 Chr.12: 8221686-8221686 on Build GRCh38
rs780048473 Chr.12: 8221945-8221945 on Build GRCh38
rs11044099 Chr.12: 8221993-8221993 on Build GRCh38
rs765702576 Chr.12: 8222049-8222049 on Build GRCh38
rs751027178 Chr.12: 8221928-8221928 on Build GRCh38
rs764565725 Chr.12: 8221981-8221981 on Build GRCh38
rs768616228 Chr.12: 8221910-8221910 on Build GRCh38
rs753153521 Chr.12: 8221865-8221865 on Build GRCh38
rs150580216 Chr.12: 8222114-8222114 on Build GRCh38
rs749500454 Chr.12: 8221804-8221804 on Build GRCh38
rs747082840 Chr.12: 8221951-8221951 on Build GRCh38
rs771576074 Chr.12: 8222096-8222096 on Build GRCh38
rs767813589 Chr.12: 8222117-8222117 on Build GRCh38

View Full Product Details