Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131910621-131911121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ULK1
Entrez Gene ID
8408
Gene Name
unc-51 like autophagy activating kinase 1
Gene Aliases
ATG1, ATG1A, UNC51, Unc51.1, hATG1
UniGene
Hs.47061
Gene Chromosome Location
Chr.12: 131894734-131923162 on Build GRCh38
Transcripts
NM_003565.2 XM_011538798.2 XM_011538799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2228879 c.893C>T p.S298L Chr.12: 131910745-131910745 on Build GRCh38
4739152 c.948G>A p.P316P Chr.12: 131910800-131910800 on Build GRCh38
3980919 c.865C>A p.P289T Chr.12: 131910717-131910717 on Build GRCh38
1706464 c.877C>T p.P293S Chr.12: 131910729-131910729 on Build GRCh38
5955537 c.943C>A p.P315T Chr.12: 131910795-131910795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051853717 Chr.12: 131911078-131911078 on Build GRCh38
rs199554361 Chr.12: 131910840-131910840 on Build GRCh38
rs775402283 Chr.12: 131910787-131910787 on Build GRCh38
rs534840424 Chr.12: 131910857-131910857 on Build GRCh38
rs770611096 Chr.12: 131910847-131910847 on Build GRCh38
rs972627414 Chr.12: 131911038-131911038 on Build GRCh38
rs763382915 Chr.12: 131910695-131910695 on Build GRCh38
rs765649041 Chr.12: 131910883-131910883 on Build GRCh38
rs901324702 Chr.12: 131910919-131910919 on Build GRCh38
rs776078607 Chr.12: 131910777-131910777 on Build GRCh38
rs571147302 Chr.12: 131910752-131910752 on Build GRCh38
rs867591923 Chr.12: 131911008-131911008 on Build GRCh38
rs778607264 Chr.12: 131910753-131910754 on Build GRCh38
rs919362427 Chr.12: 131911058-131911058 on Build GRCh38
rs961313595 Chr.12: 131911017-131911017 on Build GRCh38
rs979885193 Chr.12: 131911070-131911070 on Build GRCh38
rs114939754 Chr.12: 131910654-131910654 on Build GRCh38
rs1028791013 Chr.12: 131910968-131910968 on Build GRCh38
rs533414058 Chr.12: 131910772-131910772 on Build GRCh38
rs769057532 Chr.12: 131910753-131910753 on Build GRCh38
rs56364352 Chr.12: 131910745-131910745 on Build GRCh38
rs759014625 Chr.12: 131910780-131910780 on Build GRCh38
rs760166758 Chr.12: 131910702-131910702 on Build GRCh38
rs935634042 Chr.12: 131910874-131910874 on Build GRCh38
rs577879661 Chr.12: 131910864-131910864 on Build GRCh38
rs767820098 Chr.12: 131910799-131910799 on Build GRCh38
rs890944496 Chr.12: 131910881-131910881 on Build GRCh38
rs757464419 Chr.12: 131910797-131910797 on Build GRCh38
rs143119597 Chr.12: 131910719-131910719 on Build GRCh38
rs557619156 Chr.12: 131910862-131910862 on Build GRCh38
rs746321637 Chr.12: 131910686-131910686 on Build GRCh38
rs1025123889 Chr.12: 131910983-131910985 on Build GRCh38
rs762661519 Chr.12: 131910791-131910791 on Build GRCh38
rs774399202 Chr.12: 131910660-131910660 on Build GRCh38
rs771680318 Chr.12: 131910852-131910854 on Build GRCh38
rs763893816 Chr.12: 131910792-131910792 on Build GRCh38
rs759419078 Chr.12: 131910712-131910712 on Build GRCh38
rs200532196 Chr.12: 131910717-131910717 on Build GRCh38
rs569760619 Chr.12: 131910800-131910800 on Build GRCh38
rs753784991 Chr.12: 131910709-131910709 on Build GRCh38
rs371104284 Chr.12: 131910991-131910991 on Build GRCh38
rs926668361 Chr.12: 131910861-131910861 on Build GRCh38
rs970869251 Chr.12: 131911009-131911009 on Build GRCh38
rs570751199 Chr.12: 131910672-131910672 on Build GRCh38
rs899506914 Chr.12: 131910942-131910942 on Build GRCh38
rs942029456 Chr.12: 131910863-131910863 on Build GRCh38
rs770000281 Chr.12: 131910855-131910855 on Build GRCh38
rs542632724 Chr.12: 131910962-131910962 on Build GRCh38
rs963413167 Chr.12: 131911055-131911055 on Build GRCh38
rs200950480 Chr.12: 131911097-131911097 on Build GRCh38
rs780689767 Chr.12: 131910746-131910746 on Build GRCh38
rs1000898244 Chr.12: 131910961-131910961 on Build GRCh38
rs1054566145 Chr.12: 131911083-131911083 on Build GRCh38
rs905542691 Chr.12: 131910873-131910873 on Build GRCh38
rs777925034 Chr.12: 131910722-131910722 on Build GRCh38
rs781766194 Chr.12: 131910667-131910667 on Build GRCh38
rs370624303 Chr.12: 131910720-131910720 on Build GRCh38
rs565801324 Chr.12: 131910853-131910853 on Build GRCh38
rs775959911 Chr.12: 131910693-131910693 on Build GRCh38
rs1054046611 Chr.12: 131910896-131910896 on Build GRCh38
rs751740080 Chr.12: 131910724-131910724 on Build GRCh38
rs552636789 Chr.12: 131910903-131910903 on Build GRCh38
rs772742263 Chr.12: 131910762-131910762 on Build GRCh38
rs756859690 Chr.12: 131910734-131910734 on Build GRCh38
rs943817140 Chr.12: 131911086-131911086 on Build GRCh38
rs1007911075 Chr.12: 131910678-131910678 on Build GRCh38
rs745493050 Chr.12: 131910821-131910821 on Build GRCh38
rs935764209 Chr.12: 131911077-131911077 on Build GRCh38
rs774241521 Chr.12: 131911071-131911071 on Build GRCh38
rs755314212 Chr.12: 131910829-131910829 on Build GRCh38
rs115925722 Chr.12: 131911073-131911073 on Build GRCh38
rs984566949 Chr.12: 131910977-131910977 on Build GRCh38
rs559507880 Chr.12: 131911056-131911056 on Build GRCh38
rs147049737 Chr.12: 131910734-131910734 on Build GRCh38
rs772903689 Chr.12: 131910701-131910701 on Build GRCh38
rs897866967 Chr.12: 131910871-131910871 on Build GRCh38
rs535433315 Chr.12: 131910837-131910837 on Build GRCh38
rs750575465 Chr.12: 131910810-131910810 on Build GRCh38
rs376729575 Chr.12: 131910811-131910811 on Build GRCh38
rs749545110 Chr.12: 131911016-131911016 on Build GRCh38
rs543394579 Chr.12: 131910872-131910872 on Build GRCh38
rs746012135 Chr.12: 131910764-131910764 on Build GRCh38
rs151026522 Chr.12: 131910899-131910899 on Build GRCh38
rs771666475 Chr.12: 131910697-131910697 on Build GRCh38
rs1047307478 Chr.12: 131910880-131910880 on Build GRCh38
rs559468991 Chr.12: 131910726-131910726 on Build GRCh38
rs956895534 Chr.12: 131910748-131910748 on Build GRCh38
rs765217364 Chr.12: 131910784-131910784 on Build GRCh38
rs749740161 Chr.12: 131910826-131910826 on Build GRCh38
rs1009758666 Chr.12: 131910913-131910913 on Build GRCh38
rs770307947 Chr.12: 131910692-131910692 on Build GRCh38
rs550439192 Chr.12: 131910796-131910796 on Build GRCh38
rs535100181 Chr.12: 131911041-131911041 on Build GRCh38
rs1009303872 Chr.12: 131910981-131910981 on Build GRCh38
rs1021099800 Chr.12: 131910914-131910914 on Build GRCh38
rs954073510 Chr.12: 131910971-131910971 on Build GRCh38
rs1021078380 Chr.12: 131910655-131910655 on Build GRCh38

View Full Product Details