Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42415708-42416255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF106
Entrez Gene ID
64397
Gene Name
zinc finger protein 106
Gene Aliases
SH3BP3, ZFP106, ZNF474
UniGene
Hs.511143
Gene Chromosome Location
Chr.15: 42412437-42491197 on Build GRCh38
Transcripts
NM_001284306.1 NM_001284307.1 NM_022473.2 XM_005254591.2 XM_005254592.2 XM_017022483.1 XM_017022484.1 XM_017022485.1

SNP Detail

SNP ID Chromosome Location
rs1142536 Chr.15: 42415882-42415882 on Build GRCh38
rs944854816 Chr.15: 42415978-42415978 on Build GRCh38
rs754802580 Chr.15: 42415807-42415808 on Build GRCh38
rs574508671 Chr.15: 42416036-42416037 on Build GRCh38
rs1062016 Chr.15: 42416028-42416028 on Build GRCh38
rs868790376 Chr.15: 42416086-42416086 on Build GRCh38
rs1031264637 Chr.15: 42415781-42415781 on Build GRCh38
rs903251519 Chr.15: 42416022-42416022 on Build GRCh38
rs774041769 Chr.15: 42416186-42416186 on Build GRCh38
rs150615965 Chr.15: 42415930-42415930 on Build GRCh38
rs528762856 Chr.15: 42415922-42415922 on Build GRCh38
rs986782589 Chr.15: 42415958-42415958 on Build GRCh38
rs948621751 Chr.15: 42415907-42415907 on Build GRCh38
rs577185237 Chr.15: 42415740-42415743 on Build GRCh38
rs142723490 Chr.15: 42415892-42415892 on Build GRCh38
rs758000394 Chr.15: 42415959-42415959 on Build GRCh38
rs904437758 Chr.15: 42415739-42415739 on Build GRCh38
rs940320189 Chr.15: 42415740-42415740 on Build GRCh38
rs776333657 Chr.15: 42416011-42416011 on Build GRCh38
rs905055214 Chr.15: 42416059-42416059 on Build GRCh38
rs961720831 Chr.15: 42415928-42415928 on Build GRCh38
rs537755323 Chr.15: 42415929-42415929 on Build GRCh38
rs948781064 Chr.15: 42416078-42416078 on Build GRCh38
rs966253796 Chr.15: 42416224-42416225 on Build GRCh38
rs777950095 Chr.15: 42415831-42415831 on Build GRCh38
rs1018683762 Chr.15: 42415965-42415965 on Build GRCh38
rs772655966 Chr.15: 42416104-42416104 on Build GRCh38
rs917316302 Chr.15: 42415895-42415895 on Build GRCh38
rs1018630896 Chr.15: 42415815-42415815 on Build GRCh38
rs756718429 Chr.15: 42415830-42415830 on Build GRCh38
rs976181038 Chr.15: 42416224-42416224 on Build GRCh38
rs553210076 Chr.15: 42416027-42416027 on Build GRCh38
rs769748688 Chr.15: 42416093-42416093 on Build GRCh38
rs944845450 Chr.15: 42415814-42415814 on Build GRCh38
rs939986874 Chr.15: 42415891-42415891 on Build GRCh38
rs997026104 Chr.15: 42416023-42416023 on Build GRCh38
rs910763463 Chr.15: 42416210-42416210 on Build GRCh38
rs1040472079 Chr.15: 42415984-42415984 on Build GRCh38
rs927691385 Chr.15: 42415734-42415734 on Build GRCh38
rs1052994607 Chr.15: 42416155-42416155 on Build GRCh38
rs1015843065 Chr.15: 42416045-42416045 on Build GRCh38
rs1033702287 Chr.15: 42415757-42415757 on Build GRCh38
rs373371942 Chr.15: 42415819-42415819 on Build GRCh38
rs567475037 Chr.15: 42415806-42415806 on Build GRCh38
rs889209550 Chr.15: 42415811-42415811 on Build GRCh38
rs575065729 Chr.15: 42416052-42416052 on Build GRCh38
rs768473112 Chr.15: 42415962-42415962 on Build GRCh38
rs746131615 Chr.15: 42415934-42415934 on Build GRCh38
rs765873426 Chr.15: 42416185-42416185 on Build GRCh38
rs984128764 Chr.15: 42415875-42415875 on Build GRCh38
rs546428954 Chr.15: 42415840-42415840 on Build GRCh38
rs920819270 Chr.15: 42415798-42415798 on Build GRCh38
rs554528173 Chr.15: 42416043-42416043 on Build GRCh38
rs972338925 Chr.15: 42415745-42415745 on Build GRCh38
rs190272111 Chr.15: 42416193-42416193 on Build GRCh38
rs114274881 Chr.15: 42416074-42416074 on Build GRCh38
rs981908314 Chr.15: 42415899-42415906 on Build GRCh38
rs1010863162 Chr.15: 42416041-42416041 on Build GRCh38
rs903218880 Chr.15: 42415829-42415829 on Build GRCh38
rs992827671 Chr.15: 42416071-42416071 on Build GRCh38
rs891705471 Chr.15: 42416157-42416157 on Build GRCh38
rs115073720 Chr.15: 42416122-42416122 on Build GRCh38
rs762455063 Chr.15: 42416133-42416133 on Build GRCh38
rs1139100 Chr.15: 42415807-42415807 on Build GRCh38
rs866858824 Chr.15: 42415920-42415920 on Build GRCh38
rs1040403982 Chr.15: 42416202-42416202 on Build GRCh38
rs904447264 Chr.15: 42415921-42415921 on Build GRCh38
rs760065341 Chr.15: 42415740-42415742 on Build GRCh38
rs139959105 Chr.15: 42416201-42416201 on Build GRCh38
rs1028611923 Chr.15: 42415839-42415839 on Build GRCh38
rs930878953 Chr.15: 42415957-42415957 on Build GRCh38
rs999868367 Chr.15: 42415773-42415773 on Build GRCh38
rs548532680 Chr.15: 42415744-42415744 on Build GRCh38
rs138203640 Chr.15: 42415847-42415847 on Build GRCh38

View Full Product Details