Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43203015-43203510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB42
Entrez Gene ID
2038
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
PA, SPH5
UniGene
Hs.368642
Gene Chromosome Location
Chr.15: 43197227-43225856 on Build GRCh38
Transcripts
NM_000119.2 NM_001114134.1 XM_005254225.1 XM_011521349.2 XM_011521350.2 XM_011521351.2 XM_011521352.2 XM_011521353.2 XM_011521354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
961899 c.1750C>T p.R584* Chr.15: 43203234-43203234 on Build GRCh38
3667722 c.1857C>A p.H619Q Chr.15: 43203127-43203127 on Build GRCh38
6214101 c.1724T>C p.I575T Chr.15: 43203260-43203260 on Build GRCh38
6258539 c.1734C>A p.F578L Chr.15: 43203250-43203250 on Build GRCh38
4471745 c.1739C>T p.S580F Chr.15: 43203245-43203245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780120207 Chr.15: 43203064-43203064 on Build GRCh38
rs775129311 Chr.15: 43203112-43203112 on Build GRCh38
rs774017319 Chr.15: 43203086-43203086 on Build GRCh38
rs765829996 Chr.15: 43203234-43203234 on Build GRCh38
rs771851031 Chr.15: 43203221-43203221 on Build GRCh38
rs75424023 Chr.15: 43203222-43203222 on Build GRCh38
rs759843947 Chr.15: 43203076-43203077 on Build GRCh38
rs960007262 Chr.15: 43203224-43203224 on Build GRCh38
rs374148780 Chr.15: 43203295-43203295 on Build GRCh38
rs199778021 Chr.15: 43203208-43203208 on Build GRCh38
rs766564564 Chr.15: 43203244-43203244 on Build GRCh38
rs779026751 Chr.15: 43203188-43203188 on Build GRCh38
rs777126804 Chr.15: 43203227-43203227 on Build GRCh38
rs377666792 Chr.15: 43203301-43203301 on Build GRCh38
rs531690568 Chr.15: 43203320-43203320 on Build GRCh38
rs992710518 Chr.15: 43203063-43203063 on Build GRCh38
rs1050326214 Chr.15: 43203175-43203175 on Build GRCh38
rs1040583633 Chr.15: 43203132-43203132 on Build GRCh38
rs769717972 Chr.15: 43203072-43203072 on Build GRCh38
rs115014100 Chr.15: 43203242-43203242 on Build GRCh38
rs754319779 Chr.15: 43203150-43203150 on Build GRCh38
rs375076769 Chr.15: 43203250-43203250 on Build GRCh38
rs775989765 Chr.15: 43203298-43203298 on Build GRCh38
rs933341387 Chr.15: 43203453-43203453 on Build GRCh38
rs775832353 Chr.15: 43203212-43203212 on Build GRCh38
rs200770579 Chr.15: 43203108-43203108 on Build GRCh38
rs887665023 Chr.15: 43203461-43203461 on Build GRCh38
rs780270544 Chr.15: 43203373-43203373 on Build GRCh38
rs150037117 Chr.15: 43203233-43203233 on Build GRCh38
rs762194931 Chr.15: 43203151-43203151 on Build GRCh38
rs957083546 Chr.15: 43203345-43203345 on Build GRCh38
rs568533828 Chr.15: 43203348-43203348 on Build GRCh38
rs1036736399 Chr.15: 43203468-43203468 on Build GRCh38
rs758551627 Chr.15: 43203372-43203372 on Build GRCh38
rs745717736 Chr.15: 43203215-43203215 on Build GRCh38
rs201441200 Chr.15: 43203126-43203126 on Build GRCh38
rs548711188 Chr.15: 43203341-43203341 on Build GRCh38
rs912994169 Chr.15: 43203362-43203362 on Build GRCh38
rs770169812 Chr.15: 43203314-43203314 on Build GRCh38
rs139490049 Chr.15: 43203178-43203178 on Build GRCh38
rs552637754 Chr.15: 43203263-43203263 on Build GRCh38
rs935660366 Chr.15: 43203102-43203102 on Build GRCh38
rs373815420 Chr.15: 43203084-43203084 on Build GRCh38
rs923132760 Chr.15: 43203413-43203413 on Build GRCh38
rs372410152 Chr.15: 43203196-43203196 on Build GRCh38
rs767501910 Chr.15: 43203256-43203256 on Build GRCh38
rs931789597 Chr.15: 43203414-43203414 on Build GRCh38
rs759268312 Chr.15: 43203105-43203105 on Build GRCh38
rs762724018 Chr.15: 43203335-43203335 on Build GRCh38
rs767976512 Chr.15: 43203095-43203096 on Build GRCh38
rs746661087 Chr.15: 43203294-43203294 on Build GRCh38
rs369068948 Chr.15: 43203258-43203258 on Build GRCh38
rs755099340 Chr.15: 43203182-43203182 on Build GRCh38
rs942053350 Chr.15: 43203465-43203465 on Build GRCh38
rs777611580 Chr.15: 43203075-43203075 on Build GRCh38
rs376680136 Chr.15: 43203198-43203198 on Build GRCh38
rs749088636 Chr.15: 43203083-43203083 on Build GRCh38
rs969579987 Chr.15: 43203071-43203071 on Build GRCh38
rs751606866 Chr.15: 43203180-43203180 on Build GRCh38
rs377523204 Chr.15: 43203275-43203275 on Build GRCh38
rs71476423 Chr.15: 43203438-43203438 on Build GRCh38
rs750643111 Chr.15: 43203357-43203357 on Build GRCh38
rs757258460 Chr.15: 43203145-43203145 on Build GRCh38
rs764634978 Chr.15: 43203134-43203134 on Build GRCh38
rs115998465 Chr.15: 43203237-43203237 on Build GRCh38
rs747147980 Chr.15: 43203066-43203066 on Build GRCh38
rs76268277 Chr.15: 43203201-43203201 on Build GRCh38
rs1053809121 Chr.15: 43203137-43203137 on Build GRCh38
rs901762206 Chr.15: 43203147-43203147 on Build GRCh38
rs201925969 Chr.15: 43203325-43203325 on Build GRCh38
rs750584520 Chr.15: 43203158-43203158 on Build GRCh38
rs371132700 Chr.15: 43203285-43203285 on Build GRCh38
rs534175930 Chr.15: 43203419-43203419 on Build GRCh38
rs745630037 Chr.15: 43203266-43203266 on Build GRCh38
rs761308419 Chr.15: 43203285-43203286 on Build GRCh38
rs145043898 Chr.15: 43203199-43203199 on Build GRCh38
rs773757281 Chr.15: 43203318-43203318 on Build GRCh38
rs1040143091 Chr.15: 43203412-43203412 on Build GRCh38
rs114345862 Chr.15: 43203223-43203223 on Build GRCh38
rs564320392 Chr.15: 43203259-43203259 on Build GRCh38
rs765521438 Chr.15: 43203156-43203156 on Build GRCh38
rs1050357464 Chr.15: 43203440-43203440 on Build GRCh38
rs114354377 Chr.15: 43203127-43203127 on Build GRCh38
rs751803200 Chr.15: 43203246-43203246 on Build GRCh38
rs1006614184 Chr.15: 43203218-43203218 on Build GRCh38
rs758603689 Chr.15: 43203172-43203172 on Build GRCh38

View Full Product Details