Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40699907-40700409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT24
Entrez Gene ID
192666
Gene Name
keratin 24
Gene Aliases
K24, KA24
UniGene
Hs.87383
Gene Chromosome Location
Chr.17: 40694246-40703750 on Build GRCh38
Transcripts
NM_019016.2 XM_006721739.3 XM_011524460.2 XM_017024299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1212806 c.956C>T p.A319V Chr.17: 40700283-40700283 on Build GRCh38
302516 c.898G>A p.V300I Chr.17: 40700341-40700341 on Build GRCh38
6697934 c.1004G>A p.R335Q Chr.17: 40700235-40700235 on Build GRCh38
3983419 c.881C>G p.S294C Chr.17: 40700358-40700358 on Build GRCh38
6146621 c.957G>T p.A319A Chr.17: 40700282-40700282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186788337 Chr.17: 40700344-40700344 on Build GRCh38
rs909612861 Chr.17: 40700176-40700176 on Build GRCh38
rs753804541 Chr.17: 40700132-40700132 on Build GRCh38
rs140665011 Chr.17: 40700044-40700044 on Build GRCh38
rs959920280 Chr.17: 40700268-40700268 on Build GRCh38
rs201880280 Chr.17: 40700174-40700174 on Build GRCh38
rs747242596 Chr.17: 40700156-40700156 on Build GRCh38
rs750253983 Chr.17: 40700121-40700121 on Build GRCh38
rs373917694 Chr.17: 40700118-40700118 on Build GRCh38
rs749766317 Chr.17: 40700185-40700185 on Build GRCh38
rs74542796 Chr.17: 40700256-40700256 on Build GRCh38
rs776478694 Chr.17: 40700282-40700282 on Build GRCh38
rs766255140 Chr.17: 40700131-40700131 on Build GRCh38
rs998069317 Chr.17: 40700217-40700217 on Build GRCh38
rs150081982 Chr.17: 40700235-40700235 on Build GRCh38
rs754199352 Chr.17: 40700325-40700325 on Build GRCh38
rs564578139 Chr.17: 40700039-40700039 on Build GRCh38
rs16966138 Chr.17: 40700045-40700045 on Build GRCh38
rs763859365 Chr.17: 40700009-40700009 on Build GRCh38
rs979445396 Chr.17: 40700313-40700313 on Build GRCh38
rs751196832 Chr.17: 40700011-40700011 on Build GRCh38
rs779246628 Chr.17: 40700335-40700335 on Build GRCh38
rs778014038 Chr.17: 40700050-40700050 on Build GRCh38
rs761492060 Chr.17: 40700203-40700203 on Build GRCh38
rs767226342 Chr.17: 40700029-40700029 on Build GRCh38
rs79328812 Chr.17: 40700259-40700259 on Build GRCh38
rs756483222 Chr.17: 40700241-40700241 on Build GRCh38
rs778315563 Chr.17: 40700158-40700158 on Build GRCh38
rs756019131 Chr.17: 40700128-40700128 on Build GRCh38
rs1053081601 Chr.17: 40700193-40700193 on Build GRCh38
rs961667164 Chr.17: 40700254-40700254 on Build GRCh38
rs751630576 Chr.17: 40700234-40700234 on Build GRCh38
rs763546162 Chr.17: 40700088-40700088 on Build GRCh38
rs376820063 Chr.17: 40700370-40700370 on Build GRCh38
rs765136319 Chr.17: 40700301-40700301 on Build GRCh38
rs376890305 Chr.17: 40700120-40700120 on Build GRCh38
rs764232150 Chr.17: 40700225-40700225 on Build GRCh38
rs762664016 Chr.17: 40700004-40700004 on Build GRCh38
rs181321605 Chr.17: 40700112-40700112 on Build GRCh38
rs773427084 Chr.17: 40699964-40699964 on Build GRCh38
rs750618241 Chr.17: 40700318-40700318 on Build GRCh38
rs138394829 Chr.17: 40700341-40700341 on Build GRCh38
rs575550470 Chr.17: 40700350-40700350 on Build GRCh38
rs201002972 Chr.17: 40700250-40700250 on Build GRCh38
rs777127010 Chr.17: 40699974-40699974 on Build GRCh38
rs948135062 Chr.17: 40700312-40700312 on Build GRCh38
rs539260389 Chr.17: 40700016-40700016 on Build GRCh38
rs372772790 Chr.17: 40700052-40700052 on Build GRCh38
rs770308152 Chr.17: 40700175-40700175 on Build GRCh38
rs762507336 Chr.17: 40700084-40700084 on Build GRCh38
rs200473377 Chr.17: 40700126-40700126 on Build GRCh38
rs753715066 Chr.17: 40700222-40700222 on Build GRCh38
rs367620848 Chr.17: 40700219-40700219 on Build GRCh38
rs759487802 Chr.17: 40700283-40700283 on Build GRCh38
rs201916933 Chr.17: 40700275-40700275 on Build GRCh38
rs772317576 Chr.17: 40699962-40699962 on Build GRCh38
rs916701749 Chr.17: 40700296-40700296 on Build GRCh38
rs1028765447 Chr.17: 40700230-40700230 on Build GRCh38
rs770698814 Chr.17: 40700280-40700280 on Build GRCh38
rs776464675 Chr.17: 40700369-40700369 on Build GRCh38
rs755388730 Chr.17: 40700326-40700326 on Build GRCh38
rs757490372 Chr.17: 40700160-40700160 on Build GRCh38
rs773797299 Chr.17: 40700200-40700200 on Build GRCh38
rs972015572 Chr.17: 40700255-40700255 on Build GRCh38
rs1029722649 Chr.17: 40700155-40700155 on Build GRCh38
rs756465261 Chr.17: 40700320-40700320 on Build GRCh38
rs770465291 Chr.17: 40700073-40700073 on Build GRCh38
rs760135405 Chr.17: 40699997-40699997 on Build GRCh38
rs779018759 Chr.17: 40699947-40699947 on Build GRCh38
rs147665347 Chr.17: 40700255-40700256 on Build GRCh38
rs780532514 Chr.17: 40700324-40700324 on Build GRCh38
rs750345581 Chr.17: 40700032-40700032 on Build GRCh38
rs777726427 Chr.17: 40700137-40700137 on Build GRCh38
rs775121232 Chr.17: 40700078-40700078 on Build GRCh38
rs530427640 Chr.17: 40700136-40700136 on Build GRCh38
rs376788689 Chr.17: 40700276-40700276 on Build GRCh38
rs778459686 Chr.17: 40700253-40700253 on Build GRCh38
rs935586941 Chr.17: 40700189-40700189 on Build GRCh38
rs766164927 Chr.17: 40700221-40700221 on Build GRCh38
rs776919542 Chr.17: 40700059-40700059 on Build GRCh38
rs199620916 Chr.17: 40700267-40700267 on Build GRCh38
rs370798217 Chr.17: 40700236-40700236 on Build GRCh38
rs781666931 Chr.17: 40700170-40700170 on Build GRCh38
rs374391631 Chr.17: 40700265-40700265 on Build GRCh38
rs770612859 Chr.17: 40700364-40700364 on Build GRCh38
rs374585479 Chr.17: 40699949-40699949 on Build GRCh38
rs373782671 Chr.17: 40700213-40700213 on Build GRCh38
rs769664866 Chr.17: 40700377-40700377 on Build GRCh38
rs768158213 Chr.17: 40700195-40700195 on Build GRCh38
rs775207016 Chr.17: 40699999-40699999 on Build GRCh38
rs878997392 Chr.17: 40699992-40699992 on Build GRCh38
rs375745897 Chr.17: 40699998-40699998 on Build GRCh38
rs141461077 Chr.17: 40700100-40700100 on Build GRCh38
rs112898776 Chr.17: 40700214-40700214 on Build GRCh38
rs370056385 Chr.17: 40699967-40699967 on Build GRCh38
rs778500545 Chr.17: 40700051-40700051 on Build GRCh38
rs759451226 Chr.17: 40700223-40700223 on Build GRCh38
rs746661881 Chr.17: 40700360-40700360 on Build GRCh38
rs752704369 Chr.17: 40700314-40700314 on Build GRCh38
rs761050874 Chr.17: 40699965-40699965 on Build GRCh38
rs190018370 Chr.17: 40700049-40700049 on Build GRCh38
rs376325577 Chr.17: 40700055-40700055 on Build GRCh38
rs776459239 Chr.17: 40700130-40700131 on Build GRCh38
rs771600549 Chr.17: 40700263-40700263 on Build GRCh38
rs928491720 Chr.17: 40699987-40699987 on Build GRCh38
rs200612551 Chr.17: 40700063-40700063 on Build GRCh38
rs151290359 Chr.17: 40700043-40700043 on Build GRCh38
rs201003553 Chr.17: 40700096-40700096 on Build GRCh38
rs776104765 Chr.17: 40700180-40700180 on Build GRCh38
rs761762639 Chr.17: 40700315-40700315 on Build GRCh38
rs200097414 Chr.17: 40700348-40700348 on Build GRCh38
rs771668201 Chr.17: 40700206-40700206 on Build GRCh38
rs749795235 Chr.17: 40700251-40700251 on Build GRCh38

View Full Product Details