Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59833213-59833732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK1
Entrez Gene ID
51361
Gene Name
hook microtubule-tethering protein 1
Gene Aliases
HK1
UniGene
Hs.378836
Gene Chromosome Location
Chr.1: 59814861-59876378 on Build GRCh38
Transcripts
NM_015888.4 XM_006710676.1 XM_011541562.2 XM_011541563.1 XM_017001424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7091397 c.328G>A p.E110K Chr.1: 59833459-59833459 on Build GRCh38
4009135 c.303T>C p.L101L Chr.1: 59833434-59833434 on Build GRCh38
4690937 c.347A>G p.E116G Chr.1: 59833478-59833478 on Build GRCh38
1637794 c.319C>A p.Q107K Chr.1: 59833450-59833450 on Build GRCh38
6540477 c.335C>T p.S112L Chr.1: 59833466-59833466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746157591 Chr.1: 59833485-59833485 on Build GRCh38
rs985013518 Chr.1: 59833704-59833704 on Build GRCh38
rs911078588 Chr.1: 59833551-59833551 on Build GRCh38
rs966519046 Chr.1: 59833261-59833261 on Build GRCh38
rs1022470068 Chr.1: 59833251-59833251 on Build GRCh38
rs965704983 Chr.1: 59833666-59833666 on Build GRCh38
rs905370762 Chr.1: 59833702-59833702 on Build GRCh38
rs749402766 Chr.1: 59833515-59833515 on Build GRCh38
rs375139877 Chr.1: 59833349-59833349 on Build GRCh38
rs377010469 Chr.1: 59833577-59833577 on Build GRCh38
rs1010614553 Chr.1: 59833244-59833244 on Build GRCh38
rs769186324 Chr.1: 59833375-59833375 on Build GRCh38
rs772547410 Chr.1: 59833569-59833569 on Build GRCh38
rs151210206 Chr.1: 59833495-59833495 on Build GRCh38
rs968473346 Chr.1: 59833512-59833512 on Build GRCh38
rs764119596 Chr.1: 59833457-59833457 on Build GRCh38
rs768579712 Chr.1: 59833520-59833520 on Build GRCh38
rs759178002 Chr.1: 59833431-59833431 on Build GRCh38
rs775706227 Chr.1: 59833371-59833371 on Build GRCh38
rs768136356 Chr.1: 59833409-59833409 on Build GRCh38
rs374844840 Chr.1: 59833558-59833558 on Build GRCh38
rs929971315 Chr.1: 59833607-59833607 on Build GRCh38
rs781479752 Chr.1: 59833362-59833362 on Build GRCh38
rs907227675 Chr.1: 59833479-59833479 on Build GRCh38
rs375815725 Chr.1: 59833382-59833382 on Build GRCh38
rs907950248 Chr.1: 59833398-59833398 on Build GRCh38
rs115210278 Chr.1: 59833687-59833687 on Build GRCh38
rs905424437 Chr.1: 59833459-59833459 on Build GRCh38
rs547895157 Chr.1: 59833700-59833700 on Build GRCh38
rs959992069 Chr.1: 59833489-59833489 on Build GRCh38
rs767710740 Chr.1: 59833414-59833414 on Build GRCh38
rs1047610858 Chr.1: 59833380-59833380 on Build GRCh38
rs76651308 Chr.1: 59833332-59833332 on Build GRCh38
rs974676598 Chr.1: 59833335-59833335 on Build GRCh38
rs759233010 Chr.1: 59833580-59833580 on Build GRCh38
rs112094734 Chr.1: 59833686-59833686 on Build GRCh38
rs764889448 Chr.1: 59833587-59833587 on Build GRCh38
rs1020490478 Chr.1: 59833593-59833593 on Build GRCh38
rs41312662 Chr.1: 59833453-59833453 on Build GRCh38
rs909963592 Chr.1: 59833638-59833638 on Build GRCh38
rs757088201 Chr.1: 59833473-59833473 on Build GRCh38
rs532538600 Chr.1: 59833691-59833691 on Build GRCh38
rs751382629 Chr.1: 59833464-59833464 on Build GRCh38
rs556232811 Chr.1: 59833311-59833311 on Build GRCh38
rs999311446 Chr.1: 59833621-59833621 on Build GRCh38
rs181332350 Chr.1: 59833274-59833274 on Build GRCh38
rs74649729 Chr.1: 59833333-59833333 on Build GRCh38
rs72921691 Chr.1: 59833366-59833366 on Build GRCh38
rs764797480 Chr.1: 59833432-59833432 on Build GRCh38
rs780835674 Chr.1: 59833480-59833480 on Build GRCh38
rs760936570 Chr.1: 59833590-59833590 on Build GRCh38
rs902241983 Chr.1: 59833601-59833601 on Build GRCh38
rs760237276 Chr.1: 59833388-59833388 on Build GRCh38
rs201087676 Chr.1: 59833438-59833438 on Build GRCh38
rs542543055 Chr.1: 59833367-59833367 on Build GRCh38
rs1047109588 Chr.1: 59833627-59833627 on Build GRCh38
rs780332318 Chr.1: 59833498-59833498 on Build GRCh38
rs921724380 Chr.1: 59833339-59833339 on Build GRCh38
rs978613285 Chr.1: 59833285-59833285 on Build GRCh38
rs764062652 Chr.1: 59833555-59833555 on Build GRCh38
rs776364680 Chr.1: 59833418-59833418 on Build GRCh38
rs553593495 Chr.1: 59833344-59833344 on Build GRCh38
rs1018592610 Chr.1: 59833650-59833650 on Build GRCh38
rs774930150 Chr.1: 59833554-59833554 on Build GRCh38
rs753330033 Chr.1: 59833649-59833649 on Build GRCh38
rs372826449 Chr.1: 59833482-59833482 on Build GRCh38
rs749853289 Chr.1: 59833374-59833374 on Build GRCh38

View Full Product Details