Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64602641-64603062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6617296 c.460C>T p.R154* Chr.1: 64602855-64602855 on Build GRCh38
6617277 c.484C>T p.R162W Chr.1: 64602879-64602879 on Build GRCh38
6449815 c.485G>A p.R162Q Chr.1: 64602880-64602880 on Build GRCh38
6451936 c.503G>A p.R168Q Chr.1: 64602898-64602898 on Build GRCh38
6617281 c.461G>A p.R154Q Chr.1: 64602856-64602856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773192353 Chr.1: 64602780-64602780 on Build GRCh38
rs559035954 Chr.1: 64602709-64602709 on Build GRCh38
rs934834207 Chr.1: 64602842-64602842 on Build GRCh38
rs929783401 Chr.1: 64602822-64602822 on Build GRCh38
rs780741848 Chr.1: 64602791-64602791 on Build GRCh38
rs1055140432 Chr.1: 64602846-64602846 on Build GRCh38
rs757886843 Chr.1: 64602764-64602764 on Build GRCh38
rs369437288 Chr.1: 64602893-64602893 on Build GRCh38
rs1027348757 Chr.1: 64602988-64602988 on Build GRCh38
rs867681376 Chr.1: 64602771-64602771 on Build GRCh38
rs144100875 Chr.1: 64602975-64602975 on Build GRCh38
rs774620530 Chr.1: 64602835-64602835 on Build GRCh38
rs1003827165 Chr.1: 64602936-64602936 on Build GRCh38
rs780870638 Chr.1: 64602978-64602978 on Build GRCh38
rs6665437 Chr.1: 64602955-64602955 on Build GRCh38
rs751042895 Chr.1: 64602881-64602881 on Build GRCh38
rs372693053 Chr.1: 64602783-64602783 on Build GRCh38
rs1026884179 Chr.1: 64602710-64602710 on Build GRCh38
rs746179035 Chr.1: 64602802-64602802 on Build GRCh38
rs989600710 Chr.1: 64602987-64602987 on Build GRCh38
rs761819776 Chr.1: 64602855-64602855 on Build GRCh38
rs897016479 Chr.1: 64602815-64602815 on Build GRCh38
rs1054556 Chr.1: 64602880-64602880 on Build GRCh38
rs59926290 Chr.1: 64602924-64602924 on Build GRCh38
rs536404538 Chr.1: 64602989-64602989 on Build GRCh38
rs550241032 Chr.1: 64602925-64602925 on Build GRCh38
rs770985910 Chr.1: 64602957-64602957 on Build GRCh38
rs756726292 Chr.1: 64602883-64602883 on Build GRCh38
rs991496993 Chr.1: 64602732-64602732 on Build GRCh38
rs754776559 Chr.1: 64602964-64602964 on Build GRCh38
rs59976163 Chr.1: 64603023-64603023 on Build GRCh38
rs762999785 Chr.1: 64602818-64602819 on Build GRCh38
rs911911887 Chr.1: 64603028-64603028 on Build GRCh38
rs764044897 Chr.1: 64602879-64602879 on Build GRCh38
rs761632257 Chr.1: 64602754-64602754 on Build GRCh38
rs1041333180 Chr.1: 64602797-64602797 on Build GRCh38
rs752623290 Chr.1: 64602874-64602874 on Build GRCh38
rs376759587 Chr.1: 64602987-64602987 on Build GRCh38
rs756889525 Chr.1: 64602789-64602789 on Build GRCh38
rs765070362 Chr.1: 64602870-64602870 on Build GRCh38
rs932018323 Chr.1: 64602730-64602730 on Build GRCh38
rs749876404 Chr.1: 64602829-64602829 on Build GRCh38
rs199943361 Chr.1: 64602867-64602867 on Build GRCh38
rs952181064 Chr.1: 64602720-64602720 on Build GRCh38
rs943988425 Chr.1: 64602796-64602796 on Build GRCh38
rs527836063 Chr.1: 64602809-64602809 on Build GRCh38
rs780694131 Chr.1: 64602826-64602826 on Build GRCh38
rs773308577 Chr.1: 64602866-64602866 on Build GRCh38
rs756542793 Chr.1: 64602898-64602898 on Build GRCh38
rs554748772 Chr.1: 64603015-64603015 on Build GRCh38
rs768825442 Chr.1: 64602830-64602830 on Build GRCh38
rs572582155 Chr.1: 64602976-64602976 on Build GRCh38
rs953467400 Chr.1: 64603020-64603020 on Build GRCh38
rs762930702 Chr.1: 64602878-64602878 on Build GRCh38
rs796399258 Chr.1: 64602785-64602785 on Build GRCh38
rs777327603 Chr.1: 64602768-64602768 on Build GRCh38
rs961297219 Chr.1: 64602701-64602701 on Build GRCh38
rs539058064 Chr.1: 64602956-64602956 on Build GRCh38
rs373504906 Chr.1: 64602897-64602897 on Build GRCh38
rs752641787 Chr.1: 64602759-64602759 on Build GRCh38
rs772296106 Chr.1: 64602856-64602856 on Build GRCh38
rs957078708 Chr.1: 64602699-64602699 on Build GRCh38
rs776598960 Chr.1: 64602750-64602750 on Build GRCh38
rs755492834 Chr.1: 64602933-64602933 on Build GRCh38

View Full Product Details