Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145850669-145850165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6736
Entrez Gene ID
102466191
Gene Name
microRNA 6736
Gene Aliases
hsa-mir-6736
UniGene
Gene Chromosome Location
Chr.1: 145850587-145850645 on Build GRCh38
Transcripts
NR_106794.1 NM_006099.3

Gene Symbol
PIAS3
Entrez Gene ID
10401
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
ZMIZ5
UniGene
Hs.435761
Gene Chromosome Location
Chr.1: 145848522-145859081 on Build GRCh38
Transcripts
NR_106794.1 NM_006099.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6271351 c.1478C>A p.S493* Chr.1: 145850557-145850557 on Build GRCh38
3399671 c.1529G>A p.S510N Chr.1: 145850506-145850506 on Build GRCh38
5911468 c.1475C>T p.S492F Chr.1: 145850560-145850560 on Build GRCh38
346536 c.1466A>T p.H489L Chr.1: 145850569-145850569 on Build GRCh38
2121109 c.1465C>T p.H489Y Chr.1: 145850570-145850570 on Build GRCh38
View More

View Full Product Details