Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 5891371-5891915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4X
Entrez Gene ID
57502
Gene Name
neuroligin 4, X-linked
Gene Aliases
ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
UniGene
Hs.21107
Gene Chromosome Location
Chr.X: 5890026-6228882 on Build GRCh38
Transcripts
NM_001282145.1 NM_001282146.1 NM_020742.3 NM_181332.2 XM_005274564.2 XM_005274565.2 XM_005274566.4 XM_006724504.3 XM_011545547.2 XM_011545548.2 XM_017029690.1 XM_017029691.1 XM_017029692.1 XM_017029693.1

SNP Detail

SNP ID Chromosome Location
rs761221692 Chr.X: 5891768-5891769 on Build GRCh38
rs974462671 Chr.X: 5891792-5891792 on Build GRCh38
rs1038574838 Chr.X: 5891606-5891609 on Build GRCh38
rs1025642868 Chr.X: 5891618-5891618 on Build GRCh38
rs41305199 Chr.X: 5891533-5891533 on Build GRCh38
rs948283927 Chr.X: 5891606-5891606 on Build GRCh38
rs946831991 Chr.X: 5891447-5891447 on Build GRCh38
rs866108602 Chr.X: 5891708-5891708 on Build GRCh38
rs902345822 Chr.X: 5891434-5891434 on Build GRCh38
rs1051632622 Chr.X: 5891412-5891412 on Build GRCh38
rs933043273 Chr.X: 5891405-5891405 on Build GRCh38
rs759547470 Chr.X: 5891631-5891631 on Build GRCh38
rs771124404 Chr.X: 5891560-5891560 on Build GRCh38
rs747959552 Chr.X: 5891556-5891556 on Build GRCh38
rs1017204573 Chr.X: 5891704-5891704 on Build GRCh38
rs1012270818 Chr.X: 5891535-5891535 on Build GRCh38
rs758366733 Chr.X: 5891424-5891424 on Build GRCh38
rs1053711429 Chr.X: 5891482-5891482 on Build GRCh38
rs751049210 Chr.X: 5891594-5891594 on Build GRCh38
rs990012787 Chr.X: 5891894-5891894 on Build GRCh38
rs999320144 Chr.X: 5891448-5891448 on Build GRCh38
rs147323238 Chr.X: 5891751-5891751 on Build GRCh38
rs951554144 Chr.X: 5891628-5891628 on Build GRCh38
rs923651333 Chr.X: 5891879-5891879 on Build GRCh38
rs989980070 Chr.X: 5891425-5891425 on Build GRCh38
rs767545507 Chr.X: 5891690-5891690 on Build GRCh38
rs933097098 Chr.X: 5891870-5891870 on Build GRCh38
rs1039085953 Chr.X: 5891397-5891397 on Build GRCh38
rs984222942 Chr.X: 5891677-5891677 on Build GRCh38
rs186141923 Chr.X: 5891579-5891579 on Build GRCh38
rs963952986 Chr.X: 5891893-5891893 on Build GRCh38
rs921724995 Chr.X: 5891403-5891403 on Build GRCh38
rs1025350923 Chr.X: 5891812-5891812 on Build GRCh38
rs562212004 Chr.X: 5891873-5891873 on Build GRCh38
rs921586949 Chr.X: 5891847-5891847 on Build GRCh38
rs376804890 Chr.X: 5891871-5891871 on Build GRCh38
rs923956491 Chr.X: 5891490-5891490 on Build GRCh38
rs1042447282 Chr.X: 5891473-5891473 on Build GRCh38
rs994339431 Chr.X: 5891753-5891753 on Build GRCh38
rs1024039949 Chr.X: 5891537-5891537 on Build GRCh38
rs1018354711 Chr.X: 5891886-5891886 on Build GRCh38
rs939329379 Chr.X: 5891522-5891522 on Build GRCh38

View Full Product Details