Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 5892595-5893109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4X
Entrez Gene ID
57502
Gene Name
neuroligin 4, X-linked
Gene Aliases
ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
UniGene
Hs.21107
Gene Chromosome Location
Chr.X: 5890026-6228882 on Build GRCh38
Transcripts
NM_001282145.1 NM_001282146.1 NM_020742.3 NM_181332.2 XM_005274564.2 XM_005274565.2 XM_005274566.4 XM_006724504.3 XM_011545547.2 XM_011545548.2 XM_017029690.1 XM_017029691.1 XM_017029692.1 XM_017029693.1

SNP Detail

SNP ID Chromosome Location
rs776444310 Chr.X: 5892791-5892791 on Build GRCh38
rs1039404939 Chr.X: 5892676-5892683 on Build GRCh38
rs541646409 Chr.X: 5893045-5893045 on Build GRCh38
rs750719645 Chr.X: 5892718-5892718 on Build GRCh38
rs374449420 Chr.X: 5892907-5892907 on Build GRCh38
rs182561696 Chr.X: 5892694-5892694 on Build GRCh38
rs4999056 Chr.X: 5892997-5892997 on Build GRCh38
rs773200947 Chr.X: 5892807-5892807 on Build GRCh38
rs1030713987 Chr.X: 5893006-5893006 on Build GRCh38
rs377394394 Chr.X: 5892943-5892943 on Build GRCh38
rs886042929 Chr.X: 5893009-5893009 on Build GRCh38
rs754072429 Chr.X: 5892986-5892986 on Build GRCh38
rs760914699 Chr.X: 5892855-5892855 on Build GRCh38
rs4995611 Chr.X: 5892908-5892908 on Build GRCh38
rs754206681 Chr.X: 5892897-5892897 on Build GRCh38
rs749124096 Chr.X: 5893077-5893077 on Build GRCh38
rs773004622 Chr.X: 5892967-5892967 on Build GRCh38
rs3810688 Chr.X: 5892715-5892715 on Build GRCh38
rs752745113 Chr.X: 5892853-5892853 on Build GRCh38
rs141662515 Chr.X: 5893042-5893042 on Build GRCh38
rs765147883 Chr.X: 5892802-5892802 on Build GRCh38
rs369528412 Chr.X: 5893051-5893051 on Build GRCh38
rs764361221 Chr.X: 5892984-5892984 on Build GRCh38
rs780768055 Chr.X: 5892958-5892958 on Build GRCh38
rs140331762 Chr.X: 5892778-5892778 on Build GRCh38
rs762127085 Chr.X: 5892987-5892987 on Build GRCh38
rs775611375 Chr.X: 5892975-5892975 on Build GRCh38
rs769512850 Chr.X: 5892963-5892963 on Build GRCh38
rs758987323 Chr.X: 5892930-5892930 on Build GRCh38
rs956579844 Chr.X: 5893014-5893014 on Build GRCh38
rs762881868 Chr.X: 5892832-5892832 on Build GRCh38
rs751178982 Chr.X: 5892779-5892779 on Build GRCh38
rs764265488 Chr.X: 5892871-5892871 on Build GRCh38
rs4995613 Chr.X: 5892880-5892880 on Build GRCh38
rs899525787 Chr.X: 5892699-5892699 on Build GRCh38
rs989634767 Chr.X: 5892658-5892658 on Build GRCh38
rs1011168104 Chr.X: 5892707-5892707 on Build GRCh38
rs770890880 Chr.X: 5892973-5892973 on Build GRCh38
rs754514506 Chr.X: 5893034-5893034 on Build GRCh38
rs758932180 Chr.X: 5892998-5892998 on Build GRCh38
rs927909199 Chr.X: 5892761-5892761 on Build GRCh38
rs764592813 Chr.X: 5892637-5892637 on Build GRCh38
rs912708848 Chr.X: 5892650-5892650 on Build GRCh38
rs149065055 Chr.X: 5893050-5893050 on Build GRCh38
rs762832258 Chr.X: 5892968-5892968 on Build GRCh38
rs969639237 Chr.X: 5892711-5892711 on Build GRCh38
rs781005111 Chr.X: 5892979-5892979 on Build GRCh38
rs777508939 Chr.X: 5893074-5893074 on Build GRCh38
rs75350246 Chr.X: 5892988-5892988 on Build GRCh38
rs760718761 Chr.X: 5892980-5892980 on Build GRCh38
rs780463669 Chr.X: 5893004-5893004 on Build GRCh38
rs999173635 Chr.X: 5892852-5892852 on Build GRCh38
rs750889869 Chr.X: 5892928-5892928 on Build GRCh38
rs777673585 Chr.X: 5893021-5893021 on Build GRCh38
rs947118416 Chr.X: 5893039-5893039 on Build GRCh38
rs3810687 Chr.X: 5892643-5892643 on Build GRCh38
rs960107435 Chr.X: 5892862-5892862 on Build GRCh38
rs4995609 Chr.X: 5892951-5892951 on Build GRCh38
rs748520373 Chr.X: 5893019-5893048 on Build GRCh38
rs770882050 Chr.X: 5893078-5893078 on Build GRCh38
rs761564202 Chr.X: 5892797-5892797 on Build GRCh38
rs747933546 Chr.X: 5892959-5892959 on Build GRCh38
rs768213977 Chr.X: 5892781-5892781 on Build GRCh38
rs765660407 Chr.X: 5892749-5892752 on Build GRCh38
rs908546493 Chr.X: 5892803-5892803 on Build GRCh38
rs866564006 Chr.X: 5892869-5892869 on Build GRCh38
rs981041123 Chr.X: 5893081-5893081 on Build GRCh38
rs956478176 Chr.X: 5892651-5892651 on Build GRCh38
rs768321834 Chr.X: 5892947-5892947 on Build GRCh38
rs764978540 Chr.X: 5892638-5892638 on Build GRCh38
rs758171012 Chr.X: 5892762-5892762 on Build GRCh38
rs779554641 Chr.X: 5892914-5892914 on Build GRCh38
rs767531295 Chr.X: 5892841-5892841 on Build GRCh38
rs746531523 Chr.X: 5892944-5892944 on Build GRCh38

View Full Product Details