Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 22311156-22311665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMY2FP
Entrez Gene ID
159162
Gene Name
RNA binding motif protein, Y-linked, family 2, member F pseudogene
Gene Aliases
-
UniGene
Hs.711527
Gene Chromosome Location
Chr.Y: 22308859-22316205 on Build GRCh38
Transcripts
NR_002193.2

SNP Detail

SNP ID Chromosome Location
rs752991270 Chr.Y: 22311431-22311431 on Build GRCh38
rs770257166 Chr.Y: 22311572-22311572 on Build GRCh38
rs763723550 Chr.Y: 22311456-22311456 on Build GRCh38
rs751147051 Chr.Y: 22311474-22311474 on Build GRCh38
rs766945744 Chr.Y: 22311359-22311359 on Build GRCh38
rs758058189 Chr.Y: 22311445-22311445 on Build GRCh38
rs781239429 Chr.Y: 22311523-22311523 on Build GRCh38
rs776976749 Chr.Y: 22311376-22311376 on Build GRCh38
rs113724139 Chr.Y: 22311488-22311488 on Build GRCh38
rs200216074 Chr.Y: 22311419-22311419 on Build GRCh38
rs760036894 Chr.Y: 22311413-22311413 on Build GRCh38

View Full Product Details