Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16483747-16484124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR3
Entrez Gene ID
125972
Gene Name
calreticulin 3
Gene Aliases
CMH19, CRT2, CT93
UniGene
Hs.304020
Gene Chromosome Location
Chr.19: 16479057-16496192 on Build GRCh38
Transcripts
NM_145046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992462 c.588A>G p.I196M Chr.19: 16484020-16484020 on Build GRCh38
992461 c.591G>T p.E197D Chr.19: 16484017-16484017 on Build GRCh38
4279129 c.641C>T p.S214L Chr.19: 16483967-16483967 on Build GRCh38
474301 c.578C>A p.S193Y Chr.19: 16484030-16484030 on Build GRCh38
3530025 c.557T>C p.I186T Chr.19: 16484051-16484051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776906320 Chr.19: 16483999-16483999 on Build GRCh38
rs965608374 Chr.19: 16483873-16483873 on Build GRCh38
rs765297065 Chr.19: 16483935-16483935 on Build GRCh38
rs1035181108 Chr.19: 16484063-16484063 on Build GRCh38
rs111870908 Chr.19: 16483989-16483989 on Build GRCh38
rs560967718 Chr.19: 16483906-16483906 on Build GRCh38
rs1051163114 Chr.19: 16483875-16483875 on Build GRCh38
rs978883194 Chr.19: 16483881-16483881 on Build GRCh38
rs376733366 Chr.19: 16484100-16484100 on Build GRCh38
rs746871999 Chr.19: 16483982-16483982 on Build GRCh38
rs112527758 Chr.19: 16483890-16483890 on Build GRCh38
rs377026384 Chr.19: 16483969-16483969 on Build GRCh38
rs145046810 Chr.19: 16484014-16484014 on Build GRCh38
rs374187284 Chr.19: 16484091-16484091 on Build GRCh38
rs764373762 Chr.19: 16483951-16483951 on Build GRCh38
rs375866420 Chr.19: 16483892-16483892 on Build GRCh38
rs762301226 Chr.19: 16484047-16484047 on Build GRCh38
rs776696174 Chr.19: 16483930-16483930 on Build GRCh38
rs764707663 Chr.19: 16484028-16484028 on Build GRCh38
rs552364605 Chr.19: 16484092-16484092 on Build GRCh38
rs149108772 Chr.19: 16484029-16484029 on Build GRCh38
rs529644089 Chr.19: 16483962-16483962 on Build GRCh38
rs200669493 Chr.19: 16483966-16483966 on Build GRCh38
rs771137388 Chr.19: 16483919-16483919 on Build GRCh38
rs572074650 Chr.19: 16483878-16483878 on Build GRCh38
rs748099861 Chr.19: 16483975-16483975 on Build GRCh38
rs763397517 Chr.19: 16483950-16483950 on Build GRCh38
rs540693852 Chr.19: 16483880-16483880 on Build GRCh38
rs752204583 Chr.19: 16484097-16484097 on Build GRCh38
rs369853144 Chr.19: 16483884-16483884 on Build GRCh38
rs772004741 Chr.19: 16483896-16483896 on Build GRCh38
rs372889302 Chr.19: 16483960-16483960 on Build GRCh38
rs1009771719 Chr.19: 16483788-16483788 on Build GRCh38
rs778627769 Chr.19: 16484038-16484041 on Build GRCh38
rs772004043 Chr.19: 16483976-16483976 on Build GRCh38
rs1019856459 Chr.19: 16483809-16483809 on Build GRCh38
rs373585740 Chr.19: 16483889-16483889 on Build GRCh38
rs769879360 Chr.19: 16484004-16484004 on Build GRCh38
rs775484097 Chr.19: 16484007-16484007 on Build GRCh38
rs947671532 Chr.19: 16483833-16483833 on Build GRCh38
rs1034788641 Chr.19: 16484017-16484017 on Build GRCh38
rs143671909 Chr.19: 16484088-16484088 on Build GRCh38
rs747656642 Chr.19: 16484044-16484044 on Build GRCh38
rs760836196 Chr.19: 16483908-16483908 on Build GRCh38
rs369476772 Chr.19: 16484062-16484062 on Build GRCh38
rs550089146 Chr.19: 16483967-16483967 on Build GRCh38
rs940668458 Chr.19: 16483923-16483923 on Build GRCh38
rs1052287139 Chr.19: 16483895-16483895 on Build GRCh38
rs150376706 Chr.19: 16484000-16484000 on Build GRCh38
rs777454147 Chr.19: 16483981-16483981 on Build GRCh38
rs368652672 Chr.19: 16483941-16483941 on Build GRCh38
rs564650089 Chr.19: 16483794-16483794 on Build GRCh38

View Full Product Details