Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49959122-49959703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGLEC11
Entrez Gene ID
114132
Gene Name
sialic acid binding Ig like lectin 11
Gene Aliases
-
UniGene
Hs.661852
Gene Chromosome Location
Chr.19: 49948993-49961172 on Build GRCh38
Transcripts
NM_001135163.1 NM_052884.2 XM_005258476.3 XM_005258477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726022 c.988G>T p.G330W Chr.19: 49959429-49959429 on Build GRCh38
4596287 c.935G>A p.W312* Chr.19: 49959482-49959482 on Build GRCh38
80521 c.1008G>A p.A336A Chr.19: 49959409-49959409 on Build GRCh38
3835416 c.987A>G p.S329S Chr.19: 49959430-49959430 on Build GRCh38
3537063 c.978C>A p.A326A Chr.19: 49959439-49959439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565125962 Chr.19: 49959440-49959440 on Build GRCh38
rs200227828 Chr.19: 49959430-49959430 on Build GRCh38
rs1008637083 Chr.19: 49959246-49959246 on Build GRCh38
rs199521132 Chr.19: 49959448-49959448 on Build GRCh38
rs1028975763 Chr.19: 49959181-49959181 on Build GRCh38
rs759136292 Chr.19: 49959519-49959519 on Build GRCh38
rs201747553 Chr.19: 49959452-49959452 on Build GRCh38
rs575148848 Chr.19: 49959328-49959328 on Build GRCh38
rs995644306 Chr.19: 49959167-49959167 on Build GRCh38
rs769673458 Chr.19: 49959426-49959426 on Build GRCh38
rs563064545 Chr.19: 49959475-49959475 on Build GRCh38
rs773748696 Chr.19: 49959566-49959566 on Build GRCh38
rs780048480 Chr.19: 49959466-49959466 on Build GRCh38
rs770403078 Chr.19: 49959435-49959435 on Build GRCh38
rs560971741 Chr.19: 49959346-49959346 on Build GRCh38
rs898643141 Chr.19: 49959161-49959161 on Build GRCh38
rs777765302 Chr.19: 49959494-49959494 on Build GRCh38
rs200945898 Chr.19: 49959451-49959451 on Build GRCh38
rs758411373 Chr.19: 49959410-49959410 on Build GRCh38
rs758493747 Chr.19: 49959464-49959464 on Build GRCh38
rs765772147 Chr.19: 49959453-49959453 on Build GRCh38
rs767305033 Chr.19: 49959439-49959439 on Build GRCh38
rs199735090 Chr.19: 49959342-49959342 on Build GRCh38
rs902487924 Chr.19: 49959676-49959679 on Build GRCh38
rs749229969 Chr.19: 49959502-49959502 on Build GRCh38
rs777534180 Chr.19: 49959668-49959670 on Build GRCh38
rs925373495 Chr.19: 49959339-49959339 on Build GRCh38
rs572827488 Chr.19: 49959381-49959381 on Build GRCh38
rs916479200 Chr.19: 49959356-49959356 on Build GRCh38
rs747883983 Chr.19: 49959480-49959480 on Build GRCh38
rs557296049 Chr.19: 49959279-49959279 on Build GRCh38
rs201944046 Chr.19: 49959445-49959445 on Build GRCh38
rs112095721 Chr.19: 49959603-49959603 on Build GRCh38
rs112719752 Chr.19: 49959150-49959150 on Build GRCh38
rs364111 Chr.19: 49959383-49959383 on Build GRCh38
rs550818529 Chr.19: 49959471-49959471 on Build GRCh38
rs771792082 Chr.19: 49959520-49959520 on Build GRCh38
rs150164114 Chr.19: 49959425-49959425 on Build GRCh38
rs754233728 Chr.19: 49959254-49959254 on Build GRCh38
rs775285253 Chr.19: 49959529-49959529 on Build GRCh38
rs750957387 Chr.19: 49959454-49959454 on Build GRCh38
rs775587153 Chr.19: 49959317-49959317 on Build GRCh38
rs769585278 Chr.19: 49959493-49959493 on Build GRCh38
rs779585178 Chr.19: 49959399-49959399 on Build GRCh38
rs765753786 Chr.19: 49959541-49959541 on Build GRCh38
rs747255098 Chr.19: 49959414-49959414 on Build GRCh38
rs774029699 Chr.19: 49959436-49959436 on Build GRCh38
rs954359890 Chr.19: 49959213-49959213 on Build GRCh38
rs369690804 Chr.19: 49959142-49959146 on Build GRCh38
rs1020306982 Chr.19: 49959676-49959677 on Build GRCh38
rs538925742 Chr.19: 49959260-49959260 on Build GRCh38
rs751775195 Chr.19: 49959469-49959469 on Build GRCh38
rs768404125 Chr.19: 49959327-49959327 on Build GRCh38
rs370983001 Chr.19: 49959293-49959293 on Build GRCh38
rs577959438 Chr.19: 49959256-49959256 on Build GRCh38
rs762396089 Chr.19: 49959348-49959348 on Build GRCh38
rs111729530 Chr.19: 49959632-49959632 on Build GRCh38
rs113344458 Chr.19: 49959481-49959481 on Build GRCh38
rs751548313 Chr.19: 49959675-49959675 on Build GRCh38
rs766002290 Chr.19: 49959360-49959360 on Build GRCh38
rs773690874 Chr.19: 49959510-49959510 on Build GRCh38
rs553183502 Chr.19: 49959223-49959223 on Build GRCh38
rs978149815 Chr.19: 49959314-49959314 on Build GRCh38
rs966705529 Chr.19: 49959284-49959284 on Build GRCh38
rs773554566 Chr.19: 49959638-49959641 on Build GRCh38
rs761788292 Chr.19: 49959335-49959335 on Build GRCh38
rs759148240 Chr.19: 49959438-49959438 on Build GRCh38
rs751178318 Chr.19: 49959362-49959362 on Build GRCh38
rs543110804 Chr.19: 49959303-49959303 on Build GRCh38
rs530073857 Chr.19: 49959564-49959564 on Build GRCh38
rs995631114 Chr.19: 49959544-49959544 on Build GRCh38
rs750459116 Chr.19: 49959409-49959409 on Build GRCh38
rs764599551 Chr.19: 49959379-49959379 on Build GRCh38
rs780316906 Chr.19: 49959413-49959413 on Build GRCh38
rs889960298 Chr.19: 49959638-49959638 on Build GRCh38
rs764696561 Chr.19: 49959341-49959341 on Build GRCh38
rs762262036 Chr.19: 49959530-49959530 on Build GRCh38
rs1008443101 Chr.19: 49959673-49959673 on Build GRCh38
rs768418797 Chr.19: 49959421-49959421 on Build GRCh38
rs770440448 Chr.19: 49959504-49959504 on Build GRCh38
rs940378906 Chr.19: 49959478-49959478 on Build GRCh38
rs1049959246 Chr.19: 49959577-49959578 on Build GRCh38
rs758387412 Chr.19: 49959344-49959344 on Build GRCh38
rs766916551 Chr.19: 49959250-49959250 on Build GRCh38
rs746749510 Chr.19: 49959322-49959322 on Build GRCh38
rs773149758 Chr.19: 49959427-49959427 on Build GRCh38

View Full Product Details