Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152722459-152723037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYCT1
Entrez Gene ID
80177
Gene Name
myc target 1
Gene Aliases
MTLC
UniGene
Hs.18160
Gene Chromosome Location
Chr.6: 152697895-152747171 on Build GRCh38
Transcripts
NM_025107.2 XM_011536149.2 XM_011536150.2 XM_011536151.2 XM_011536152.2 XM_017011319.1

SNP Detail

SNP ID Chromosome Location
rs941899758 Chr.6: 152722618-152722618 on Build GRCh38
rs544821481 Chr.6: 152722891-152722891 on Build GRCh38
rs1030317822 Chr.6: 152722721-152722721 on Build GRCh38
rs968603990 Chr.6: 152722671-152722671 on Build GRCh38
rs567887166 Chr.6: 152722674-152722674 on Build GRCh38
rs918969119 Chr.6: 152722882-152722882 on Build GRCh38
rs146967867 Chr.6: 152722505-152722505 on Build GRCh38
rs137991007 Chr.6: 152722719-152722719 on Build GRCh38
rs917925503 Chr.6: 152722784-152722784 on Build GRCh38
rs1041575977 Chr.6: 152722930-152722930 on Build GRCh38
rs1051379740 Chr.6: 152722655-152722655 on Build GRCh38
rs942231289 Chr.6: 152722619-152722619 on Build GRCh38
rs149091531 Chr.6: 152722994-152722994 on Build GRCh38
rs949665651 Chr.6: 152722643-152722643 on Build GRCh38
rs867616580 Chr.6: 152722679-152722679 on Build GRCh38
rs749919199 Chr.6: 152722942-152722942 on Build GRCh38
rs747174841 Chr.6: 152722860-152722860 on Build GRCh38
rs775928321 Chr.6: 152722559-152722559 on Build GRCh38
rs972543511 Chr.6: 152723003-152723003 on Build GRCh38
rs919525869 Chr.6: 152722632-152722632 on Build GRCh38
rs1036899632 Chr.6: 152722631-152722632 on Build GRCh38
rs919400415 Chr.6: 152722816-152722816 on Build GRCh38
rs757281346 Chr.6: 152722879-152722879 on Build GRCh38
rs909272423 Chr.6: 152722610-152722610 on Build GRCh38
rs35529547 Chr.6: 152722582-152722583 on Build GRCh38
rs892920090 Chr.6: 152722906-152722907 on Build GRCh38
rs775358515 Chr.6: 152722986-152722986 on Build GRCh38
rs1007524588 Chr.6: 152722978-152722978 on Build GRCh38
rs976089673 Chr.6: 152722499-152722499 on Build GRCh38
rs986168287 Chr.6: 152722602-152722602 on Build GRCh38
rs1007249717 Chr.6: 152722899-152722899 on Build GRCh38
rs774370043 Chr.6: 152722720-152722720 on Build GRCh38
rs1055571024 Chr.6: 152722675-152722675 on Build GRCh38
rs575307938 Chr.6: 152722628-152722629 on Build GRCh38
rs947984780 Chr.6: 152722558-152722558 on Build GRCh38
rs917802457 Chr.6: 152722543-152722543 on Build GRCh38
rs367571372 Chr.6: 152723002-152723002 on Build GRCh38
rs565292684 Chr.6: 152722783-152722783 on Build GRCh38
rs953395090 Chr.6: 152722741-152722741 on Build GRCh38
rs1036069941 Chr.6: 152722631-152722631 on Build GRCh38
rs897616211 Chr.6: 152722638-152722638 on Build GRCh38
rs924045589 Chr.6: 152722501-152722501 on Build GRCh38
rs1020530971 Chr.6: 152722699-152722699 on Build GRCh38
rs772739646 Chr.6: 152722741-152722742 on Build GRCh38
rs376924226 Chr.6: 152722556-152722556 on Build GRCh38
rs748405604 Chr.6: 152722744-152722744 on Build GRCh38
rs1011415696 Chr.6: 152722694-152722694 on Build GRCh38
rs1018555157 Chr.6: 152722670-152722670 on Build GRCh38
rs992193180 Chr.6: 152722533-152722533 on Build GRCh38
rs949688332 Chr.6: 152722825-152722825 on Build GRCh38
rs1000999617 Chr.6: 152722948-152722948 on Build GRCh38
rs997276084 Chr.6: 152722954-152722954 on Build GRCh38
rs1001071511 Chr.6: 152722691-152722691 on Build GRCh38
rs956782496 Chr.6: 152722776-152722776 on Build GRCh38
rs9397552 Chr.6: 152722915-152722915 on Build GRCh38
rs986073296 Chr.6: 152722762-152722762 on Build GRCh38
rs941862492 Chr.6: 152722863-152722863 on Build GRCh38
rs536870756 Chr.6: 152722892-152722892 on Build GRCh38
rs909172778 Chr.6: 152722853-152722853 on Build GRCh38
rs909303861 Chr.6: 152722764-152722764 on Build GRCh38
rs909026934 Chr.6: 152722606-152722606 on Build GRCh38
rs191594983 Chr.6: 152722785-152722785 on Build GRCh38
rs1036163434 Chr.6: 152722877-152722877 on Build GRCh38
rs888764012 Chr.6: 152722898-152722898 on Build GRCh38
rs547000868 Chr.6: 152722770-152722770 on Build GRCh38
rs981078428 Chr.6: 152722795-152722795 on Build GRCh38
rs980831312 Chr.6: 152723007-152723007 on Build GRCh38
rs555501621 Chr.6: 152722901-152722901 on Build GRCh38
rs752952321 Chr.6: 152722641-152722641 on Build GRCh38
rs527654700 Chr.6: 152722934-152722934 on Build GRCh38
rs368143240 Chr.6: 152722667-152722667 on Build GRCh38
rs771416802 Chr.6: 152722742-152722742 on Build GRCh38
rs1044877174 Chr.6: 152722592-152722592 on Build GRCh38
rs113721272 Chr.6: 152722977-152722977 on Build GRCh38
rs553300226 Chr.6: 152722967-152722967 on Build GRCh38
rs969444644 Chr.6: 152722793-152722793 on Build GRCh38
rs891352557 Chr.6: 152722663-152722663 on Build GRCh38
rs557461679 Chr.6: 152722886-152722886 on Build GRCh38

View Full Product Details