Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74359757-74360297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP2
Entrez Gene ID
7461
Gene Name
CAP-Gly domain containing linker protein 2
Gene Aliases
CLIP, CLIP-115, CYLN2, WBSCR3, WBSCR4, WSCR3, WSCR4
UniGene
Hs.647018
Gene Chromosome Location
Chr.7: 74289437-74405943 on Build GRCh38
Transcripts
NM_003388.4 NM_032421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630404 c.1216T>C p.Y406H Chr.7: 74360175-74360175 on Build GRCh38
1648021 c.1230C>T p.A410A Chr.7: 74360189-74360189 on Build GRCh38
6379065 c.1309G>T p.E437* Chr.7: 74360268-74360268 on Build GRCh38
4672825 c.1274G>A p.R425Q Chr.7: 74360233-74360233 on Build GRCh38
5141552 c.1231G>A p.E411K Chr.7: 74360190-74360190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781834378 Chr.7: 74360237-74360237 on Build GRCh38
rs999258666 Chr.7: 74359959-74359959 on Build GRCh38
rs972964522 Chr.7: 74360130-74360130 on Build GRCh38
rs918928234 Chr.7: 74359826-74359826 on Build GRCh38
rs954343845 Chr.7: 74360057-74360057 on Build GRCh38
rs782323424 Chr.7: 74360180-74360180 on Build GRCh38
rs542092183 Chr.7: 74360163-74360163 on Build GRCh38
rs782250917 Chr.7: 74360047-74360047 on Build GRCh38
rs782177238 Chr.7: 74360177-74360177 on Build GRCh38
rs782013291 Chr.7: 74360182-74360182 on Build GRCh38
rs116564168 Chr.7: 74359896-74359896 on Build GRCh38
rs782384622 Chr.7: 74360190-74360190 on Build GRCh38
rs1017603560 Chr.7: 74360089-74360089 on Build GRCh38
rs782414566 Chr.7: 74360008-74360008 on Build GRCh38
rs782624222 Chr.7: 74360157-74360157 on Build GRCh38
rs938397464 Chr.7: 74359884-74359884 on Build GRCh38
rs1028435820 Chr.7: 74360040-74360040 on Build GRCh38
rs746645546 Chr.7: 74359906-74359906 on Build GRCh38
rs938366494 Chr.7: 74360160-74360160 on Build GRCh38
rs367705587 Chr.7: 74360232-74360232 on Build GRCh38
rs984341432 Chr.7: 74360059-74360059 on Build GRCh38
rs782368353 Chr.7: 74360150-74360150 on Build GRCh38
rs564040624 Chr.7: 74360189-74360189 on Build GRCh38
rs533430096 Chr.7: 74360228-74360228 on Build GRCh38
rs971746685 Chr.7: 74359819-74359819 on Build GRCh38
rs948603989 Chr.7: 74359936-74359936 on Build GRCh38
rs904473332 Chr.7: 74359949-74359949 on Build GRCh38
rs904357431 Chr.7: 74360192-74360192 on Build GRCh38
rs1031643392 Chr.7: 74359997-74359997 on Build GRCh38
rs576970300 Chr.7: 74359911-74359911 on Build GRCh38
rs145022768 Chr.7: 74359938-74359938 on Build GRCh38
rs200008575 Chr.7: 74360229-74360229 on Build GRCh38
rs367722388 Chr.7: 74360128-74360128 on Build GRCh38
rs371915307 Chr.7: 74360134-74360134 on Build GRCh38
rs560402454 Chr.7: 74360219-74360219 on Build GRCh38
rs781808011 Chr.7: 74360146-74360146 on Build GRCh38
rs781829765 Chr.7: 74360125-74360125 on Build GRCh38
rs782612705 Chr.7: 74360176-74360176 on Build GRCh38
rs202062702 Chr.7: 74360211-74360211 on Build GRCh38
rs571139366 Chr.7: 74360039-74360039 on Build GRCh38
rs780959185 Chr.7: 74360090-74360090 on Build GRCh38
rs547249580 Chr.7: 74360233-74360233 on Build GRCh38
rs559283649 Chr.7: 74359953-74359953 on Build GRCh38
rs374931670 Chr.7: 74360150-74360150 on Build GRCh38
rs1056819368 Chr.7: 74359894-74359894 on Build GRCh38
rs782443943 Chr.7: 74360265-74360265 on Build GRCh38
rs576431893 Chr.7: 74359856-74359856 on Build GRCh38
rs915691959 Chr.7: 74360168-74360168 on Build GRCh38
rs782461129 Chr.7: 74360172-74360172 on Build GRCh38
rs550854768 Chr.7: 74359811-74359811 on Build GRCh38
rs782068816 Chr.7: 74360220-74360220 on Build GRCh38
rs374448871 Chr.7: 74360206-74360206 on Build GRCh38

View Full Product Details