Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34555845-34556342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDEF
Entrez Gene ID
25803
Gene Name
SAM pointed domain containing ETS transcription factor
Gene Aliases
PDEF, bA375E1.3
UniGene
Hs.485158
Gene Chromosome Location
Chr.6: 34537802-34556333 on Build GRCh38
Transcripts
NM_001252294.1 NM_012391.2

SNP Detail

SNP ID Chromosome Location
rs1036559660 Chr.6: 34556126-34556126 on Build GRCh38
rs1046010389 Chr.6: 34556161-34556161 on Build GRCh38
rs189542727 Chr.6: 34556321-34556321 on Build GRCh38
rs903373910 Chr.6: 34556232-34556232 on Build GRCh38
rs1026107990 Chr.6: 34555864-34555864 on Build GRCh38
rs544000631 Chr.6: 34555921-34555921 on Build GRCh38
rs911742654 Chr.6: 34556120-34556120 on Build GRCh38
rs957407917 Chr.6: 34556016-34556016 on Build GRCh38
rs559440579 Chr.6: 34556082-34556082 on Build GRCh38
rs921854762 Chr.6: 34556041-34556041 on Build GRCh38
rs936077009 Chr.6: 34556104-34556104 on Build GRCh38
rs540755799 Chr.6: 34556062-34556062 on Build GRCh38
rs931876022 Chr.6: 34556048-34556048 on Build GRCh38
rs374569801 Chr.6: 34556243-34556243 on Build GRCh38
rs906189007 Chr.6: 34556162-34556162 on Build GRCh38
rs139219168 Chr.6: 34556318-34556318 on Build GRCh38
rs146836954 Chr.6: 34556194-34556194 on Build GRCh38
rs532770564 Chr.6: 34556046-34556046 on Build GRCh38
rs1033772503 Chr.6: 34556179-34556179 on Build GRCh38
rs565503643 Chr.6: 34555985-34555985 on Build GRCh38
rs1024549837 Chr.6: 34556224-34556224 on Build GRCh38
rs999030498 Chr.6: 34556251-34556251 on Build GRCh38
rs1007932365 Chr.6: 34556124-34556124 on Build GRCh38
rs555257320 Chr.6: 34555907-34555907 on Build GRCh38
rs867565202 Chr.6: 34556186-34556186 on Build GRCh38
rs974739698 Chr.6: 34556294-34556294 on Build GRCh38
rs894749699 Chr.6: 34556195-34556195 on Build GRCh38
rs940515184 Chr.6: 34556122-34556122 on Build GRCh38
rs3822984 Chr.6: 34556277-34556277 on Build GRCh38
rs1055431092 Chr.6: 34556148-34556148 on Build GRCh38
rs529654687 Chr.6: 34556133-34556133 on Build GRCh38
rs1024872070 Chr.6: 34555899-34555899 on Build GRCh38
rs530727813 Chr.6: 34556247-34556247 on Build GRCh38
rs547784461 Chr.6: 34556149-34556149 on Build GRCh38
rs541091729 Chr.6: 34556033-34556033 on Build GRCh38
rs994566450 Chr.6: 34556219-34556219 on Build GRCh38
rs1053238106 Chr.6: 34556113-34556113 on Build GRCh38
rs9296118 Chr.6: 34555920-34555920 on Build GRCh38
rs746490461 Chr.6: 34555910-34555910 on Build GRCh38
rs975378173 Chr.6: 34556018-34556018 on Build GRCh38
rs987364491 Chr.6: 34555892-34555892 on Build GRCh38
rs1001912109 Chr.6: 34556166-34556166 on Build GRCh38
rs898881739 Chr.6: 34556207-34556207 on Build GRCh38
rs185496493 Chr.6: 34556291-34556291 on Build GRCh38
rs551924747 Chr.6: 34556265-34556265 on Build GRCh38
rs930302245 Chr.6: 34556138-34556138 on Build GRCh38
rs977938445 Chr.6: 34555929-34555929 on Build GRCh38
rs144082188 Chr.6: 34555894-34555894 on Build GRCh38
rs1014488192 Chr.6: 34556200-34556200 on Build GRCh38
rs965067323 Chr.6: 34556285-34556285 on Build GRCh38
rs111892902 Chr.6: 34555891-34555891 on Build GRCh38
rs957418089 Chr.6: 34556266-34556266 on Build GRCh38

View Full Product Details