Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35342426-35342953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARD
Entrez Gene ID
5467
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
UniGene
Hs.696032
Gene Chromosome Location
Chr.6: 35342558-35428191 on Build GRCh38
Transcripts
NM_001171818.1 NM_001171819.1 NM_001171820.1 NM_006238.4 NM_177435.2 XM_005249193.1 XM_006715120.1 XM_006715123.1 XM_011514707.1 XM_011514709.1 XM_011514710.1 XM_017010971.1 XM_017010972.1 XM_017010973.1 XM_017010974.1

SNP Detail

SNP ID Chromosome Location
rs988102900 Chr.6: 35342570-35342570 on Build GRCh38
rs535130327 Chr.6: 35342451-35342451 on Build GRCh38
rs753988439 Chr.6: 35342541-35342541 on Build GRCh38
rs948362988 Chr.6: 35342794-35342794 on Build GRCh38
rs564138934 Chr.6: 35342767-35342767 on Build GRCh38
rs891891150 Chr.6: 35342871-35342871 on Build GRCh38
rs545959242 Chr.6: 35342753-35342753 on Build GRCh38
rs1022363112 Chr.6: 35342845-35342845 on Build GRCh38
rs946983941 Chr.6: 35342586-35342586 on Build GRCh38
rs1009676000 Chr.6: 35342844-35342844 on Build GRCh38
rs919431079 Chr.6: 35342742-35342742 on Build GRCh38
rs1002791265 Chr.6: 35342826-35342826 on Build GRCh38
rs541385080 Chr.6: 35342601-35342601 on Build GRCh38
rs928011305 Chr.6: 35342557-35342557 on Build GRCh38
rs957048775 Chr.6: 35342675-35342675 on Build GRCh38
rs1046243972 Chr.6: 35342878-35342878 on Build GRCh38
rs907077278 Chr.6: 35342818-35342818 on Build GRCh38
rs944844958 Chr.6: 35342594-35342594 on Build GRCh38
rs906465911 Chr.6: 35342706-35342706 on Build GRCh38
rs1022814207 Chr.6: 35342635-35342635 on Build GRCh38
rs970346690 Chr.6: 35342585-35342585 on Build GRCh38
rs776977027 Chr.6: 35342875-35342875 on Build GRCh38
rs951925141 Chr.6: 35342501-35342501 on Build GRCh38
rs202245321 Chr.6: 35342593-35342593 on Build GRCh38
rs1007519632 Chr.6: 35342478-35342478 on Build GRCh38
rs928975837 Chr.6: 35342621-35342621 on Build GRCh38
rs1040136439 Chr.6: 35342595-35342595 on Build GRCh38
rs201616498 Chr.6: 35342657-35342657 on Build GRCh38
rs556645720 Chr.6: 35342484-35342484 on Build GRCh38
rs866028947 Chr.6: 35342850-35342850 on Build GRCh38
rs891618515 Chr.6: 35342831-35342831 on Build GRCh38
rs915245156 Chr.6: 35342577-35342577 on Build GRCh38
rs1022331832 Chr.6: 35342678-35342678 on Build GRCh38
rs9658058 Chr.6: 35342655-35342655 on Build GRCh38
rs982237434 Chr.6: 35342549-35342549 on Build GRCh38
rs200241121 Chr.6: 35342565-35342565 on Build GRCh38
rs140749896 Chr.6: 35342519-35342519 on Build GRCh38
rs1025281449 Chr.6: 35342705-35342705 on Build GRCh38
rs11571502 Chr.6: 35342634-35342634 on Build GRCh38
rs1009894857 Chr.6: 35342547-35342547 on Build GRCh38
rs912123925 Chr.6: 35342806-35342806 on Build GRCh38
rs181894752 Chr.6: 35342734-35342734 on Build GRCh38
rs9658059 Chr.6: 35342689-35342689 on Build GRCh38
rs910764975 Chr.6: 35342695-35342695 on Build GRCh38
rs539518483 Chr.6: 35342901-35342902 on Build GRCh38
rs200323112 Chr.6: 35342596-35342596 on Build GRCh38
rs201191408 Chr.6: 35342590-35342590 on Build GRCh38
rs531052740 Chr.6: 35342747-35342747 on Build GRCh38
rs956740820 Chr.6: 35342566-35342566 on Build GRCh38
rs1032573113 Chr.6: 35342522-35342522 on Build GRCh38
rs9658060 Chr.6: 35342872-35342872 on Build GRCh38
rs907049863 Chr.6: 35342645-35342645 on Build GRCh38
rs150373160 Chr.6: 35342902-35342903 on Build GRCh38
rs887431162 Chr.6: 35342627-35342627 on Build GRCh38
rs546581318 Chr.6: 35342864-35342864 on Build GRCh38
rs202006248 Chr.6: 35342559-35342559 on Build GRCh38
rs201136524 Chr.6: 35342622-35342622 on Build GRCh38
rs1002143959 Chr.6: 35342723-35342723 on Build GRCh38
rs1048779997 Chr.6: 35342762-35342762 on Build GRCh38
rs11571503 Chr.6: 35342797-35342797 on Build GRCh38
rs1053578595 Chr.6: 35342868-35342868 on Build GRCh38
rs537630935 Chr.6: 35342524-35342524 on Build GRCh38
rs1000223218 Chr.6: 35342686-35342686 on Build GRCh38
rs1031958915 Chr.6: 35342830-35342830 on Build GRCh38
rs796403394 Chr.6: 35342497-35342497 on Build GRCh38
rs986366754 Chr.6: 35342587-35342587 on Build GRCh38
rs944899012 Chr.6: 35342720-35342720 on Build GRCh38
rs968020094 Chr.6: 35342687-35342687 on Build GRCh38
rs542286834 Chr.6: 35342661-35342661 on Build GRCh38
rs760456727 Chr.6: 35342506-35342506 on Build GRCh38
rs971620162 Chr.6: 35342528-35342528 on Build GRCh38
rs370185256 Chr.6: 35342805-35342805 on Build GRCh38
rs966734381 Chr.6: 35342902-35342902 on Build GRCh38
rs968052702 Chr.6: 35342857-35342857 on Build GRCh38
rs988218814 Chr.6: 35342677-35342677 on Build GRCh38
rs973561557 Chr.6: 35342748-35342748 on Build GRCh38
rs986095578 Chr.6: 35342866-35342866 on Build GRCh38
rs1009410715 Chr.6: 35342694-35342694 on Build GRCh38
rs1035114626 Chr.6: 35342665-35342665 on Build GRCh38

View Full Product Details