Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35426487-35426988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPARD
Entrez Gene ID
5467
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB
UniGene
Hs.696032
Gene Chromosome Location
Chr.6: 35342558-35428191 on Build GRCh38
Transcripts
NM_001171818.1 NM_001171819.1 NM_001171820.1 NM_006238.4 XM_005249193.1 XM_006715120.1 XM_006715123.1 XM_011514707.1 XM_011514709.1 XM_011514710.1 XM_017010971.1 XM_017010972.1 XM_017010973.1 XM_017010974.1 XM_017010975.1

SNP Detail

SNP ID Chromosome Location
rs9658170 Chr.6: 35426727-35426727 on Build GRCh38
rs922256837 Chr.6: 35426923-35426923 on Build GRCh38
rs913373418 Chr.6: 35426627-35426627 on Build GRCh38
rs201127360 Chr.6: 35426840-35426840 on Build GRCh38
rs570085469 Chr.6: 35426577-35426577 on Build GRCh38
rs201744699 Chr.6: 35426714-35426714 on Build GRCh38
rs1014981271 Chr.6: 35426957-35426957 on Build GRCh38
rs952203672 Chr.6: 35426720-35426720 on Build GRCh38
rs199779515 Chr.6: 35426952-35426952 on Build GRCh38
rs1012280649 Chr.6: 35426707-35426707 on Build GRCh38
rs34735061 Chr.6: 35426659-35426660 on Build GRCh38
rs201459821 Chr.6: 35426588-35426588 on Build GRCh38
rs946870724 Chr.6: 35426632-35426632 on Build GRCh38
rs563382262 Chr.6: 35426758-35426758 on Build GRCh38
rs9658171 Chr.6: 35426821-35426821 on Build GRCh38
rs200943709 Chr.6: 35426663-35426663 on Build GRCh38
rs199855568 Chr.6: 35426609-35426609 on Build GRCh38
rs201418104 Chr.6: 35426871-35426871 on Build GRCh38
rs9658169 Chr.6: 35426712-35426712 on Build GRCh38
rs200623190 Chr.6: 35426646-35426646 on Build GRCh38
rs74806287 Chr.6: 35426666-35426666 on Build GRCh38
rs1015347896 Chr.6: 35426736-35426736 on Build GRCh38
rs200496552 Chr.6: 35426866-35426866 on Build GRCh38
rs577059360 Chr.6: 35426620-35426620 on Build GRCh38
rs536316020 Chr.6: 35426527-35426527 on Build GRCh38
rs199616216 Chr.6: 35426917-35426917 on Build GRCh38
rs202166822 Chr.6: 35426865-35426865 on Build GRCh38
rs759893973 Chr.6: 35426887-35426887 on Build GRCh38
rs200368320 Chr.6: 35426893-35426893 on Build GRCh38
rs902689582 Chr.6: 35426657-35426657 on Build GRCh38
rs199549196 Chr.6: 35426880-35426880 on Build GRCh38
rs1035287087 Chr.6: 35426672-35426672 on Build GRCh38
rs574509586 Chr.6: 35426677-35426677 on Build GRCh38
rs1004930228 Chr.6: 35426929-35426929 on Build GRCh38
rs922551369 Chr.6: 35426793-35426793 on Build GRCh38
rs910941351 Chr.6: 35426892-35426892 on Build GRCh38
rs200113596 Chr.6: 35426818-35426818 on Build GRCh38
rs534926010 Chr.6: 35426655-35426655 on Build GRCh38
rs45621342 Chr.6: 35426961-35426961 on Build GRCh38
rs200422292 Chr.6: 35426575-35426575 on Build GRCh38
rs201455626 Chr.6: 35426650-35426650 on Build GRCh38
rs946452434 Chr.6: 35426835-35426835 on Build GRCh38
rs9658173 Chr.6: 35426894-35426894 on Build GRCh38
rs201962025 Chr.6: 35426556-35426556 on Build GRCh38
rs201073235 Chr.6: 35426756-35426756 on Build GRCh38
rs901072727 Chr.6: 35426689-35426689 on Build GRCh38
rs191172020 Chr.6: 35426531-35426531 on Build GRCh38
rs960209626 Chr.6: 35426612-35426612 on Build GRCh38
rs1040952319 Chr.6: 35426682-35426682 on Build GRCh38
rs980354286 Chr.6: 35426580-35426580 on Build GRCh38
rs960302536 Chr.6: 35426851-35426851 on Build GRCh38
rs148098583 Chr.6: 35426819-35426819 on Build GRCh38
rs200717927 Chr.6: 35426924-35426924 on Build GRCh38
rs9658172 Chr.6: 35426833-35426833 on Build GRCh38
rs887784260 Chr.6: 35426926-35426926 on Build GRCh38
rs200421322 Chr.6: 35426600-35426600 on Build GRCh38
rs199537437 Chr.6: 35426662-35426662 on Build GRCh38
rs988972559 Chr.6: 35426615-35426615 on Build GRCh38
rs201656538 Chr.6: 35426760-35426760 on Build GRCh38
rs199574588 Chr.6: 35426590-35426590 on Build GRCh38
rs201468320 Chr.6: 35426605-35426605 on Build GRCh38
rs747421811 Chr.6: 35426603-35426603 on Build GRCh38
rs201475851 Chr.6: 35426934-35426934 on Build GRCh38
rs1016724493 Chr.6: 35426761-35426761 on Build GRCh38
rs758132321 Chr.6: 35426888-35426888 on Build GRCh38
rs1006347459 Chr.6: 35426728-35426728 on Build GRCh38
rs558516477 Chr.6: 35426619-35426619 on Build GRCh38
rs17847856 Chr.6: 35426832-35426832 on Build GRCh38
rs929941974 Chr.6: 35426953-35426953 on Build GRCh38

View Full Product Details