Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36301749-36302253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA1
Entrez Gene ID
285848
Gene Name
patatin like phospholipase domain containing 1
Gene Aliases
ARCI10, dJ50J22.1
UniGene
Hs.407002
Gene Chromosome Location
Chr.6: 36242523-36313955 on Build GRCh38
Transcripts
NM_001145716.2 NM_001145717.1 NM_173676.2 XM_011514519.2 XM_011514520.2 XM_017010776.1 XM_017010777.1 XM_017010778.1 XM_017010779.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079181 c.1142C>T p.P381L Chr.6: 36302227-36302227 on Build GRCh38
7115998 c.878G>A p.R293H Chr.6: 36301960-36301960 on Build GRCh38
6893063 c.953G>A p.G318E Chr.6: 36302038-36302038 on Build GRCh38
3697410 c.898C>T p.R300W Chr.6: 36301983-36301983 on Build GRCh38
7079183 c.1145C>T p.P382L Chr.6: 36302227-36302227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766001985 Chr.6: 36302148-36302148 on Build GRCh38
rs760594841 Chr.6: 36301986-36301990 on Build GRCh38
rs376611583 Chr.6: 36302033-36302033 on Build GRCh38
rs201804225 Chr.6: 36302054-36302054 on Build GRCh38
rs376104376 Chr.6: 36301812-36301812 on Build GRCh38
rs143843185 Chr.6: 36302038-36302038 on Build GRCh38
rs561407491 Chr.6: 36302173-36302173 on Build GRCh38
rs886061375 Chr.6: 36302047-36302047 on Build GRCh38
rs536510288 Chr.6: 36302001-36302001 on Build GRCh38
rs774988230 Chr.6: 36302045-36302045 on Build GRCh38
rs761075511 Chr.6: 36302147-36302147 on Build GRCh38
rs55788625 Chr.6: 36301799-36301799 on Build GRCh38
rs369897469 Chr.6: 36302083-36302083 on Build GRCh38
rs201231660 Chr.6: 36301898-36301898 on Build GRCh38
rs367595414 Chr.6: 36301823-36301823 on Build GRCh38
rs78257597 Chr.6: 36302070-36302070 on Build GRCh38
rs138035213 Chr.6: 36302109-36302109 on Build GRCh38
rs373648600 Chr.6: 36302099-36302099 on Build GRCh38
rs751140258 Chr.6: 36301935-36301935 on Build GRCh38
rs763059905 Chr.6: 36302039-36302039 on Build GRCh38
rs376134805 Chr.6: 36302199-36302199 on Build GRCh38
rs368719584 Chr.6: 36302062-36302062 on Build GRCh38
rs778158406 Chr.6: 36301977-36301977 on Build GRCh38
rs55882160 Chr.6: 36301798-36301798 on Build GRCh38
rs146664996 Chr.6: 36302089-36302089 on Build GRCh38
rs934207245 Chr.6: 36301806-36301806 on Build GRCh38
rs762355904 Chr.6: 36302121-36302121 on Build GRCh38
rs1013220255 Chr.6: 36301910-36301910 on Build GRCh38
rs763319302 Chr.6: 36301972-36301972 on Build GRCh38
rs868165790 Chr.6: 36302167-36302167 on Build GRCh38
rs778797684 Chr.6: 36302000-36302000 on Build GRCh38
rs780963967 Chr.6: 36301905-36301905 on Build GRCh38
rs773246334 Chr.6: 36302007-36302008 on Build GRCh38
rs376807751 Chr.6: 36302082-36302082 on Build GRCh38
rs760708335 Chr.6: 36302064-36302075 on Build GRCh38
rs984725679 Chr.6: 36302013-36302013 on Build GRCh38
rs918735179 Chr.6: 36301792-36301792 on Build GRCh38
rs754166062 Chr.6: 36301983-36301983 on Build GRCh38
rs986868922 Chr.6: 36302162-36302162 on Build GRCh38
rs148922973 Chr.6: 36302025-36302025 on Build GRCh38
rs754910405 Chr.6: 36301991-36301991 on Build GRCh38
rs753139564 Chr.6: 36301868-36301868 on Build GRCh38
rs769951055 Chr.6: 36302087-36302087 on Build GRCh38
rs937830523 Chr.6: 36302011-36302011 on Build GRCh38
rs752563994 Chr.6: 36302077-36302077 on Build GRCh38
rs571759549 Chr.6: 36301934-36301934 on Build GRCh38
rs202194175 Chr.6: 36302158-36302158 on Build GRCh38
rs34598813 Chr.6: 36301948-36301948 on Build GRCh38
rs199756414 Chr.6: 36301973-36301973 on Build GRCh38
rs375612206 Chr.6: 36301856-36301856 on Build GRCh38
rs746018691 Chr.6: 36302031-36302031 on Build GRCh38
rs776728407 Chr.6: 36301839-36301839 on Build GRCh38
rs764308393 Chr.6: 36301885-36301885 on Build GRCh38
rs146577333 Chr.6: 36301946-36301946 on Build GRCh38
rs768854376 Chr.6: 36302102-36302102 on Build GRCh38
rs779379285 Chr.6: 36301987-36301987 on Build GRCh38
rs986801659 Chr.6: 36301978-36301978 on Build GRCh38
rs532084398 Chr.6: 36302193-36302193 on Build GRCh38
rs370317617 Chr.6: 36301821-36301821 on Build GRCh38
rs182227800 Chr.6: 36302040-36302040 on Build GRCh38
rs1013356400 Chr.6: 36302229-36302229 on Build GRCh38
rs762339323 Chr.6: 36302050-36302050 on Build GRCh38
rs769812955 Chr.6: 36301810-36301810 on Build GRCh38
rs113967622 Chr.6: 36301979-36301979 on Build GRCh38
rs771191324 Chr.6: 36301951-36301951 on Build GRCh38
rs975551743 Chr.6: 36301963-36301963 on Build GRCh38
rs776341023 Chr.6: 36302150-36302150 on Build GRCh38
rs765828334 Chr.6: 36301862-36301862 on Build GRCh38
rs778887479 Chr.6: 36302074-36302074 on Build GRCh38
rs898315427 Chr.6: 36301845-36301845 on Build GRCh38
rs544026479 Chr.6: 36302194-36302194 on Build GRCh38
rs781396380 Chr.6: 36302020-36302020 on Build GRCh38
rs765162711 Chr.6: 36301961-36301961 on Build GRCh38
rs373763742 Chr.6: 36301822-36301822 on Build GRCh38
rs761325452 Chr.6: 36301838-36301838 on Build GRCh38
rs372573034 Chr.6: 36301906-36301906 on Build GRCh38
rs148164327 Chr.6: 36302068-36302068 on Build GRCh38
rs773486191 Chr.6: 36302145-36302145 on Build GRCh38
rs750100853 Chr.6: 36301984-36301984 on Build GRCh38
rs143487095 Chr.6: 36302151-36302151 on Build GRCh38
rs758291531 Chr.6: 36302172-36302172 on Build GRCh38
rs531791387 Chr.6: 36301872-36301872 on Build GRCh38
rs975899270 Chr.6: 36301782-36301782 on Build GRCh38
rs759728136 Chr.6: 36301960-36301960 on Build GRCh38
rs571808408 Chr.6: 36302103-36302103 on Build GRCh38
rs112858348 Chr.6: 36301950-36301950 on Build GRCh38
rs752011907 Chr.6: 36301974-36301974 on Build GRCh38
rs146796609 Chr.6: 36301989-36301989 on Build GRCh38
rs908508975 Chr.6: 36301980-36301980 on Build GRCh38
rs908477731 Chr.6: 36301815-36301815 on Build GRCh38
rs143604894 Chr.6: 36301985-36301985 on Build GRCh38
rs751279110 Chr.6: 36301897-36301897 on Build GRCh38
rs893355470 Chr.6: 36302088-36302088 on Build GRCh38
rs145856102 Chr.6: 36302169-36302169 on Build GRCh38
rs776079082 Chr.6: 36301965-36301965 on Build GRCh38
rs200207071 Chr.6: 36302123-36302123 on Build GRCh38
rs776818332 Chr.6: 36301959-36301959 on Build GRCh38
rs749463513 Chr.6: 36302195-36302195 on Build GRCh38
rs199548644 Chr.6: 36302019-36302019 on Build GRCh38
rs1038214508 Chr.6: 36301836-36301836 on Build GRCh38
rs775006894 Chr.6: 36302111-36302111 on Build GRCh38
rs10947600 Chr.6: 36301826-36301826 on Build GRCh38
rs77787922 Chr.6: 36301857-36301857 on Build GRCh38
rs371888522 Chr.6: 36301930-36301930 on Build GRCh38
rs372538424 Chr.6: 36302063-36302063 on Build GRCh38
rs867297816 Chr.6: 36302022-36302022 on Build GRCh38
rs750119342 Chr.6: 36302060-36302060 on Build GRCh38

View Full Product Details