Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27870967-27871475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARA5
Entrez Gene ID
286133
Gene Name
scavenger receptor class A member 5
Gene Aliases
NET33, Tesr
UniGene
Hs.591833
Gene Chromosome Location
Chr.8: 27869882-27992852 on Build GRCh38
Transcripts
NM_173833.5

SNP Detail

SNP ID Chromosome Location
rs1030734150 Chr.8: 27871367-27871367 on Build GRCh38
rs896168606 Chr.8: 27871250-27871250 on Build GRCh38
rs991209279 Chr.8: 27871209-27871209 on Build GRCh38
rs78416608 Chr.8: 27871333-27871333 on Build GRCh38
rs4732782 Chr.8: 27871384-27871384 on Build GRCh38
rs954757074 Chr.8: 27871422-27871422 on Build GRCh38
rs71519657 Chr.8: 27871200-27871200 on Build GRCh38
rs917986384 Chr.8: 27871031-27871031 on Build GRCh38
rs116655504 Chr.8: 27871313-27871313 on Build GRCh38
rs968612686 Chr.8: 27871171-27871171 on Build GRCh38
rs534927888 Chr.8: 27871371-27871371 on Build GRCh38
rs1013900632 Chr.8: 27871280-27871280 on Build GRCh38
rs187682982 Chr.8: 27871073-27871073 on Build GRCh38
rs923315014 Chr.8: 27871126-27871126 on Build GRCh38
rs972905232 Chr.8: 27871239-27871239 on Build GRCh38
rs918024450 Chr.8: 27871228-27871228 on Build GRCh38
rs761802781 Chr.8: 27871151-27871151 on Build GRCh38
rs890023800 Chr.8: 27871352-27871352 on Build GRCh38
rs931519831 Chr.8: 27871038-27871038 on Build GRCh38
rs1016335200 Chr.8: 27871114-27871114 on Build GRCh38
rs774085392 Chr.8: 27871128-27871128 on Build GRCh38
rs996288772 Chr.8: 27871364-27871364 on Build GRCh38
rs989106631 Chr.8: 27871135-27871135 on Build GRCh38
rs964741782 Chr.8: 27871363-27871363 on Build GRCh38
rs760502239 Chr.8: 27871285-27871285 on Build GRCh38
rs989466638 Chr.8: 27871424-27871424 on Build GRCh38
rs753557108 Chr.8: 27871074-27871074 on Build GRCh38
rs1006258140 Chr.8: 27871084-27871084 on Build GRCh38
rs144997388 Chr.8: 27871166-27871166 on Build GRCh38
rs149921029 Chr.8: 27871036-27871036 on Build GRCh38
rs528803301 Chr.8: 27871035-27871035 on Build GRCh38
rs924992002 Chr.8: 27871206-27871206 on Build GRCh38
rs533763299 Chr.8: 27871238-27871238 on Build GRCh38
rs113666475 Chr.8: 27870991-27870991 on Build GRCh38
rs990959433 Chr.8: 27871028-27871028 on Build GRCh38
rs1001905255 Chr.8: 27870999-27870999 on Build GRCh38
rs938096970 Chr.8: 27871000-27871000 on Build GRCh38
rs1010040807 Chr.8: 27871154-27871154 on Build GRCh38
rs526301 Chr.8: 27871080-27871080 on Build GRCh38
rs913373409 Chr.8: 27871156-27871156 on Build GRCh38
rs955100401 Chr.8: 27871132-27871132 on Build GRCh38
rs78889390 Chr.8: 27871443-27871443 on Build GRCh38
rs937747994 Chr.8: 27871229-27871229 on Build GRCh38
rs149124800 Chr.8: 27871359-27871359 on Build GRCh38
rs1034676956 Chr.8: 27871202-27871202 on Build GRCh38
rs768223603 Chr.8: 27871117-27871117 on Build GRCh38
rs964772384 Chr.8: 27871115-27871115 on Build GRCh38
rs567775227 Chr.8: 27871215-27871215 on Build GRCh38
rs1026551808 Chr.8: 27871079-27871079 on Build GRCh38
rs949420098 Chr.8: 27871235-27871235 on Build GRCh38
rs950574303 Chr.8: 27871083-27871083 on Build GRCh38
rs889501435 Chr.8: 27871051-27871051 on Build GRCh38
rs1048539805 Chr.8: 27871045-27871045 on Build GRCh38
rs1047676651 Chr.8: 27871286-27871286 on Build GRCh38

View Full Product Details