Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28353032-28353556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF395
Entrez Gene ID
55893
Gene Name
zinc finger protein 395
Gene Aliases
HDBP-2, HDBP2, HDRF-2, PBF, PRF-1, PRF1, Si-1-8-14
UniGene
Hs.435535
Gene Chromosome Location
Chr.8: 28345585-28386460 on Build GRCh38
Transcripts
NM_018660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1737247 c.819+4C>T p.? Chr.8: 28353169-28353169 on Build GRCh38
2154686 c.754G>A p.D252N Chr.8: 28353238-28353238 on Build GRCh38
3899396 c.638G>A p.G213D Chr.8: 28353354-28353354 on Build GRCh38
184586 c.700delC p.H234fs*43 Chr.8: 28353292-28353292 on Build GRCh38
6994458 c.587C>T p.S196F Chr.8: 28353405-28353405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752312111 Chr.8: 28353235-28353235 on Build GRCh38
rs563932326 Chr.8: 28353203-28353203 on Build GRCh38
rs557476742 Chr.8: 28353080-28353080 on Build GRCh38
rs962691432 Chr.8: 28353427-28353427 on Build GRCh38
rs527253158 Chr.8: 28353114-28353114 on Build GRCh38
rs549592223 Chr.8: 28353322-28353322 on Build GRCh38
rs777362321 Chr.8: 28353434-28353434 on Build GRCh38
rs766339235 Chr.8: 28353129-28353129 on Build GRCh38
rs776867373 Chr.8: 28353312-28353312 on Build GRCh38
rs778547157 Chr.8: 28353292-28353292 on Build GRCh38
rs201787778 Chr.8: 28353137-28353137 on Build GRCh38
rs891061405 Chr.8: 28353510-28353510 on Build GRCh38
rs925648511 Chr.8: 28353515-28353515 on Build GRCh38
rs932609238 Chr.8: 28353181-28353181 on Build GRCh38
rs766187939 Chr.8: 28353205-28353205 on Build GRCh38
rs973198643 Chr.8: 28353057-28353057 on Build GRCh38
rs559344073 Chr.8: 28353107-28353107 on Build GRCh38
rs528992700 Chr.8: 28353362-28353362 on Build GRCh38
rs774832287 Chr.8: 28353349-28353349 on Build GRCh38
rs973789881 Chr.8: 28353430-28353430 on Build GRCh38
rs779513576 Chr.8: 28353162-28353162 on Build GRCh38
rs1008042153 Chr.8: 28353246-28353246 on Build GRCh38
rs548968299 Chr.8: 28353363-28353363 on Build GRCh38
rs757654362 Chr.8: 28353277-28353277 on Build GRCh38
rs200322543 Chr.8: 28353437-28353437 on Build GRCh38
rs371451550 Chr.8: 28353125-28353125 on Build GRCh38
rs571744613 Chr.8: 28353403-28353403 on Build GRCh38
rs766882746 Chr.8: 28353221-28353221 on Build GRCh38
rs755719000 Chr.8: 28353245-28353245 on Build GRCh38
rs200668886 Chr.8: 28353381-28353381 on Build GRCh38
rs887332515 Chr.8: 28353393-28353393 on Build GRCh38
rs145352684 Chr.8: 28353292-28353292 on Build GRCh38
rs921792273 Chr.8: 28353242-28353242 on Build GRCh38
rs747148637 Chr.8: 28353302-28353302 on Build GRCh38
rs766650844 Chr.8: 28353253-28353253 on Build GRCh38
rs758797339 Chr.8: 28353392-28353392 on Build GRCh38
rs987289665 Chr.8: 28353448-28353448 on Build GRCh38
rs774128952 Chr.8: 28353443-28353443 on Build GRCh38
rs759478170 Chr.8: 28353216-28353216 on Build GRCh38
rs754232698 Chr.8: 28353272-28353272 on Build GRCh38
rs756695317 Chr.8: 28353444-28353444 on Build GRCh38
rs926823646 Chr.8: 28353090-28353090 on Build GRCh38
rs868387201 Chr.8: 28353287-28353288 on Build GRCh38
rs746664260 Chr.8: 28353438-28353438 on Build GRCh38
rs370835109 Chr.8: 28353151-28353151 on Build GRCh38
rs977288864 Chr.8: 28353503-28353503 on Build GRCh38
rs772210875 Chr.8: 28353169-28353169 on Build GRCh38
rs777988297 Chr.8: 28353147-28353147 on Build GRCh38
rs991235667 Chr.8: 28353533-28353533 on Build GRCh38
rs148606352 Chr.8: 28353399-28353399 on Build GRCh38
rs768735105 Chr.8: 28353311-28353311 on Build GRCh38
rs552142076 Chr.8: 28353402-28353402 on Build GRCh38
rs142368525 Chr.8: 28353350-28353350 on Build GRCh38
rs987833909 Chr.8: 28353290-28353290 on Build GRCh38
rs768986191 Chr.8: 28353196-28353196 on Build GRCh38
rs951501152 Chr.8: 28353184-28353184 on Build GRCh38
rs967298888 Chr.8: 28353494-28353494 on Build GRCh38
rs775651845 Chr.8: 28353183-28353183 on Build GRCh38
rs1014947714 Chr.8: 28353157-28353157 on Build GRCh38
rs371302969 Chr.8: 28353152-28353152 on Build GRCh38
rs137923010 Chr.8: 28353341-28353341 on Build GRCh38
rs780811598 Chr.8: 28353475-28353475 on Build GRCh38
rs368497423 Chr.8: 28353188-28353188 on Build GRCh38
rs747627304 Chr.8: 28353413-28353413 on Build GRCh38
rs943925547 Chr.8: 28353517-28353517 on Build GRCh38
rs375975466 Chr.8: 28353134-28353134 on Build GRCh38
rs777522426 Chr.8: 28353250-28353250 on Build GRCh38
rs770648077 Chr.8: 28353327-28353327 on Build GRCh38
rs756707002 Chr.8: 28353197-28353197 on Build GRCh38
rs763543727 Chr.8: 28353355-28353355 on Build GRCh38
rs755806363 Chr.8: 28353145-28353145 on Build GRCh38
rs780656102 Chr.8: 28353398-28353398 on Build GRCh38
rs768191349 Chr.8: 28353291-28353292 on Build GRCh38
rs560842903 Chr.8: 28353102-28353102 on Build GRCh38
rs746407530 Chr.8: 28353168-28353168 on Build GRCh38
rs911813169 Chr.8: 28353458-28353458 on Build GRCh38
rs1014419156 Chr.8: 28353387-28353387 on Build GRCh38
rs779991389 Chr.8: 28353298-28353298 on Build GRCh38
rs751463118 Chr.8: 28353211-28353211 on Build GRCh38
rs1022309228 Chr.8: 28353297-28353297 on Build GRCh38
rs771882762 Chr.8: 28353447-28353447 on Build GRCh38
rs6558057 Chr.8: 28353096-28353096 on Build GRCh38
rs370376491 Chr.8: 28353133-28353133 on Build GRCh38
rs752227626 Chr.8: 28353141-28353141 on Build GRCh38
rs945732329 Chr.8: 28353109-28353109 on Build GRCh38
rs565681323 Chr.8: 28353391-28353391 on Build GRCh38
rs939776377 Chr.8: 28353418-28353418 on Build GRCh38
rs573928317 Chr.8: 28353084-28353084 on Build GRCh38
rs73570782 Chr.8: 28353356-28353356 on Build GRCh38
rs763807352 Chr.8: 28353098-28353098 on Build GRCh38
rs543541450 Chr.8: 28353064-28353064 on Build GRCh38
rs774149075 Chr.8: 28353333-28353333 on Build GRCh38
rs745788207 Chr.8: 28353529-28353529 on Build GRCh38
rs779423873 Chr.8: 28353287-28353287 on Build GRCh38
rs759457939 Chr.8: 28353338-28353338 on Build GRCh38
rs775449786 Chr.8: 28353454-28353454 on Build GRCh38

View Full Product Details