Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72592762-72593297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKA1
Entrez Gene ID
5255
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Gene Aliases
PHKA
UniGene
Hs.201379
Gene Chromosome Location
Chr.X: 72578814-72714181 on Build GRCh38
Transcripts
NM_001122670.1 NM_001172436.1 NM_002637.3 XM_006724661.2

SNP Detail

SNP ID Chromosome Location
rs200283229 Chr.X: 72593205-72593205 on Build GRCh38
rs781947160 Chr.X: 72593070-72593070 on Build GRCh38
rs151120187 Chr.X: 72593204-72593204 on Build GRCh38
rs782038473 Chr.X: 72593271-72593271 on Build GRCh38
rs373128694 Chr.X: 72593097-72593097 on Build GRCh38
rs903271211 Chr.X: 72593095-72593095 on Build GRCh38
rs140225187 Chr.X: 72593227-72593227 on Build GRCh38
rs377499516 Chr.X: 72593103-72593103 on Build GRCh38
rs1038924115 Chr.X: 72592887-72592887 on Build GRCh38
rs782022487 Chr.X: 72593143-72593143 on Build GRCh38
rs782073125 Chr.X: 72593266-72593266 on Build GRCh38
rs782183196 Chr.X: 72593192-72593192 on Build GRCh38
rs782591584 Chr.X: 72592952-72592952 on Build GRCh38
rs781856424 Chr.X: 72593178-72593178 on Build GRCh38
rs899845193 Chr.X: 72592908-72592908 on Build GRCh38
rs782453258 Chr.X: 72593198-72593198 on Build GRCh38
rs201554965 Chr.X: 72593075-72593075 on Build GRCh38
rs1011496648 Chr.X: 72592967-72592967 on Build GRCh38
rs782104707 Chr.X: 72593059-72593059 on Build GRCh38
rs782706413 Chr.X: 72593122-72593122 on Build GRCh38
rs782653573 Chr.X: 72593116-72593116 on Build GRCh38
rs781924822 Chr.X: 72593262-72593262 on Build GRCh38
rs781958344 Chr.X: 72593083-72593083 on Build GRCh38
rs782623805 Chr.X: 72593184-72593184 on Build GRCh38
rs782204354 Chr.X: 72593257-72593257 on Build GRCh38
rs782514332 Chr.X: 72593181-72593181 on Build GRCh38
rs145272281 Chr.X: 72593267-72593267 on Build GRCh38
rs782709783 Chr.X: 72593065-72593066 on Build GRCh38
rs370590996 Chr.X: 72593158-72593158 on Build GRCh38
rs192812741 Chr.X: 72592824-72592824 on Build GRCh38
rs782374250 Chr.X: 72593220-72593220 on Build GRCh38
rs184743934 Chr.X: 72593177-72593177 on Build GRCh38
rs781811981 Chr.X: 72593137-72593137 on Build GRCh38
rs782360415 Chr.X: 72593259-72593259 on Build GRCh38
rs373895720 Chr.X: 72593185-72593185 on Build GRCh38

View Full Product Details