Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78124788-78125282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGK1
Entrez Gene ID
5230
Gene Name
phosphoglycerate kinase 1
Gene Aliases
HEL-S-68p, MIG10, PGKA
UniGene
Hs.78771
Gene Chromosome Location
Chr.X: 78104169-78126827 on Build GRCh38
Transcripts
NM_000291.3

SNP Detail

SNP ID Chromosome Location
rs782701299 Chr.X: 78125144-78125144 on Build GRCh38
rs372350486 Chr.X: 78125065-78125065 on Build GRCh38
rs890279411 Chr.X: 78124999-78124999 on Build GRCh38
rs782248660 Chr.X: 78124845-78124845 on Build GRCh38
rs782347543 Chr.X: 78125064-78125064 on Build GRCh38
rs1057234314 Chr.X: 78124846-78124846 on Build GRCh38
rs915956435 Chr.X: 78124916-78124916 on Build GRCh38
rs782290996 Chr.X: 78124861-78124861 on Build GRCh38
rs1047587122 Chr.X: 78124990-78124990 on Build GRCh38
rs137852537 Chr.X: 78124896-78124896 on Build GRCh38
rs992960131 Chr.X: 78125215-78125215 on Build GRCh38
rs137852530 Chr.X: 78124992-78124992 on Build GRCh38
rs371615868 Chr.X: 78124981-78124981 on Build GRCh38
rs782030143 Chr.X: 78125066-78125066 on Build GRCh38
rs369320831 Chr.X: 78125000-78125000 on Build GRCh38
rs183087139 Chr.X: 78124985-78124985 on Build GRCh38
rs781922586 Chr.X: 78124934-78124934 on Build GRCh38
rs782574227 Chr.X: 78125002-78125002 on Build GRCh38
rs960196613 Chr.X: 78125186-78125186 on Build GRCh38
rs916012043 Chr.X: 78125224-78125224 on Build GRCh38
rs782762254 Chr.X: 78124987-78124987 on Build GRCh38
rs781813870 Chr.X: 78124823-78124823 on Build GRCh38
rs782512699 Chr.X: 78124986-78124986 on Build GRCh38
rs782136810 Chr.X: 78125057-78125057 on Build GRCh38
rs137852533 Chr.X: 78124883-78124883 on Build GRCh38
rs964399706 Chr.X: 78125118-78125118 on Build GRCh38
rs1008641570 Chr.X: 78125050-78125050 on Build GRCh38
rs782819714 Chr.X: 78124925-78124925 on Build GRCh38
rs17303413 Chr.X: 78124810-78124810 on Build GRCh38
rs782388400 Chr.X: 78124851-78124851 on Build GRCh38
rs782424374 Chr.X: 78124876-78124876 on Build GRCh38
rs782144480 Chr.X: 78124922-78124922 on Build GRCh38
rs727504084 Chr.X: 78125051-78125051 on Build GRCh38
rs782272102 Chr.X: 78124865-78124865 on Build GRCh38
rs782797917 Chr.X: 78125042-78125042 on Build GRCh38
rs782633105 Chr.X: 78124835-78124835 on Build GRCh38
rs782173677 Chr.X: 78125083-78125083 on Build GRCh38
rs782471954 Chr.X: 78125029-78125029 on Build GRCh38
rs782201994 Chr.X: 78125010-78125010 on Build GRCh38
rs150956842 Chr.X: 78125033-78125033 on Build GRCh38
rs782566461 Chr.X: 78125128-78125128 on Build GRCh38
rs367749559 Chr.X: 78124866-78124866 on Build GRCh38
rs782576397 Chr.X: 78124854-78124855 on Build GRCh38
rs782303151 Chr.X: 78124821-78124821 on Build GRCh38
rs782072317 Chr.X: 78124963-78124963 on Build GRCh38
rs781974818 Chr.X: 78125185-78125185 on Build GRCh38
rs924810345 Chr.X: 78125155-78125155 on Build GRCh38
rs782002575 Chr.X: 78124915-78124915 on Build GRCh38
rs782652022 Chr.X: 78124854-78124854 on Build GRCh38
rs782479790 Chr.X: 78124824-78124824 on Build GRCh38
rs1010588426 Chr.X: 78125126-78125126 on Build GRCh38
rs782358944 Chr.X: 78124889-78124889 on Build GRCh38
rs781889755 Chr.X: 78125127-78125127 on Build GRCh38
rs1017311455 Chr.X: 78125055-78125055 on Build GRCh38

View Full Product Details