Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 85306089-85306601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
UniGene
Hs.267038
Gene Chromosome Location
Chr.X: 85277389-85379743 on Build GRCh38
Transcripts
NM_001307940.1 NM_024921.3 XM_005262203.3

SNP Detail

SNP ID Chromosome Location
rs753557062 Chr.X: 85306167-85306167 on Build GRCh38
rs899139281 Chr.X: 85306434-85306434 on Build GRCh38
rs1028905013 Chr.X: 85306457-85306457 on Build GRCh38
rs770250275 Chr.X: 85306300-85306300 on Build GRCh38
rs756579325 Chr.X: 85306498-85306498 on Build GRCh38
rs375249982 Chr.X: 85306363-85306363 on Build GRCh38
rs771883809 Chr.X: 85306361-85306361 on Build GRCh38
rs747737798 Chr.X: 85306352-85306352 on Build GRCh38
rs989610455 Chr.X: 85306535-85306535 on Build GRCh38
rs766200497 Chr.X: 85306380-85306380 on Build GRCh38
rs943624237 Chr.X: 85306444-85306444 on Build GRCh38
rs1057271614 Chr.X: 85306234-85306234 on Build GRCh38
rs868215608 Chr.X: 85306282-85306282 on Build GRCh38
rs746932126 Chr.X: 85306126-85306126 on Build GRCh38
rs766362960 Chr.X: 85306150-85306150 on Build GRCh38
rs12012683 Chr.X: 85306522-85306522 on Build GRCh38
rs757679160 Chr.X: 85306241-85306241 on Build GRCh38
rs144515909 Chr.X: 85306326-85306326 on Build GRCh38
rs200080276 Chr.X: 85306218-85306218 on Build GRCh38
rs760282121 Chr.X: 85306365-85306365 on Build GRCh38
rs139385491 Chr.X: 85306198-85306198 on Build GRCh38
rs754846192 Chr.X: 85306171-85306171 on Build GRCh38
rs554164450 Chr.X: 85306256-85306256 on Build GRCh38
rs914275381 Chr.X: 85306428-85306428 on Build GRCh38
rs752977075 Chr.X: 85306176-85306176 on Build GRCh38
rs773361558 Chr.X: 85306499-85306499 on Build GRCh38
rs901993495 Chr.X: 85306493-85306493 on Build GRCh38
rs363780 Chr.X: 85306459-85306459 on Build GRCh38
rs111797102 Chr.X: 85306310-85306310 on Build GRCh38
rs751799715 Chr.X: 85306442-85306442 on Build GRCh38
rs779002542 Chr.X: 85306175-85306175 on Build GRCh38
rs776596654 Chr.X: 85306296-85306296 on Build GRCh38
rs531173335 Chr.X: 85306320-85306320 on Build GRCh38
rs781742296 Chr.X: 85306273-85306273 on Build GRCh38
rs746212940 Chr.X: 85306299-85306299 on Build GRCh38
rs766415515 Chr.X: 85306147-85306148 on Build GRCh38
rs141327405 Chr.X: 85306388-85306388 on Build GRCh38
rs758621935 Chr.X: 85306189-85306189 on Build GRCh38
rs750328897 Chr.X: 85306217-85306217 on Build GRCh38

View Full Product Details