Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100593803-100594300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNMD
Entrez Gene ID
64102
Gene Name
tenomodulin
Gene Aliases
BRICD4, CHM1L, TEM
UniGene
Hs.132957
Gene Chromosome Location
Chr.X: 100584793-100599885 on Build GRCh38
Transcripts
NM_022144.2

SNP Detail

SNP ID Chromosome Location
rs779320174 Chr.X: 100594257-100594257 on Build GRCh38
rs372659004 Chr.X: 100593933-100593933 on Build GRCh38
rs777845557 Chr.X: 100593916-100593916 on Build GRCh38
rs749419680 Chr.X: 100594244-100594244 on Build GRCh38
rs368264185 Chr.X: 100594256-100594256 on Build GRCh38
rs762845483 Chr.X: 100594036-100594037 on Build GRCh38
rs373430804 Chr.X: 100593851-100593851 on Build GRCh38
rs762108249 Chr.X: 100594035-100594035 on Build GRCh38
rs780972964 Chr.X: 100594211-100594211 on Build GRCh38
rs972248454 Chr.X: 100594036-100594036 on Build GRCh38
rs370268358 Chr.X: 100593899-100593899 on Build GRCh38
rs748625552 Chr.X: 100594259-100594259 on Build GRCh38
rs764913432 Chr.X: 100594053-100594053 on Build GRCh38
rs894918267 Chr.X: 100594099-100594099 on Build GRCh38
rs370445533 Chr.X: 100594058-100594058 on Build GRCh38
rs751466748 Chr.X: 100594220-100594220 on Build GRCh38
rs781289023 Chr.X: 100593963-100593963 on Build GRCh38
rs769866085 Chr.X: 100593982-100593982 on Build GRCh38
rs910076073 Chr.X: 100593922-100593922 on Build GRCh38
rs754481263 Chr.X: 100593861-100593861 on Build GRCh38
rs956053710 Chr.X: 100594216-100594216 on Build GRCh38
rs1011987247 Chr.X: 100594120-100594120 on Build GRCh38
rs762788417 Chr.X: 100594081-100594081 on Build GRCh38
rs999722086 Chr.X: 100593892-100593892 on Build GRCh38
rs767884458 Chr.X: 100594048-100594048 on Build GRCh38
rs773181070 Chr.X: 100594203-100594203 on Build GRCh38
rs764973968 Chr.X: 100594055-100594055 on Build GRCh38
rs780219677 Chr.X: 100594221-100594221 on Build GRCh38
rs199720373 Chr.X: 100593876-100593876 on Build GRCh38
rs769192135 Chr.X: 100594023-100594023 on Build GRCh38
rs746061867 Chr.X: 100593970-100593970 on Build GRCh38
rs776193813 Chr.X: 100593863-100593864 on Build GRCh38
rs34561593 Chr.X: 100594132-100594133 on Build GRCh38
rs2073162 Chr.X: 100594020-100594020 on Build GRCh38
rs747058779 Chr.X: 100593827-100593827 on Build GRCh38
rs774899523 Chr.X: 100594031-100594031 on Build GRCh38
rs971969965 Chr.X: 100594128-100594128 on Build GRCh38
rs768624882 Chr.X: 100593924-100593924 on Build GRCh38
rs1049578127 Chr.X: 100593907-100593907 on Build GRCh38
rs757946626 Chr.X: 100594061-100594061 on Build GRCh38
rs375390954 Chr.X: 100594068-100594068 on Build GRCh38
rs757051341 Chr.X: 100593951-100593951 on Build GRCh38
rs990133762 Chr.X: 100594248-100594248 on Build GRCh38
rs753483163 Chr.X: 100593857-100593857 on Build GRCh38
rs758472019 Chr.X: 100593914-100593914 on Build GRCh38
rs775490194 Chr.X: 100593990-100593990 on Build GRCh38
rs746090412 Chr.X: 100594214-100594214 on Build GRCh38
rs752699389 Chr.X: 100593895-100593895 on Build GRCh38
rs914629865 Chr.X: 100593847-100593847 on Build GRCh38
rs2073163 Chr.X: 100594054-100594054 on Build GRCh38
rs768470160 Chr.X: 100594212-100594212 on Build GRCh38
rs766280893 Chr.X: 100594122-100594122 on Build GRCh38
rs774222415 Chr.X: 100594216-100594218 on Build GRCh38
rs1003128826 Chr.X: 100594189-100594189 on Build GRCh38
rs775126660 Chr.X: 100593932-100593934 on Build GRCh38

View Full Product Details