Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100849912-100850414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOX1
Entrez Gene ID
27035
Gene Name
NADPH oxidase 1
Gene Aliases
GP91-2, MOX1, NOH-1, NOH1
UniGene
Hs.592227
Gene Chromosome Location
Chr.X: 100843324-100874345 on Build GRCh38
Transcripts
NM_001271815.1 NM_007052.4 NM_013955.2 XM_017029407.1

SNP Detail

SNP ID Chromosome Location
rs201130779 Chr.X: 100850278-100850278 on Build GRCh38
rs768290899 Chr.X: 100850341-100850341 on Build GRCh38
rs200198157 Chr.X: 100850217-100850217 on Build GRCh38
rs751521758 Chr.X: 100849978-100849978 on Build GRCh38
rs772571550 Chr.X: 100850164-100850164 on Build GRCh38
rs950297676 Chr.X: 100850290-100850290 on Build GRCh38
rs747275050 Chr.X: 100850342-100850343 on Build GRCh38
rs747350924 Chr.X: 100850169-100850169 on Build GRCh38
rs371695525 Chr.X: 100849981-100849981 on Build GRCh38
rs755237207 Chr.X: 100850271-100850271 on Build GRCh38
rs775730307 Chr.X: 100850191-100850191 on Build GRCh38
rs150382983 Chr.X: 100850177-100850177 on Build GRCh38
rs866136739 Chr.X: 100850242-100850242 on Build GRCh38
rs771457982 Chr.X: 100850360-100850360 on Build GRCh38
rs766334007 Chr.X: 100850387-100850387 on Build GRCh38
rs903175269 Chr.X: 100850050-100850050 on Build GRCh38
rs778002828 Chr.X: 100850168-100850168 on Build GRCh38
rs34688635 Chr.X: 100850206-100850206 on Build GRCh38
rs773768401 Chr.X: 100850343-100850343 on Build GRCh38
rs34220544 Chr.X: 100850061-100850061 on Build GRCh38
rs373968764 Chr.X: 100849948-100849948 on Build GRCh38
rs1054396453 Chr.X: 100850345-100850345 on Build GRCh38
rs916078319 Chr.X: 100850288-100850288 on Build GRCh38
rs756432456 Chr.X: 100850126-100850126 on Build GRCh38
rs780888793 Chr.X: 100850203-100850203 on Build GRCh38
rs36078841 Chr.X: 100850043-100850043 on Build GRCh38
rs759123631 Chr.X: 100849952-100849952 on Build GRCh38
rs1476011 Chr.X: 100850019-100850019 on Build GRCh38
rs1049160662 Chr.X: 100850297-100850297 on Build GRCh38
rs754179284 Chr.X: 100850259-100850259 on Build GRCh38
rs760591404 Chr.X: 100850229-100850229 on Build GRCh38
rs773237228 Chr.X: 100850379-100850379 on Build GRCh38
rs747483659 Chr.X: 100850300-100850300 on Build GRCh38
rs1014322522 Chr.X: 100850008-100850008 on Build GRCh38
rs1044257353 Chr.X: 100850145-100850145 on Build GRCh38
rs762499829 Chr.X: 100849973-100849973 on Build GRCh38
rs372154041 Chr.X: 100850319-100850319 on Build GRCh38
rs777826017 Chr.X: 100850114-100850115 on Build GRCh38
rs748222060 Chr.X: 100849970-100849971 on Build GRCh38
rs775005293 Chr.X: 100849957-100849957 on Build GRCh38
rs369560908 Chr.X: 100850218-100850218 on Build GRCh38
rs2071756 Chr.X: 100850340-100850340 on Build GRCh38
rs778395877 Chr.X: 100850282-100850282 on Build GRCh38
rs973753137 Chr.X: 100850231-100850231 on Build GRCh38
rs35404864 Chr.X: 100850151-100850151 on Build GRCh38
rs886520767 Chr.X: 100850298-100850298 on Build GRCh38
rs917182438 Chr.X: 100850065-100850065 on Build GRCh38
rs748661380 Chr.X: 100850160-100850160 on Build GRCh38
rs1028097649 Chr.X: 100850185-100850185 on Build GRCh38
rs771121983 Chr.X: 100850138-100850138 on Build GRCh38
rs763564875 Chr.X: 100849977-100849977 on Build GRCh38
rs774598153 Chr.X: 100850150-100850150 on Build GRCh38
rs780371283 Chr.X: 100850136-100850136 on Build GRCh38
rs1052764148 Chr.X: 100850055-100850055 on Build GRCh38
rs146366505 Chr.X: 100850349-100850349 on Build GRCh38
rs766241994 Chr.X: 100850236-100850236 on Build GRCh38
rs368118380 Chr.X: 100849956-100849956 on Build GRCh38
rs772431375 Chr.X: 100850168-100850170 on Build GRCh38
rs767232289 Chr.X: 100850111-100850111 on Build GRCh38
rs749428040 Chr.X: 100850119-100850119 on Build GRCh38
rs772831048 Chr.X: 100850227-100850227 on Build GRCh38
rs920819281 Chr.X: 100850268-100850268 on Build GRCh38
rs949968833 Chr.X: 100850115-100850115 on Build GRCh38
rs771265181 Chr.X: 100850176-100850176 on Build GRCh38
rs35468984 Chr.X: 100850380-100850380 on Build GRCh38
rs757686279 Chr.X: 100850315-100850315 on Build GRCh38
rs758409543 Chr.X: 100849947-100849947 on Build GRCh38

View Full Product Details