Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101278578-101279114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF7L
Entrez Gene ID
54457
Gene Name
TATA-box binding protein associated factor 7 like
Gene Aliases
CT40, TAF2Q
UniGene
Hs.223806
Gene Chromosome Location
Chr.X: 101268253-101293071 on Build GRCh38
Transcripts
NM_001168474.1 NM_024885.3 XM_005262152.1 XM_006724664.1

SNP Detail

SNP ID Chromosome Location
rs776676794 Chr.X: 101278927-101278927 on Build GRCh38
rs200724506 Chr.X: 101278788-101278788 on Build GRCh38
rs769980295 Chr.X: 101278969-101278969 on Build GRCh38
rs4256744 Chr.X: 101278644-101278644 on Build GRCh38
rs1030187612 Chr.X: 101278756-101278756 on Build GRCh38
rs747850730 Chr.X: 101278895-101278895 on Build GRCh38
rs1022823574 Chr.X: 101278766-101278766 on Build GRCh38
rs779828419 Chr.X: 101278782-101278782 on Build GRCh38
rs749882927 Chr.X: 101278960-101278960 on Build GRCh38
rs191139490 Chr.X: 101278718-101278718 on Build GRCh38
rs778749822 Chr.X: 101278990-101278990 on Build GRCh38
rs905778062 Chr.X: 101278856-101278856 on Build GRCh38
rs768604863 Chr.X: 101278676-101278676 on Build GRCh38
rs768498072 Chr.X: 101278898-101278898 on Build GRCh38
rs746914198 Chr.X: 101278642-101278642 on Build GRCh38
rs775917566 Chr.X: 101279071-101279071 on Build GRCh38
rs770112787 Chr.X: 101279057-101279057 on Build GRCh38
rs372945691 Chr.X: 101278983-101278983 on Build GRCh38
rs777726028 Chr.X: 101279021-101279021 on Build GRCh38
rs771591889 Chr.X: 101279035-101279035 on Build GRCh38
rs757988636 Chr.X: 101278689-101278689 on Build GRCh38
rs748001398 Chr.X: 101278941-101278941 on Build GRCh38
rs900037065 Chr.X: 101278649-101278649 on Build GRCh38
rs769268408 Chr.X: 101279008-101279008 on Build GRCh38
rs373791048 Chr.X: 101278979-101278979 on Build GRCh38
rs780800971 Chr.X: 101278846-101278846 on Build GRCh38
rs998354929 Chr.X: 101278748-101278748 on Build GRCh38
rs745587413 Chr.X: 101278684-101278684 on Build GRCh38
rs377646158 Chr.X: 101279012-101279012 on Build GRCh38
rs767141710 Chr.X: 101279076-101279076 on Build GRCh38
rs1019121463 Chr.X: 101279043-101279043 on Build GRCh38
rs747032990 Chr.X: 101279025-101279025 on Build GRCh38
rs777318216 Chr.X: 101279044-101279044 on Build GRCh38
rs188012163 Chr.X: 101278628-101278628 on Build GRCh38
rs769061711 Chr.X: 101279040-101279040 on Build GRCh38
rs756059819 Chr.X: 101278962-101278962 on Build GRCh38
rs780187425 Chr.X: 101278607-101278607 on Build GRCh38
rs951477283 Chr.X: 101278858-101278858 on Build GRCh38
rs533472671 Chr.X: 101278906-101278906 on Build GRCh38
rs760096781 Chr.X: 101279052-101279052 on Build GRCh38
rs767005178 Chr.X: 101278959-101278959 on Build GRCh38
rs994787446 Chr.X: 101278891-101278891 on Build GRCh38
rs1016903373 Chr.X: 101278910-101278910 on Build GRCh38
rs761378280 Chr.X: 101279075-101279075 on Build GRCh38

View Full Product Details